Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 25049392-25049888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STXBP6
Entrez Gene ID
29091
Gene Name
syntaxin binding protein 6
Gene Aliases
HSPC156, amisyn
UniGene
Hs.508958
Gene Chromosome Location
Chr.14: 24809654-25050297 on Build GRCh38
Transcripts
NM_001304476.1 NM_014178.7 XM_011536669.2 XM_017021231.1 XM_017021233.1 XM_017021236.1 XM_017021242.1

SNP Detail

SNP ID Chromosome Location
rs933489408 Chr.14: 25049551-25049551 on Build GRCh38
rs904557887 Chr.14: 25049832-25049832 on Build GRCh38
rs557966316 Chr.14: 25049621-25049621 on Build GRCh38
rs535491506 Chr.14: 25049571-25049571 on Build GRCh38
rs1026157887 Chr.14: 25049485-25049485 on Build GRCh38
rs114746317 Chr.14: 25049522-25049522 on Build GRCh38
rs531661727 Chr.14: 25049594-25049594 on Build GRCh38
rs912077400 Chr.14: 25049777-25049777 on Build GRCh38
rs76944633 Chr.14: 25049713-25049713 on Build GRCh38
rs910679086 Chr.14: 25049693-25049693 on Build GRCh38
rs984030134 Chr.14: 25049552-25049552 on Build GRCh38
rs913226785 Chr.14: 25049866-25049866 on Build GRCh38
rs867793947 Chr.14: 25049655-25049655 on Build GRCh38
rs906356577 Chr.14: 25049493-25049493 on Build GRCh38
rs954793004 Chr.14: 25049870-25049870 on Build GRCh38
rs922194324 Chr.14: 25049796-25049796 on Build GRCh38
rs939059928 Chr.14: 25049749-25049749 on Build GRCh38
rs1039293063 Chr.14: 25049576-25049576 on Build GRCh38
rs891880767 Chr.14: 25049644-25049644 on Build GRCh38
rs776058648 Chr.14: 25049564-25049564 on Build GRCh38
rs1003898410 Chr.14: 25049680-25049680 on Build GRCh38
rs1054432980 Chr.14: 25049637-25049637 on Build GRCh38
rs1054461281 Chr.14: 25049863-25049863 on Build GRCh38
rs977516318 Chr.14: 25049663-25049663 on Build GRCh38
rs536788624 Chr.14: 25049686-25049686 on Build GRCh38
rs1010784597 Chr.14: 25049632-25049632 on Build GRCh38
rs1021244315 Chr.14: 25049662-25049662 on Build GRCh38
rs188313068 Chr.14: 25049495-25049495 on Build GRCh38
rs779154243 Chr.14: 25049517-25049517 on Build GRCh38
rs138931318 Chr.14: 25049521-25049521 on Build GRCh38
rs928936067 Chr.14: 25049726-25049726 on Build GRCh38
rs531132830 Chr.14: 25049544-25049544 on Build GRCh38
rs906294395 Chr.14: 25049746-25049746 on Build GRCh38
rs771105811 Chr.14: 25049500-25049500 on Build GRCh38
rs1047508547 Chr.14: 25049604-25049604 on Build GRCh38
rs867240937 Chr.14: 25049628-25049628 on Build GRCh38
rs1031606466 Chr.14: 25049865-25049865 on Build GRCh38
rs929073274 Chr.14: 25049597-25049597 on Build GRCh38
rs987615083 Chr.14: 25049692-25049692 on Build GRCh38
rs982045945 Chr.14: 25049510-25049510 on Build GRCh38
rs1043647791 Chr.14: 25049829-25049829 on Build GRCh38
rs936201205 Chr.14: 25049823-25049823 on Build GRCh38
rs1011275412 Chr.14: 25049810-25049810 on Build GRCh38
rs906387311 Chr.14: 25049608-25049608 on Build GRCh38
rs1022053475 Chr.14: 25049646-25049646 on Build GRCh38
rs180675933 Chr.14: 25049611-25049611 on Build GRCh38
rs969212107 Chr.14: 25049528-25049528 on Build GRCh38
rs551300732 Chr.14: 25049518-25049518 on Build GRCh38
rs954906657 Chr.14: 25049682-25049682 on Build GRCh38
rs1055369621 Chr.14: 25049757-25049757 on Build GRCh38
rs1047541337 Chr.14: 25049735-25049735 on Build GRCh38
rs894945701 Chr.14: 25049781-25049781 on Build GRCh38
rs559563045 Chr.14: 25049786-25049786 on Build GRCh38
rs891864543 Chr.14: 25049462-25049462 on Build GRCh38
rs564747561 Chr.14: 25049650-25049650 on Build GRCh38
rs79012860 Chr.14: 25049559-25049559 on Build GRCh38
rs943608362 Chr.14: 25049569-25049569 on Build GRCh38
rs920362830 Chr.14: 25049544-25049544 on Build GRCh38
rs1033578430 Chr.14: 25049496-25049496 on Build GRCh38
rs998802455 Chr.14: 25049862-25049862 on Build GRCh38
rs895074066 Chr.14: 25049622-25049622 on Build GRCh38
rs974885849 Chr.14: 25049791-25049791 on Build GRCh38
rs868242356 Chr.14: 25049538-25049538 on Build GRCh38
rs1031699829 Chr.14: 25049667-25049667 on Build GRCh38
rs746182949 Chr.14: 25049536-25049536 on Build GRCh38
rs577313917 Chr.14: 25049668-25049668 on Build GRCh38
rs114867359 Chr.14: 25049699-25049699 on Build GRCh38
rs79870238 Chr.14: 25049705-25049705 on Build GRCh38
rs369813043 Chr.14: 25049609-25049609 on Build GRCh38
rs950561466 Chr.14: 25049719-25049719 on Build GRCh38

View Full Product Details