Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 34764914-34765447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAZ1A
Entrez Gene ID
11177
Gene Name
bromodomain adjacent to zinc finger domain 1A
Gene Aliases
ACF1, WALp1, WCRF180, hACF1
UniGene
Hs.509140
Gene Chromosome Location
Chr.14: 34752731-34875647 on Build GRCh38
Transcripts
NM_013448.2 NM_182648.1 XM_011536374.1 XM_011536376.2 XM_017020942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6521433 c.3475G>T p.D1159Y Chr.14: 34765095-34765095 on Build GRCh38
2195878 c.3344G>T p.R1115L Chr.14: 34765226-34765226 on Build GRCh38
6053806 c.3320G>A p.R1107H Chr.14: 34765250-34765250 on Build GRCh38
4791474 c.3319C>T p.R1107C Chr.14: 34765251-34765251 on Build GRCh38
3744313 c.3543G>A p.K1181K Chr.14: 34765027-34765027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765670997 Chr.14: 34765157-34765157 on Build GRCh38
rs2275144 Chr.14: 34765003-34765003 on Build GRCh38
rs779430182 Chr.14: 34765031-34765031 on Build GRCh38
rs757525684 Chr.14: 34765080-34765080 on Build GRCh38
rs747932979 Chr.14: 34765078-34765078 on Build GRCh38
rs140590013 Chr.14: 34765268-34765268 on Build GRCh38
rs773143111 Chr.14: 34765083-34765083 on Build GRCh38
rs189951617 Chr.14: 34765409-34765409 on Build GRCh38
rs17102719 Chr.14: 34765376-34765376 on Build GRCh38
rs746250203 Chr.14: 34765059-34765059 on Build GRCh38
rs757917967 Chr.14: 34765046-34765046 on Build GRCh38
rs896110362 Chr.14: 34765398-34765398 on Build GRCh38
rs1036431139 Chr.14: 34765275-34765275 on Build GRCh38
rs756315491 Chr.14: 34764998-34764998 on Build GRCh38
rs113263332 Chr.14: 34765342-34765342 on Build GRCh38
rs757738575 Chr.14: 34765018-34765018 on Build GRCh38
rs777531640 Chr.14: 34765238-34765238 on Build GRCh38
rs777387877 Chr.14: 34764984-34764985 on Build GRCh38
rs766951291 Chr.14: 34765303-34765303 on Build GRCh38
rs769564606 Chr.14: 34765228-34765228 on Build GRCh38
rs768776081 Chr.14: 34765261-34765261 on Build GRCh38
rs762873715 Chr.14: 34765094-34765094 on Build GRCh38
rs1013754358 Chr.14: 34765334-34765334 on Build GRCh38
rs542047836 Chr.14: 34765096-34765096 on Build GRCh38
rs934848911 Chr.14: 34765375-34765375 on Build GRCh38
rs1057335359 Chr.14: 34765122-34765122 on Build GRCh38
rs752984957 Chr.14: 34764985-34764985 on Build GRCh38
rs761940413 Chr.14: 34765276-34765276 on Build GRCh38
rs751135556 Chr.14: 34765310-34765310 on Build GRCh38
rs560883807 Chr.14: 34765379-34765379 on Build GRCh38
rs988531892 Chr.14: 34764937-34764937 on Build GRCh38
rs764340514 Chr.14: 34765138-34765138 on Build GRCh38
rs934795848 Chr.14: 34765110-34765110 on Build GRCh38
rs779518198 Chr.14: 34765092-34765092 on Build GRCh38
rs544583566 Chr.14: 34765315-34765315 on Build GRCh38
rs760763324 Chr.14: 34765251-34765251 on Build GRCh38
rs779757454 Chr.14: 34765212-34765212 on Build GRCh38
rs759562367 Chr.14: 34764969-34764969 on Build GRCh38
rs780108216 Chr.14: 34765354-34765354 on Build GRCh38
rs754618049 Chr.14: 34765217-34765217 on Build GRCh38
rs564495112 Chr.14: 34765335-34765335 on Build GRCh38
rs765687280 Chr.14: 34765277-34765277 on Build GRCh38
rs924667529 Chr.14: 34765371-34765371 on Build GRCh38
rs767739547 Chr.14: 34764977-34764977 on Build GRCh38
rs751033693 Chr.14: 34765196-34765196 on Build GRCh38
rs752453154 Chr.14: 34765318-34765318 on Build GRCh38
rs377009627 Chr.14: 34765329-34765329 on Build GRCh38
rs779373792 Chr.14: 34765010-34765011 on Build GRCh38
rs200319497 Chr.14: 34765082-34765082 on Build GRCh38
rs750790103 Chr.14: 34765284-34765284 on Build GRCh38
rs755562203 Chr.14: 34764936-34764937 on Build GRCh38
rs745388986 Chr.14: 34765044-34765044 on Build GRCh38
rs771599911 Chr.14: 34765005-34765007 on Build GRCh38
rs746343548 Chr.14: 34765289-34765289 on Build GRCh38
rs758956493 Chr.14: 34765210-34765210 on Build GRCh38
rs779702413 Chr.14: 34765048-34765048 on Build GRCh38
rs921729277 Chr.14: 34765107-34765107 on Build GRCh38
rs749786112 Chr.14: 34764997-34764998 on Build GRCh38
rs1012569503 Chr.14: 34765152-34765152 on Build GRCh38
rs562160763 Chr.14: 34765227-34765227 on Build GRCh38
rs370664036 Chr.14: 34765052-34765052 on Build GRCh38
rs200755247 Chr.14: 34765258-34765258 on Build GRCh38
rs775527075 Chr.14: 34765106-34765106 on Build GRCh38
rs754452249 Chr.14: 34765312-34765312 on Build GRCh38
rs754178887 Chr.14: 34765147-34765147 on Build GRCh38
rs772257354 Chr.14: 34765344-34765344 on Build GRCh38
rs763591475 Chr.14: 34765292-34765292 on Build GRCh38
rs760929467 Chr.14: 34765123-34765123 on Build GRCh38
rs757587766 Chr.14: 34765153-34765153 on Build GRCh38
rs1026452155 Chr.14: 34765252-34765252 on Build GRCh38
rs746714250 Chr.14: 34765060-34765060 on Build GRCh38
rs780846512 Chr.14: 34765226-34765226 on Build GRCh38
rs774324301 Chr.14: 34764968-34764968 on Build GRCh38
rs921384535 Chr.14: 34764947-34764947 on Build GRCh38
rs754200906 Chr.14: 34765016-34765016 on Build GRCh38
rs749273209 Chr.14: 34765240-34765240 on Build GRCh38
rs765188052 Chr.14: 34764948-34764948 on Build GRCh38
rs867399795 Chr.14: 34765158-34765158 on Build GRCh38
rs746483325 Chr.14: 34765216-34765216 on Build GRCh38
rs147149723 Chr.14: 34765250-34765250 on Build GRCh38
rs1018879976 Chr.14: 34765294-34765294 on Build GRCh38
rs768329825 Chr.14: 34765073-34765073 on Build GRCh38
rs145046055 Chr.14: 34765077-34765077 on Build GRCh38

View Full Product Details