Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 36678915-36679464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A21
Entrez Gene ID
89874
Gene Name
solute carrier family 25 member 21
Gene Aliases
ODC, ODC1
UniGene
Hs.730857
Gene Chromosome Location
Chr.14: 36677921-37172660 on Build GRCh38
Transcripts
NM_001171170.1 NM_030631.3

SNP Detail

SNP ID Chromosome Location
rs902145808 Chr.14: 36679293-36679293 on Build GRCh38
rs561785441 Chr.14: 36679250-36679250 on Build GRCh38
rs937523608 Chr.14: 36678983-36678983 on Build GRCh38
rs915462440 Chr.14: 36679100-36679100 on Build GRCh38
rs1012495817 Chr.14: 36679151-36679151 on Build GRCh38
rs899077106 Chr.14: 36679175-36679175 on Build GRCh38
rs951442313 Chr.14: 36679407-36679407 on Build GRCh38
rs971835150 Chr.14: 36678979-36678979 on Build GRCh38
rs869281740 Chr.14: 36679016-36679017 on Build GRCh38
rs914752111 Chr.14: 36679224-36679224 on Build GRCh38
rs775390006 Chr.14: 36679139-36679139 on Build GRCh38
rs370956621 Chr.14: 36679180-36679181 on Build GRCh38
rs550721306 Chr.14: 36679284-36679284 on Build GRCh38
rs750680922 Chr.14: 36679348-36679348 on Build GRCh38
rs937490879 Chr.14: 36679190-36679190 on Build GRCh38
rs5807898 Chr.14: 36679015-36679016 on Build GRCh38
rs756536717 Chr.14: 36679371-36679371 on Build GRCh38
rs1018067264 Chr.14: 36679240-36679240 on Build GRCh38
rs981569842 Chr.14: 36678942-36678943 on Build GRCh38
rs17104978 Chr.14: 36679431-36679431 on Build GRCh38
rs954729664 Chr.14: 36679215-36679215 on Build GRCh38
rs149325199 Chr.14: 36678950-36678950 on Build GRCh38
rs564161819 Chr.14: 36679136-36679136 on Build GRCh38
rs879381449 Chr.14: 36679180-36679181 on Build GRCh38
rs567055094 Chr.14: 36679404-36679404 on Build GRCh38
rs995803297 Chr.14: 36679373-36679374 on Build GRCh38
rs530641486 Chr.14: 36679258-36679258 on Build GRCh38
rs386381127 Chr.14: 36679017-36679018 on Build GRCh38
rs545678409 Chr.14: 36679173-36679173 on Build GRCh38
rs550228962 Chr.14: 36679436-36679437 on Build GRCh38
rs113789011 Chr.14: 36679178-36679179 on Build GRCh38
rs775337481 Chr.14: 36679018-36679018 on Build GRCh38
rs894180184 Chr.14: 36679383-36679383 on Build GRCh38
rs951426974 Chr.14: 36679160-36679160 on Build GRCh38
rs898348460 Chr.14: 36679369-36679370 on Build GRCh38
rs939493835 Chr.14: 36679363-36679363 on Build GRCh38
rs945645220 Chr.14: 36679106-36679106 on Build GRCh38
rs3061566 Chr.14: 36679018-36679019 on Build GRCh38
rs141752368 Chr.14: 36679294-36679294 on Build GRCh38
rs755152754 Chr.14: 36679121-36679122 on Build GRCh38
rs963265423 Chr.14: 36678958-36678958 on Build GRCh38
rs1035769969 Chr.14: 36679423-36679423 on Build GRCh38
rs906720677 Chr.14: 36679114-36679114 on Build GRCh38
rs958744684 Chr.14: 36679428-36679428 on Build GRCh38
rs1037326909 Chr.14: 36679414-36679414 on Build GRCh38
rs928251886 Chr.14: 36679344-36679344 on Build GRCh38
rs919119033 Chr.14: 36678988-36678988 on Build GRCh38
rs61994921 Chr.14: 36679011-36679011 on Build GRCh38
rs761852269 Chr.14: 36679280-36679280 on Build GRCh38
rs754786807 Chr.14: 36679283-36679284 on Build GRCh38
rs1048200 Chr.14: 36679180-36679180 on Build GRCh38
rs549719359 Chr.14: 36679368-36679369 on Build GRCh38
rs148038041 Chr.14: 36679128-36679128 on Build GRCh38
rs555784150 Chr.14: 36679091-36679091 on Build GRCh38
rs954813889 Chr.14: 36679000-36679000 on Build GRCh38
rs372555719 Chr.14: 36678974-36678974 on Build GRCh38
rs1050781166 Chr.14: 36679347-36679348 on Build GRCh38
rs547267555 Chr.14: 36679375-36679375 on Build GRCh38
rs539477713 Chr.14: 36679303-36679303 on Build GRCh38
rs183106288 Chr.14: 36679228-36679228 on Build GRCh38
rs759296234 Chr.14: 36679118-36679118 on Build GRCh38
rs575344321 Chr.14: 36679107-36679107 on Build GRCh38
rs17104977 Chr.14: 36679154-36679154 on Build GRCh38
rs902178122 Chr.14: 36679024-36679024 on Build GRCh38
rs1048199 Chr.14: 36679043-36679043 on Build GRCh38
rs188526122 Chr.14: 36679432-36679432 on Build GRCh38
rs1002615515 Chr.14: 36679116-36679116 on Build GRCh38
rs144585547 Chr.14: 36679050-36679050 on Build GRCh38
rs1684 Chr.14: 36679181-36679181 on Build GRCh38

View Full Product Details