Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 44927922-44928458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL28
Entrez Gene ID
54813
Gene Name
kelch like family member 28
Gene Aliases
BTBD5
UniGene
Hs.550906
Gene Chromosome Location
Chr.14: 44924310-44961978 on Build GRCh38
Transcripts
NM_001308112.1 NM_017658.4 XM_005267770.3 XM_011536847.2 XM_011536849.2

SNP Detail

SNP ID Chromosome Location
rs1007461115 Chr.14: 44928127-44928127 on Build GRCh38
rs906163987 Chr.14: 44928180-44928180 on Build GRCh38
rs6572290 Chr.14: 44927962-44927962 on Build GRCh38
rs79851852 Chr.14: 44928152-44928152 on Build GRCh38
rs886888408 Chr.14: 44928240-44928240 on Build GRCh38
rs187317062 Chr.14: 44928185-44928185 on Build GRCh38
rs1054055918 Chr.14: 44928080-44928080 on Build GRCh38
rs1016503024 Chr.14: 44927956-44927956 on Build GRCh38
rs759067400 Chr.14: 44928217-44928217 on Build GRCh38
rs987899040 Chr.14: 44928344-44928344 on Build GRCh38
rs760437512 Chr.14: 44927957-44927957 on Build GRCh38
rs199601468 Chr.14: 44927997-44927997 on Build GRCh38
rs1045006775 Chr.14: 44928176-44928176 on Build GRCh38
rs10129456 Chr.14: 44928404-44928404 on Build GRCh38
rs1029214997 Chr.14: 44928332-44928332 on Build GRCh38
rs998212529 Chr.14: 44928196-44928196 on Build GRCh38
rs963518858 Chr.14: 44928295-44928295 on Build GRCh38
rs369559749 Chr.14: 44928010-44928010 on Build GRCh38
rs1041348598 Chr.14: 44928072-44928072 on Build GRCh38
rs924034411 Chr.14: 44928372-44928372 on Build GRCh38
rs1025618869 Chr.14: 44928198-44928198 on Build GRCh38
rs902958907 Chr.14: 44928075-44928075 on Build GRCh38
rs567286566 Chr.14: 44928431-44928431 on Build GRCh38
rs370424922 Chr.14: 44927993-44927993 on Build GRCh38
rs893936322 Chr.14: 44928104-44928104 on Build GRCh38
rs879518879 Chr.14: 44928343-44928343 on Build GRCh38
rs924038109 Chr.14: 44928043-44928043 on Build GRCh38
rs996351593 Chr.14: 44928331-44928331 on Build GRCh38
rs1005044623 Chr.14: 44928255-44928255 on Build GRCh38
rs909966649 Chr.14: 44928308-44928308 on Build GRCh38
rs987546157 Chr.14: 44928030-44928030 on Build GRCh38
rs954719953 Chr.14: 44928002-44928002 on Build GRCh38
rs915152355 Chr.14: 44928149-44928149 on Build GRCh38
rs947874893 Chr.14: 44928422-44928422 on Build GRCh38
rs999874401 Chr.14: 44928095-44928095 on Build GRCh38
rs951448575 Chr.14: 44928214-44928214 on Build GRCh38
rs1016886038 Chr.14: 44928258-44928258 on Build GRCh38
rs921900081 Chr.14: 44927985-44927985 on Build GRCh38
rs989325208 Chr.14: 44928400-44928400 on Build GRCh38
rs1046793778 Chr.14: 44928220-44928220 on Build GRCh38
rs935158977 Chr.14: 44928057-44928057 on Build GRCh38
rs1053890650 Chr.14: 44928097-44928097 on Build GRCh38
rs551567585 Chr.14: 44928064-44928064 on Build GRCh38
rs957038161 Chr.14: 44928389-44928389 on Build GRCh38

View Full Product Details