Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50721086-50721603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIN
Entrez Gene ID
51199
Gene Name
ninein
Gene Aliases
SCKL7
UniGene
Hs.310429
Gene Chromosome Location
Chr.14: 50719763-50831121 on Build GRCh38
Transcripts
NM_020921.3 XM_011536817.1 XM_011536822.1

SNP Detail

SNP ID Chromosome Location
rs368803967 Chr.14: 50721390-50721396 on Build GRCh38
rs997365028 Chr.14: 50721126-50721126 on Build GRCh38
rs1056669417 Chr.14: 50721125-50721125 on Build GRCh38
rs942043697 Chr.14: 50721375-50721375 on Build GRCh38
rs761805671 Chr.14: 50721537-50721537 on Build GRCh38
rs905899611 Chr.14: 50721214-50721214 on Build GRCh38
rs946188760 Chr.14: 50721291-50721291 on Build GRCh38
rs918516679 Chr.14: 50721374-50721374 on Build GRCh38
rs533640180 Chr.14: 50721383-50721383 on Build GRCh38
rs552564031 Chr.14: 50721111-50721111 on Build GRCh38
rs987839980 Chr.14: 50721441-50721441 on Build GRCh38
rs953258633 Chr.14: 50721161-50721161 on Build GRCh38
rs905933619 Chr.14: 50721492-50721492 on Build GRCh38
rs891786383 Chr.14: 50721536-50721536 on Build GRCh38
rs1033242031 Chr.14: 50721241-50721241 on Build GRCh38
rs568859145 Chr.14: 50721117-50721117 on Build GRCh38
rs138342257 Chr.14: 50721325-50721325 on Build GRCh38
rs1010411460 Chr.14: 50721270-50721270 on Build GRCh38
rs896314076 Chr.14: 50721191-50721191 on Build GRCh38
rs4534750 Chr.14: 50721406-50721406 on Build GRCh38
rs929918894 Chr.14: 50721381-50721381 on Build GRCh38
rs1030200160 Chr.14: 50721160-50721160 on Build GRCh38
rs185609961 Chr.14: 50721202-50721202 on Build GRCh38
rs753917470 Chr.14: 50721524-50721524 on Build GRCh38
rs1020604077 Chr.14: 50721450-50721450 on Build GRCh38
rs1044439849 Chr.14: 50721207-50721207 on Build GRCh38
rs1000121254 Chr.14: 50721235-50721235 on Build GRCh38
rs973697379 Chr.14: 50721358-50721358 on Build GRCh38
rs559712698 Chr.14: 50721350-50721350 on Build GRCh38
rs553940949 Chr.14: 50721405-50721405 on Build GRCh38
rs888986746 Chr.14: 50721418-50721418 on Build GRCh38
rs140292137 Chr.14: 50721384-50721390 on Build GRCh38
rs955550506 Chr.14: 50721317-50721317 on Build GRCh38
rs567856589 Chr.14: 50721335-50721335 on Build GRCh38
rs981863423 Chr.14: 50721530-50721530 on Build GRCh38
rs1007858192 Chr.14: 50721422-50721422 on Build GRCh38
rs143827249 Chr.14: 50721436-50721436 on Build GRCh38
rs956251375 Chr.14: 50721251-50721251 on Build GRCh38
rs200555037 Chr.14: 50721347-50721350 on Build GRCh38
rs776859806 Chr.14: 50721439-50721441 on Build GRCh38
rs573996706 Chr.14: 50721361-50721362 on Build GRCh38
rs778090069 Chr.14: 50721253-50721254 on Build GRCh38
rs1030284001 Chr.14: 50721281-50721281 on Build GRCh38
rs554128814 Chr.14: 50721312-50721312 on Build GRCh38
rs8021903 Chr.14: 50721513-50721513 on Build GRCh38
rs764323413 Chr.14: 50721419-50721419 on Build GRCh38
rs988259221 Chr.14: 50721220-50721220 on Build GRCh38

View Full Product Details