Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 51969215-51969724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG2
Entrez Gene ID
54331
Gene Name
G protein subunit gamma 2
Gene Aliases
-
UniGene
Hs.187772
Gene Chromosome Location
Chr.14: 51847131-51969800 on Build GRCh38
Transcripts
NM_001243773.1 NM_001243774.1 NM_053064.4 XM_006720173.2 XM_011536846.2 XM_017021376.1 XM_017021377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6500195 c.141T>C p.Y47Y Chr.14: 51969389-51969389 on Build GRCh38
7333484 c.100C>G p.Q34E Chr.14: 51969430-51969430 on Build GRCh38
3700870 c.200T>C p.L67P Chr.14: 51969330-51969330 on Build GRCh38
6215549 c.40G>A p.V14I Chr.14: 51969490-51969490 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs958745006 Chr.14: 51969692-51969692 on Build GRCh38
rs866794142 Chr.14: 51969275-51969275 on Build GRCh38
rs914964189 Chr.14: 51969418-51969418 on Build GRCh38
rs977425452 Chr.14: 51969480-51969480 on Build GRCh38
rs1010805656 Chr.14: 51969270-51969270 on Build GRCh38
rs145926792 Chr.14: 51969405-51969405 on Build GRCh38
rs865819241 Chr.14: 51969445-51969445 on Build GRCh38
rs1000535633 Chr.14: 51969283-51969283 on Build GRCh38
rs776680178 Chr.14: 51969589-51969589 on Build GRCh38
rs929723189 Chr.14: 51969652-51969652 on Build GRCh38
rs45455697 Chr.14: 51969393-51969393 on Build GRCh38
rs182540561 Chr.14: 51969488-51969488 on Build GRCh38
rs572675907 Chr.14: 51969333-51969333 on Build GRCh38
rs929651236 Chr.14: 51969307-51969307 on Build GRCh38
rs1030477018 Chr.14: 51969310-51969310 on Build GRCh38
rs995981185 Chr.14: 51969308-51969308 on Build GRCh38
rs11157869 Chr.14: 51969392-51969392 on Build GRCh38
rs1014678582 Chr.14: 51969660-51969660 on Build GRCh38
rs954597697 Chr.14: 51969315-51969315 on Build GRCh38
rs916073639 Chr.14: 51969257-51969257 on Build GRCh38
rs954591530 Chr.14: 51969539-51969539 on Build GRCh38
rs950225322 Chr.14: 51969628-51969628 on Build GRCh38
rs138189498 Chr.14: 51969337-51969337 on Build GRCh38
rs76213020 Chr.14: 51969287-51969287 on Build GRCh38
rs904752357 Chr.14: 51969277-51969277 on Build GRCh38
rs907818373 Chr.14: 51969386-51969386 on Build GRCh38
rs927672812 Chr.14: 51969646-51969646 on Build GRCh38
rs542491696 Chr.14: 51969490-51969490 on Build GRCh38
rs904783289 Chr.14: 51969491-51969491 on Build GRCh38
rs900193624 Chr.14: 51969515-51969515 on Build GRCh38
rs748007771 Chr.14: 51969662-51969662 on Build GRCh38
rs111544575 Chr.14: 51969549-51969549 on Build GRCh38
rs45595737 Chr.14: 51969449-51969449 on Build GRCh38
rs983663759 Chr.14: 51969343-51969343 on Build GRCh38
rs1039977082 Chr.14: 51969514-51969514 on Build GRCh38
rs556056738 Chr.14: 51969297-51969297 on Build GRCh38
rs139516966 Chr.14: 51969533-51969533 on Build GRCh38
rs1004725113 Chr.14: 51969614-51969614 on Build GRCh38
rs368214525 Chr.14: 51969611-51969612 on Build GRCh38
rs981839197 Chr.14: 51969642-51969644 on Build GRCh38
rs112910664 Chr.14: 51969494-51969494 on Build GRCh38
rs45543441 Chr.14: 51969684-51969684 on Build GRCh38
rs992189457 Chr.14: 51969501-51969501 on Build GRCh38
rs973315062 Chr.14: 51969664-51969664 on Build GRCh38
rs1018066750 Chr.14: 51969385-51969385 on Build GRCh38
rs746479551 Chr.14: 51969478-51969478 on Build GRCh38
rs1005888242 Chr.14: 51969383-51969383 on Build GRCh38
rs927335448 Chr.14: 51969396-51969396 on Build GRCh38
rs996232320 Chr.14: 51969516-51969516 on Build GRCh38
rs551315275 Chr.14: 51969552-51969552 on Build GRCh38
rs543554898 Chr.14: 51969406-51969406 on Build GRCh38
rs993212185 Chr.14: 51969697-51969697 on Build GRCh38

View Full Product Details