Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52553431-52553933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR137C
Entrez Gene ID
283554
Gene Name
G protein-coupled receptor 137C
Gene Aliases
TM7SF1L2
UniGene
Hs.416214
Gene Chromosome Location
Chr.14: 52553148-52637713 on Build GRCh38
Transcripts
NM_001099652.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5077567 c.296T>C p.L99P Chr.14: 52553443-52553443 on Build GRCh38
5740101 c.386G>A p.C129Y Chr.14: 52553533-52553533 on Build GRCh38
4647766 c.297C>T p.L99L Chr.14: 52553444-52553444 on Build GRCh38
470019 c.323G>A p.G108D Chr.14: 52553470-52553470 on Build GRCh38
5740102 c.427A>G p.N143D Chr.14: 52553574-52553574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766673125 Chr.14: 52553473-52553473 on Build GRCh38
rs751364876 Chr.14: 52553633-52553633 on Build GRCh38
rs763713796 Chr.14: 52553614-52553614 on Build GRCh38
rs756360872 Chr.14: 52553582-52553582 on Build GRCh38
rs570635989 Chr.14: 52553905-52553905 on Build GRCh38
rs756450512 Chr.14: 52553634-52553634 on Build GRCh38
rs375517618 Chr.14: 52553469-52553469 on Build GRCh38
rs769056681 Chr.14: 52553506-52553506 on Build GRCh38
rs775792789 Chr.14: 52553610-52553610 on Build GRCh38
rs1004419652 Chr.14: 52553599-52553601 on Build GRCh38
rs934105267 Chr.14: 52553474-52553474 on Build GRCh38
rs763398548 Chr.14: 52553516-52553516 on Build GRCh38
rs773857877 Chr.14: 52553462-52553462 on Build GRCh38
rs970282238 Chr.14: 52553834-52553834 on Build GRCh38
rs1004351109 Chr.14: 52553807-52553808 on Build GRCh38
rs905070629 Chr.14: 52553772-52553775 on Build GRCh38
rs201785222 Chr.14: 52553475-52553475 on Build GRCh38
rs773373618 Chr.14: 52553513-52553513 on Build GRCh38
rs753358489 Chr.14: 52553567-52553567 on Build GRCh38
rs368348208 Chr.14: 52553456-52553456 on Build GRCh38
rs928627063 Chr.14: 52553845-52553845 on Build GRCh38
rs921245476 Chr.14: 52553471-52553471 on Build GRCh38
rs113975789 Chr.14: 52553487-52553487 on Build GRCh38
rs1046857479 Chr.14: 52553629-52553629 on Build GRCh38
rs375496859 Chr.14: 52553576-52553576 on Build GRCh38
rs767147076 Chr.14: 52553477-52553477 on Build GRCh38
rs757191003 Chr.14: 52553568-52553568 on Build GRCh38
rs763457316 Chr.14: 52553464-52553464 on Build GRCh38
rs777209184 Chr.14: 52553490-52553490 on Build GRCh38
rs369741603 Chr.14: 52553611-52553611 on Build GRCh38
rs980346418 Chr.14: 52553842-52553842 on Build GRCh38
rs913321524 Chr.14: 52553879-52553879 on Build GRCh38
rs781105140 Chr.14: 52553788-52553788 on Build GRCh38
rs749377591 Chr.14: 52553599-52553599 on Build GRCh38
rs950924502 Chr.14: 52553785-52553785 on Build GRCh38
rs773411526 Chr.14: 52553864-52553864 on Build GRCh38
rs946608877 Chr.14: 52553515-52553515 on Build GRCh38
rs750251127 Chr.14: 52553573-52553573 on Build GRCh38
rs757178604 Chr.14: 52553493-52553493 on Build GRCh38
rs903781617 Chr.14: 52553601-52553601 on Build GRCh38
rs1056132140 Chr.14: 52553792-52553792 on Build GRCh38
rs765221976 Chr.14: 52553713-52553713 on Build GRCh38
rs938697661 Chr.14: 52553856-52553856 on Build GRCh38
rs527677227 Chr.14: 52553870-52553870 on Build GRCh38
rs963015561 Chr.14: 52553603-52553603 on Build GRCh38
rs200851420 Chr.14: 52553459-52553459 on Build GRCh38
rs373165564 Chr.14: 52553625-52553625 on Build GRCh38
rs774640416 Chr.14: 52553531-52553531 on Build GRCh38
rs562258984 Chr.14: 52553765-52553765 on Build GRCh38
rs1045019775 Chr.14: 52553710-52553710 on Build GRCh38
rs770004651 Chr.14: 52553510-52553510 on Build GRCh38
rs1016894130 Chr.14: 52553794-52553794 on Build GRCh38
rs1043561011 Chr.14: 52553908-52553908 on Build GRCh38
rs545963736 Chr.14: 52553581-52553581 on Build GRCh38
rs201559342 Chr.14: 52553559-52553559 on Build GRCh38
rs562760224 Chr.14: 52553607-52553607 on Build GRCh38
rs753743830 Chr.14: 52553492-52553492 on Build GRCh38
rs955378867 Chr.14: 52553619-52553619 on Build GRCh38
rs779935112 Chr.14: 52553577-52553577 on Build GRCh38
rs368087789 Chr.14: 52553481-52553481 on Build GRCh38
rs372313581 Chr.14: 52553560-52553560 on Build GRCh38
rs753975518 Chr.14: 52553634-52553634 on Build GRCh38
rs915048021 Chr.14: 52553685-52553685 on Build GRCh38
rs1026075367 Chr.14: 52553779-52553779 on Build GRCh38
rs1017277004 Chr.14: 52553680-52553680 on Build GRCh38
rs375079082 Chr.14: 52553505-52553505 on Build GRCh38
rs898671635 Chr.14: 52553819-52553819 on Build GRCh38
rs934741542 Chr.14: 52553675-52553675 on Build GRCh38
rs777750722 Chr.14: 52553587-52553587 on Build GRCh38
rs748312487 Chr.14: 52553507-52553507 on Build GRCh38
rs984979827 Chr.14: 52553787-52553787 on Build GRCh38
rs745653802 Chr.14: 52553499-52553499 on Build GRCh38
rs750385768 Chr.14: 52553630-52553630 on Build GRCh38
rs766305402 Chr.14: 52553631-52553631 on Build GRCh38
rs541963216 Chr.14: 52553696-52553696 on Build GRCh38
rs761381734 Chr.14: 52553622-52553622 on Build GRCh38
rs1007517300 Chr.14: 52553655-52553655 on Build GRCh38
rs759944878 Chr.14: 52553549-52553549 on Build GRCh38
rs577150825 Chr.14: 52553561-52553561 on Build GRCh38
rs369106861 Chr.14: 52553528-52553528 on Build GRCh38
rs1017130716 Chr.14: 52553808-52553811 on Build GRCh38
rs763627535 Chr.14: 52553562-52553562 on Build GRCh38
rs1053237685 Chr.14: 52553551-52553551 on Build GRCh38
rs986866270 Chr.14: 52553484-52553484 on Build GRCh38
rs939004407 Chr.14: 52553776-52553776 on Build GRCh38
rs537475506 Chr.14: 52553835-52553839 on Build GRCh38
rs765206310 Chr.14: 52553570-52553570 on Build GRCh38
rs752589914 Chr.14: 52553820-52553821 on Build GRCh38
rs547805720 Chr.14: 52553873-52553873 on Build GRCh38
rs965723262 Chr.14: 52553683-52553683 on Build GRCh38

View Full Product Details