Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69711644-69712138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUSD6
Entrez Gene ID
9766
Gene Name
sushi domain containing 6
Gene Aliases
DRAGO, KIAA0247
UniGene
Hs.440025
Gene Chromosome Location
Chr.14: 69611593-69715144 on Build GRCh38
Transcripts
NM_014734.3 XM_011537412.1 XM_011537413.1 XM_017021838.1 XM_017021839.1

SNP Detail

SNP ID Chromosome Location
rs1002771076 Chr.14: 69712054-69712054 on Build GRCh38
rs1021359618 Chr.14: 69711814-69711814 on Build GRCh38
rs774860456 Chr.14: 69711783-69711783 on Build GRCh38
rs146806205 Chr.14: 69712053-69712053 on Build GRCh38
rs758885856 Chr.14: 69711863-69711863 on Build GRCh38
rs943583346 Chr.14: 69711855-69711855 on Build GRCh38
rs575781744 Chr.14: 69712055-69712055 on Build GRCh38
rs376732654 Chr.14: 69711840-69711841 on Build GRCh38
rs539185373 Chr.14: 69711686-69711686 on Build GRCh38
rs1006596533 Chr.14: 69712102-69712102 on Build GRCh38
rs749450103 Chr.14: 69711693-69711693 on Build GRCh38
rs1003213446 Chr.14: 69711728-69711728 on Build GRCh38
rs929324222 Chr.14: 69711950-69711950 on Build GRCh38
rs1044218929 Chr.14: 69712050-69712051 on Build GRCh38
rs140048396 Chr.14: 69711725-69711725 on Build GRCh38
rs968184123 Chr.14: 69711815-69711815 on Build GRCh38
rs3809397 Chr.14: 69712109-69712109 on Build GRCh38
rs1022888172 Chr.14: 69711720-69711720 on Build GRCh38
rs544253585 Chr.14: 69711832-69711833 on Build GRCh38
rs370334013 Chr.14: 69711856-69711856 on Build GRCh38
rs374999814 Chr.14: 69712079-69712079 on Build GRCh38
rs941836899 Chr.14: 69712032-69712032 on Build GRCh38
rs955917369 Chr.14: 69711760-69711760 on Build GRCh38
rs56352508 Chr.14: 69711799-69711799 on Build GRCh38
rs1035323908 Chr.14: 69711748-69711748 on Build GRCh38
rs891447485 Chr.14: 69712057-69712057 on Build GRCh38
rs553327891 Chr.14: 69712009-69712009 on Build GRCh38
rs923265917 Chr.14: 69711922-69711922 on Build GRCh38
rs887594366 Chr.14: 69711699-69711699 on Build GRCh38
rs191077320 Chr.14: 69712080-69712080 on Build GRCh38
rs143672909 Chr.14: 69711874-69711874 on Build GRCh38
rs748604143 Chr.14: 69711791-69711791 on Build GRCh38
rs537053416 Chr.14: 69711849-69711849 on Build GRCh38
rs3809396 Chr.14: 69711718-69711718 on Build GRCh38
rs1021322020 Chr.14: 69712068-69712068 on Build GRCh38
rs578033895 Chr.14: 69712033-69712033 on Build GRCh38
rs773585780 Chr.14: 69711915-69711915 on Build GRCh38
rs1017702150 Chr.14: 69712110-69712110 on Build GRCh38
rs575193487 Chr.14: 69712067-69712067 on Build GRCh38
rs564228706 Chr.14: 69711978-69711978 on Build GRCh38

View Full Product Details