Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73721207-73721712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMSAN1
Entrez Gene ID
91748
Gene Name
ELM2 and Myb/SANT domain containing 1
Gene Aliases
C14orf117, C14orf43, LSR68, MIDEAS, c14_5541
UniGene
Hs.594157
Gene Chromosome Location
Chr.14: 73715122-73790285 on Build GRCh38
Transcripts
NM_001043318.2 NM_194278.3 XM_005268204.1 XM_005268205.1 XM_005268206.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
309604 c.2725-3T>G p.? Chr.14: 73721512-73721512 on Build GRCh38
3754097 c.2773A>G p.S925G Chr.14: 73721461-73721461 on Build GRCh38
6865107 c.2764G>T p.E922* Chr.14: 73721470-73721470 on Build GRCh38
6030662 c.2912C>T p.T971M Chr.14: 73721322-73721322 on Build GRCh38
2137321 c.2755C>T p.P919S Chr.14: 73721479-73721479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146014844 Chr.14: 73721343-73721343 on Build GRCh38
rs771926660 Chr.14: 73721559-73721559 on Build GRCh38
rs767693003 Chr.14: 73721251-73721251 on Build GRCh38
rs761889883 Chr.14: 73721435-73721435 on Build GRCh38
rs368460541 Chr.14: 73721256-73721256 on Build GRCh38
rs748593689 Chr.14: 73721276-73721276 on Build GRCh38
rs143153494 Chr.14: 73721380-73721380 on Build GRCh38
rs947287425 Chr.14: 73721592-73721592 on Build GRCh38
rs928726430 Chr.14: 73721550-73721550 on Build GRCh38
rs370994483 Chr.14: 73721294-73721294 on Build GRCh38
rs919928567 Chr.14: 73721246-73721246 on Build GRCh38
rs745418178 Chr.14: 73721367-73721367 on Build GRCh38
rs753890164 Chr.14: 73721332-73721332 on Build GRCh38
rs776330566 Chr.14: 73721378-73721380 on Build GRCh38
rs1029264029 Chr.14: 73721619-73721619 on Build GRCh38
rs752541058 Chr.14: 73721240-73721240 on Build GRCh38
rs527753518 Chr.14: 73721430-73721430 on Build GRCh38
rs62006063 Chr.14: 73721679-73721679 on Build GRCh38
rs758867456 Chr.14: 73721352-73721352 on Build GRCh38
rs768402164 Chr.14: 73721407-73721407 on Build GRCh38
rs780228313 Chr.14: 73721538-73721538 on Build GRCh38
rs371324179 Chr.14: 73721387-73721387 on Build GRCh38
rs768639350 Chr.14: 73721372-73721374 on Build GRCh38
rs779590133 Chr.14: 73721381-73721381 on Build GRCh38
rs775171109 Chr.14: 73721288-73721288 on Build GRCh38
rs781643450 Chr.14: 73721663-73721663 on Build GRCh38
rs1018357269 Chr.14: 73721669-73721669 on Build GRCh38
rs998956099 Chr.14: 73721634-73721634 on Build GRCh38
rs760840582 Chr.14: 73721451-73721451 on Build GRCh38
rs200154652 Chr.14: 73721325-73721325 on Build GRCh38
rs1056577464 Chr.14: 73721476-73721476 on Build GRCh38
rs772157770 Chr.14: 73721445-73721445 on Build GRCh38
rs750507902 Chr.14: 73721321-73721321 on Build GRCh38
rs189818005 Chr.14: 73721580-73721580 on Build GRCh38
rs367637105 Chr.14: 73721298-73721298 on Build GRCh38
rs915738566 Chr.14: 73721584-73721584 on Build GRCh38
rs541501034 Chr.14: 73721460-73721460 on Build GRCh38
rs964388463 Chr.14: 73721238-73721238 on Build GRCh38
rs755308228 Chr.14: 73721341-73721341 on Build GRCh38
rs974732384 Chr.14: 73721508-73721508 on Build GRCh38
rs750742702 Chr.14: 73721254-73721254 on Build GRCh38
rs779289375 Chr.14: 73721274-73721274 on Build GRCh38
rs746767297 Chr.14: 73721274-73721278 on Build GRCh38
rs755639224 Chr.14: 73721522-73721522 on Build GRCh38
rs754024767 Chr.14: 73721337-73721337 on Build GRCh38
rs544407069 Chr.14: 73721528-73721528 on Build GRCh38
rs147485986 Chr.14: 73721411-73721411 on Build GRCh38
rs375900466 Chr.14: 73721475-73721475 on Build GRCh38
rs143466467 Chr.14: 73721600-73721600 on Build GRCh38
rs547463009 Chr.14: 73721616-73721616 on Build GRCh38
rs1012394127 Chr.14: 73721281-73721281 on Build GRCh38
rs377698481 Chr.14: 73721666-73721666 on Build GRCh38
rs1043385619 Chr.14: 73721594-73721594 on Build GRCh38
rs770482156 Chr.14: 73721278-73721278 on Build GRCh38
rs111761785 Chr.14: 73721512-73721512 on Build GRCh38
rs773242120 Chr.14: 73721450-73721450 on Build GRCh38
rs373693644 Chr.14: 73721567-73721567 on Build GRCh38
rs529548531 Chr.14: 73721560-73721560 on Build GRCh38
rs780517073 Chr.14: 73721262-73721262 on Build GRCh38
rs200140026 Chr.14: 73721304-73721304 on Build GRCh38
rs769463597 Chr.14: 73721287-73721287 on Build GRCh38
rs185896203 Chr.14: 73721376-73721376 on Build GRCh38
rs745572967 Chr.14: 73721283-73721283 on Build GRCh38
rs747985534 Chr.14: 73721549-73721549 on Build GRCh38
rs998593468 Chr.14: 73721395-73721395 on Build GRCh38
rs60635070 Chr.14: 73721461-73721461 on Build GRCh38
rs753087068 Chr.14: 73721488-73721488 on Build GRCh38
rs576413634 Chr.14: 73721358-73721358 on Build GRCh38
rs116418438 Chr.14: 73721236-73721236 on Build GRCh38
rs779614183 Chr.14: 73721527-73721527 on Build GRCh38
rs79954518 Chr.14: 73721504-73721504 on Build GRCh38
rs767746427 Chr.14: 73721312-73721312 on Build GRCh38
rs139139732 Chr.14: 73721370-73721370 on Build GRCh38
rs749634230 Chr.14: 73721266-73721266 on Build GRCh38
rs780672964 Chr.14: 73721280-73721280 on Build GRCh38
rs766538579 Chr.14: 73721336-73721336 on Build GRCh38
rs570493993 Chr.14: 73721643-73721643 on Build GRCh38
rs1018473753 Chr.14: 73721501-73721501 on Build GRCh38
rs540068598 Chr.14: 73721583-73721584 on Build GRCh38
rs755327753 Chr.14: 73721272-73721272 on Build GRCh38

View Full Product Details