Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74955076-74955582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGF
Entrez Gene ID
5228
Gene Name
placental growth factor
Gene Aliases
D12S1900, PGFL, PIGF, PLGF, PlGF-2, SHGC-10760
UniGene
Hs.252820
Gene Chromosome Location
Chr.14: 74941830-74955764 on Build GRCh38
Transcripts
NM_001207012.1 NM_001293643.1 NM_002632.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4613886 c.73delC p.Q25fs*83 Chr.14: 74955170-74955170 on Build GRCh38
6407835 c.62C>T p.A21V Chr.14: 74955181-74955181 on Build GRCh38
5171116 c.45C>T p.A15A Chr.14: 74955198-74955198 on Build GRCh38
5769959 c.31C>A p.L11M Chr.14: 74955212-74955212 on Build GRCh38
6407836 c.62C>T p.A21V Chr.14: 74955181-74955181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906900178 Chr.14: 74955310-74955310 on Build GRCh38
rs1020146714 Chr.14: 74955363-74955363 on Build GRCh38
rs1012065141 Chr.14: 74955138-74955138 on Build GRCh38
rs1031615108 Chr.14: 74955476-74955476 on Build GRCh38
rs45449993 Chr.14: 74955344-74955344 on Build GRCh38
rs1018472154 Chr.14: 74955189-74955189 on Build GRCh38
rs778360122 Chr.14: 74955246-74955248 on Build GRCh38
rs770668471 Chr.14: 74955327-74955327 on Build GRCh38
rs115820345 Chr.14: 74955142-74955142 on Build GRCh38
rs956976703 Chr.14: 74955285-74955285 on Build GRCh38
rs528849914 Chr.14: 74955460-74955460 on Build GRCh38
rs187938882 Chr.14: 74955118-74955118 on Build GRCh38
rs1050848677 Chr.14: 74955384-74955384 on Build GRCh38
rs909363745 Chr.14: 74955188-74955188 on Build GRCh38
rs549282502 Chr.14: 74955467-74955467 on Build GRCh38
rs771954837 Chr.14: 74955441-74955441 on Build GRCh38
rs11542848 Chr.14: 74955472-74955472 on Build GRCh38
rs953438924 Chr.14: 74955403-74955403 on Build GRCh38
rs1006475985 Chr.14: 74955411-74955411 on Build GRCh38
rs545154602 Chr.14: 74955179-74955179 on Build GRCh38
rs953740209 Chr.14: 74955175-74955175 on Build GRCh38
rs980050531 Chr.14: 74955313-74955313 on Build GRCh38
rs997625164 Chr.14: 74955383-74955383 on Build GRCh38
rs573931945 Chr.14: 74955104-74955104 on Build GRCh38
rs893050371 Chr.14: 74955339-74955339 on Build GRCh38
rs549588224 Chr.14: 74955288-74955288 on Build GRCh38
rs753380544 Chr.14: 74955136-74955137 on Build GRCh38
rs558027430 Chr.14: 74955561-74955561 on Build GRCh38
rs890781031 Chr.14: 74955165-74955165 on Build GRCh38
rs11542849 Chr.14: 74955378-74955378 on Build GRCh38
rs532908973 Chr.14: 74955253-74955253 on Build GRCh38
rs919456676 Chr.14: 74955504-74955504 on Build GRCh38
rs759812119 Chr.14: 74955147-74955147 on Build GRCh38
rs770739693 Chr.14: 74955140-74955140 on Build GRCh38
rs963987647 Chr.14: 74955471-74955471 on Build GRCh38
rs918314084 Chr.14: 74955233-74955233 on Build GRCh38
rs1030335959 Chr.14: 74955386-74955386 on Build GRCh38
rs972646723 Chr.14: 74955208-74955208 on Build GRCh38
rs751945485 Chr.14: 74955198-74955198 on Build GRCh38
rs896918248 Chr.14: 74955375-74955375 on Build GRCh38
rs764734526 Chr.14: 74955182-74955182 on Build GRCh38
rs992690460 Chr.14: 74955505-74955505 on Build GRCh38
rs1045358861 Chr.14: 74955300-74955300 on Build GRCh38
rs1024848510 Chr.14: 74955495-74955495 on Build GRCh38
rs565128099 Chr.14: 74955203-74955203 on Build GRCh38
rs369360973 Chr.14: 74955273-74955273 on Build GRCh38
rs927161621 Chr.14: 74955316-74955318 on Build GRCh38
rs531595412 Chr.14: 74955530-74955530 on Build GRCh38
rs1007981032 Chr.14: 74955432-74955432 on Build GRCh38
rs878889106 Chr.14: 74955161-74955161 on Build GRCh38
rs929743166 Chr.14: 74955299-74955299 on Build GRCh38
rs1031100615 Chr.14: 74955252-74955252 on Build GRCh38
rs565935298 Chr.14: 74955468-74955468 on Build GRCh38
rs868687247 Chr.14: 74955239-74955239 on Build GRCh38
rs903239226 Chr.14: 74955382-74955382 on Build GRCh38
rs528662573 Chr.14: 74955151-74955151 on Build GRCh38
rs1007121632 Chr.14: 74955173-74955173 on Build GRCh38
rs941177269 Chr.14: 74955334-74955335 on Build GRCh38
rs775694930 Chr.14: 74955172-74955172 on Build GRCh38
rs937249915 Chr.14: 74955316-74955316 on Build GRCh38
rs893006922 Chr.14: 74955129-74955129 on Build GRCh38
rs928278504 Chr.14: 74955559-74955559 on Build GRCh38
rs904087592 Chr.14: 74955450-74955450 on Build GRCh38
rs951114795 Chr.14: 74955520-74955520 on Build GRCh38
rs756979630 Chr.14: 74955169-74955170 on Build GRCh38

View Full Product Details