Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 89955765-89956279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TDP1
Entrez Gene ID
55775
Gene Name
tyrosyl-DNA phosphodiesterase 1
Gene Aliases
-
UniGene
Hs.209945
Gene Chromosome Location
Chr.14: 89954935-90044764 on Build GRCh38
Transcripts
NM_001008744.1 NM_018319.3 XM_005267847.2 XM_005267848.2 XM_006720197.3 XM_006720199.1 XM_011536942.2 XM_011536944.1 XM_017021439.1 XM_017021440.1 XM_017021441.1 XM_017021444.1 XM_017021446.1

SNP Detail

SNP ID Chromosome Location
rs932340039 Chr.14: 89956117-89956117 on Build GRCh38
rs956656522 Chr.14: 89955907-89955907 on Build GRCh38
rs1042761179 Chr.14: 89956248-89956248 on Build GRCh38
rs947989462 Chr.14: 89955940-89955940 on Build GRCh38
rs550113478 Chr.14: 89955856-89955856 on Build GRCh38
rs886050882 Chr.14: 89955969-89955969 on Build GRCh38
rs954069934 Chr.14: 89956241-89956241 on Build GRCh38
rs542276144 Chr.14: 89956033-89956033 on Build GRCh38
rs952654855 Chr.14: 89956200-89956200 on Build GRCh38
rs1019166628 Chr.14: 89955861-89955861 on Build GRCh38
rs879767772 Chr.14: 89956158-89956158 on Build GRCh38
rs981133873 Chr.14: 89956020-89956020 on Build GRCh38
rs902676776 Chr.14: 89956221-89956221 on Build GRCh38
rs967133285 Chr.14: 89955978-89955978 on Build GRCh38
rs926006867 Chr.14: 89955920-89955920 on Build GRCh38
rs979414749 Chr.14: 89955807-89955807 on Build GRCh38
rs35843847 Chr.14: 89956202-89956202 on Build GRCh38
rs386779854 Chr.14: 89956052-89956053 on Build GRCh38
rs943802014 Chr.14: 89956218-89956218 on Build GRCh38
rs113492506 Chr.14: 89955822-89955822 on Build GRCh38
rs2277520 Chr.14: 89955838-89955838 on Build GRCh38
rs1021636390 Chr.14: 89956066-89956066 on Build GRCh38
rs968169803 Chr.14: 89955995-89955995 on Build GRCh38
rs898225845 Chr.14: 89955843-89955843 on Build GRCh38
rs2277519 Chr.14: 89955868-89955868 on Build GRCh38
rs931059674 Chr.14: 89955949-89955949 on Build GRCh38
rs908417734 Chr.14: 89956207-89956207 on Build GRCh38
rs1014176988 Chr.14: 89956206-89956206 on Build GRCh38
rs35210768 Chr.14: 89955927-89955928 on Build GRCh38
rs776312528 Chr.14: 89956106-89956106 on Build GRCh38
rs886050880 Chr.14: 89955923-89955923 on Build GRCh38
rs943819521 Chr.14: 89956056-89956056 on Build GRCh38
rs886873955 Chr.14: 89955958-89955958 on Build GRCh38
rs540570456 Chr.14: 89955789-89955789 on Build GRCh38
rs1032545154 Chr.14: 89956239-89956239 on Build GRCh38
rs1018984906 Chr.14: 89955798-89955798 on Build GRCh38
rs36084373 Chr.14: 89955844-89955845 on Build GRCh38
rs904085382 Chr.14: 89956257-89956257 on Build GRCh38
rs992849279 Chr.14: 89955965-89955965 on Build GRCh38
rs1045318987 Chr.14: 89955839-89955839 on Build GRCh38
rs562491255 Chr.14: 89956034-89956034 on Build GRCh38
rs954041108 Chr.14: 89956086-89956086 on Build GRCh38
rs35151267 Chr.14: 89956242-89956242 on Build GRCh38
rs985540433 Chr.14: 89956102-89956102 on Build GRCh38
rs961183273 Chr.14: 89956059-89956066 on Build GRCh38
rs533696578 Chr.14: 89955959-89955959 on Build GRCh38
rs1001988614 Chr.14: 89956049-89956049 on Build GRCh38
rs932701348 Chr.14: 89955992-89955992 on Build GRCh38
rs561240946 Chr.14: 89956154-89956154 on Build GRCh38
rs979402718 Chr.14: 89956079-89956079 on Build GRCh38
rs577403021 Chr.14: 89955909-89955909 on Build GRCh38
rs570027662 Chr.14: 89956244-89956244 on Build GRCh38
rs28736874 Chr.14: 89956038-89956038 on Build GRCh38
rs914141155 Chr.14: 89955973-89955973 on Build GRCh38
rs887470072 Chr.14: 89956164-89956164 on Build GRCh38
rs924025917 Chr.14: 89955890-89955890 on Build GRCh38
rs11848712 Chr.14: 89955818-89955818 on Build GRCh38
rs886050881 Chr.14: 89955926-89955926 on Build GRCh38
rs749384201 Chr.14: 89956251-89956251 on Build GRCh38
rs1051084584 Chr.14: 89956119-89956119 on Build GRCh38
rs547981684 Chr.14: 89956100-89956100 on Build GRCh38
rs549548552 Chr.14: 89955838-89955839 on Build GRCh38
rs969753961 Chr.14: 89956183-89956183 on Build GRCh38
rs1046758245 Chr.14: 89955957-89955957 on Build GRCh38
rs34861207 Chr.14: 89956054-89956055 on Build GRCh38
rs904116681 Chr.14: 89956120-89956120 on Build GRCh38
rs573655514 Chr.14: 89956010-89956010 on Build GRCh38
rs751215358 Chr.14: 89956039-89956039 on Build GRCh38
rs565157876 Chr.14: 89955800-89955800 on Build GRCh38
rs980803297 Chr.14: 89956192-89956192 on Build GRCh38
rs966951081 Chr.14: 89956078-89956078 on Build GRCh38
rs1046704229 Chr.14: 89955849-89955849 on Build GRCh38
rs1049005245 Chr.14: 89956160-89956160 on Build GRCh38
rs1044890567 Chr.14: 89955941-89955941 on Build GRCh38
rs917817073 Chr.14: 89955926-89955926 on Build GRCh38
rs956793828 Chr.14: 89956008-89956008 on Build GRCh38
rs902083021 Chr.14: 89955972-89955972 on Build GRCh38
rs892538888 Chr.14: 89956155-89956155 on Build GRCh38
rs1031061815 Chr.14: 89955999-89955999 on Build GRCh38
rs968117490 Chr.14: 89955886-89955886 on Build GRCh38
rs369662288 Chr.14: 89955857-89955857 on Build GRCh38
rs951746308 Chr.14: 89955952-89955952 on Build GRCh38
rs545713755 Chr.14: 89956044-89956044 on Build GRCh38
rs2181599 Chr.14: 89955867-89955867 on Build GRCh38
rs925380883 Chr.14: 89955986-89955986 on Build GRCh38
rs1040282373 Chr.14: 89955934-89955934 on Build GRCh38
rs992420860 Chr.14: 89955846-89955846 on Build GRCh38
rs369947795 Chr.14: 89956012-89956012 on Build GRCh38
rs952212258 Chr.14: 89955831-89955831 on Build GRCh38
rs2277518 Chr.14: 89956053-89956053 on Build GRCh38
rs35070653 Chr.14: 89956052-89956052 on Build GRCh38
rs995243741 Chr.14: 89955844-89955844 on Build GRCh38
rs996865660 Chr.14: 89955980-89955980 on Build GRCh38
rs1013730501 Chr.14: 89956064-89956064 on Build GRCh38
rs780580574 Chr.14: 89956211-89956211 on Build GRCh38
rs1026837299 Chr.14: 89956194-89956194 on Build GRCh38
rs1029604727 Chr.14: 89955879-89955879 on Build GRCh38
rs917834014 Chr.14: 89955825-89955825 on Build GRCh38
rs912630088 Chr.14: 89956103-89956103 on Build GRCh38
rs1024971784 Chr.14: 89956163-89956163 on Build GRCh38
rs919786242 Chr.14: 89955944-89955944 on Build GRCh38
rs747909638 Chr.14: 89955889-89955889 on Build GRCh38
rs917940335 Chr.14: 89956017-89956017 on Build GRCh38
rs993159203 Chr.14: 89955946-89955946 on Build GRCh38
rs770272066 Chr.14: 89956080-89956080 on Build GRCh38
rs935233515 Chr.14: 89955820-89955820 on Build GRCh38
rs34335711 Chr.14: 89955983-89955983 on Build GRCh38
rs1001043789 Chr.14: 89955996-89955996 on Build GRCh38
rs937641622 Chr.14: 89956195-89956195 on Build GRCh38
rs979245026 Chr.14: 89955878-89955878 on Build GRCh38
rs1001009481 Chr.14: 89956122-89956122 on Build GRCh38
rs921032437 Chr.14: 89956235-89956235 on Build GRCh38
rs1027422872 Chr.14: 89955947-89955947 on Build GRCh38
rs976870770 Chr.14: 89956220-89956220 on Build GRCh38
rs773187925 Chr.14: 89955962-89955962 on Build GRCh38
rs757900354 Chr.14: 89956019-89956019 on Build GRCh38

View Full Product Details