Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91232828-91233337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR68
Entrez Gene ID
8111
Gene Name
G protein-coupled receptor 68
Gene Aliases
GPR12A, OGR1
UniGene
Hs.8882
Gene Chromosome Location
Chr.14: 91232532-91264581 on Build GRCh38
Transcripts
NM_001177676.1 NM_003485.3 XM_005268110.4 XM_005268111.3 XM_005268112.3 XM_006720262.3 XM_011537196.2 XM_011537197.2 XM_011537198.2 XM_011537199.2

SNP Detail

SNP ID Chromosome Location
rs565189614 Chr.14: 91232894-91232894 on Build GRCh38
rs556230443 Chr.14: 91233270-91233270 on Build GRCh38
rs191766147 Chr.14: 91232966-91232966 on Build GRCh38
rs1001702770 Chr.14: 91233017-91233017 on Build GRCh38
rs937508141 Chr.14: 91233307-91233307 on Build GRCh38
rs867025462 Chr.14: 91232879-91232879 on Build GRCh38
rs538247848 Chr.14: 91233087-91233087 on Build GRCh38
rs1050936868 Chr.14: 91232977-91232977 on Build GRCh38
rs960278841 Chr.14: 91233072-91233072 on Build GRCh38
rs963330430 Chr.14: 91233124-91233124 on Build GRCh38
rs868752459 Chr.14: 91233303-91233303 on Build GRCh38
rs544930097 Chr.14: 91232881-91232881 on Build GRCh38
rs573077107 Chr.14: 91233093-91233093 on Build GRCh38
rs1004000873 Chr.14: 91232995-91232995 on Build GRCh38
rs2236324 Chr.14: 91233074-91233074 on Build GRCh38
rs943905500 Chr.14: 91233219-91233219 on Build GRCh38
rs534103374 Chr.14: 91233205-91233205 on Build GRCh38
rs887019501 Chr.14: 91233261-91233262 on Build GRCh38
rs1011305318 Chr.14: 91232990-91232990 on Build GRCh38
rs969350842 Chr.14: 91233049-91233049 on Build GRCh38
rs144509908 Chr.14: 91233246-91233246 on Build GRCh38
rs536006462 Chr.14: 91233250-91233250 on Build GRCh38
rs549349898 Chr.14: 91233035-91233035 on Build GRCh38
rs547636522 Chr.14: 91232982-91232982 on Build GRCh38
rs758566974 Chr.14: 91233054-91233054 on Build GRCh38
rs139485231 Chr.14: 91233025-91233025 on Build GRCh38
rs1039836656 Chr.14: 91233217-91233217 on Build GRCh38
rs765700855 Chr.14: 91233005-91233005 on Build GRCh38
rs1039357894 Chr.14: 91232895-91232895 on Build GRCh38
rs898300669 Chr.14: 91233289-91233289 on Build GRCh38
rs544940049 Chr.14: 91233234-91233234 on Build GRCh38
rs149662142 Chr.14: 91233206-91233206 on Build GRCh38
rs928159476 Chr.14: 91233055-91233055 on Build GRCh38
rs899783024 Chr.14: 91232904-91232904 on Build GRCh38
rs1050143700 Chr.14: 91232891-91232891 on Build GRCh38
rs188734044 Chr.14: 91233126-91233126 on Build GRCh38
rs943689396 Chr.14: 91232911-91232911 on Build GRCh38
rs61741976 Chr.14: 91233118-91233118 on Build GRCh38
rs748459063 Chr.14: 91232884-91232884 on Build GRCh38
rs576285910 Chr.14: 91232868-91232868 on Build GRCh38
rs932490383 Chr.14: 91232885-91232885 on Build GRCh38
rs756058370 Chr.14: 91233183-91233183 on Build GRCh38
rs973896268 Chr.14: 91232882-91232882 on Build GRCh38
rs946467555 Chr.14: 91233152-91233152 on Build GRCh38
rs912478587 Chr.14: 91233208-91233208 on Build GRCh38
rs983413232 Chr.14: 91232854-91232854 on Build GRCh38
rs571628511 Chr.14: 91233148-91233148 on Build GRCh38
rs76458218 Chr.14: 91233271-91233271 on Build GRCh38
rs183293895 Chr.14: 91233004-91233004 on Build GRCh38
rs994946584 Chr.14: 91233291-91233291 on Build GRCh38
rs933619601 Chr.14: 91233240-91233240 on Build GRCh38
rs752905771 Chr.14: 91233007-91233007 on Build GRCh38
rs754240578 Chr.14: 91232926-91232926 on Build GRCh38

View Full Product Details