Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91782139-91782687 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TC2N
Entrez Gene ID
123036
Gene Name
tandem C2 domains, nuclear
Gene Aliases
C14orf47, C2CD1, MTAC2D1, Tac2-N
UniGene
Hs.510262
Gene Chromosome Location
Chr.14: 91779751-91867536 on Build GRCh38
Transcripts
NM_001128595.2 NM_001128596.2 NM_001289134.1 NM_152332.5

SNP Detail

SNP ID Chromosome Location
rs1043303478 Chr.14: 91782476-91782476 on Build GRCh38
rs1026188340 Chr.14: 91782507-91782507 on Build GRCh38
rs998981347 Chr.14: 91782318-91782318 on Build GRCh38
rs1005081358 Chr.14: 91782533-91782533 on Build GRCh38
rs950206561 Chr.14: 91782210-91782210 on Build GRCh38
rs534709690 Chr.14: 91782512-91782512 on Build GRCh38
rs973342923 Chr.14: 91782242-91782242 on Build GRCh38
rs892002641 Chr.14: 91782420-91782420 on Build GRCh38
rs1033391257 Chr.14: 91782336-91782336 on Build GRCh38
rs545474398 Chr.14: 91782229-91782229 on Build GRCh38
rs903262623 Chr.14: 91782228-91782228 on Build GRCh38
rs905502778 Chr.14: 91782492-91782492 on Build GRCh38
rs10498630 Chr.14: 91782541-91782541 on Build GRCh38
rs947621515 Chr.14: 91782425-91782425 on Build GRCh38
rs760550457 Chr.14: 91782352-91782356 on Build GRCh38
rs954141581 Chr.14: 91782601-91782601 on Build GRCh38
rs530197125 Chr.14: 91782181-91782181 on Build GRCh38
rs747768529 Chr.14: 91782353-91782353 on Build GRCh38
rs963290639 Chr.14: 91782238-91782238 on Build GRCh38
rs12894042 Chr.14: 91782608-91782608 on Build GRCh38
rs988879291 Chr.14: 91782653-91782653 on Build GRCh38
rs965488660 Chr.14: 91782637-91782637 on Build GRCh38
rs1045922119 Chr.14: 91782223-91782223 on Build GRCh38
rs566021491 Chr.14: 91782475-91782475 on Build GRCh38
rs1035815016 Chr.14: 91782511-91782511 on Build GRCh38
rs746476938 Chr.14: 91782619-91782619 on Build GRCh38
rs574972896 Chr.14: 91782568-91782568 on Build GRCh38
rs912649107 Chr.14: 91782396-91782396 on Build GRCh38
rs985924282 Chr.14: 91782387-91782387 on Build GRCh38
rs894618133 Chr.14: 91782176-91782176 on Build GRCh38
rs1053208164 Chr.14: 91782409-91782409 on Build GRCh38
rs575948943 Chr.14: 91782586-91782586 on Build GRCh38
rs963238733 Chr.14: 91782563-91782563 on Build GRCh38
rs549945835 Chr.14: 91782268-91782268 on Build GRCh38
rs912586681 Chr.14: 91782622-91782622 on Build GRCh38
rs867395384 Chr.14: 91782280-91782280 on Build GRCh38
rs565138738 Chr.14: 91782542-91782542 on Build GRCh38
rs192930525 Chr.14: 91782429-91782429 on Build GRCh38
rs1001213068 Chr.14: 91782495-91782495 on Build GRCh38
rs188135015 Chr.14: 91782400-91782400 on Build GRCh38
rs114965754 Chr.14: 91782406-91782406 on Build GRCh38

View Full Product Details