Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92797794-92798305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA5
Entrez Gene ID
9950
Gene Name
golgin A5
Gene Aliases
GOLIM5, RFG5, ret-II
UniGene
Hs.104320
Gene Chromosome Location
Chr.14: 92794231-92839963 on Build GRCh38
Transcripts
NM_005113.3 XM_011537420.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4896246 c.382A>G p.N128D Chr.14: 92797819-92797819 on Build GRCh38
5478788 c.519A>C p.E173D Chr.14: 92797956-92797956 on Build GRCh38
4053081 c.543C>T p.H181H Chr.14: 92797980-92797980 on Build GRCh38
6374028 c.401C>G p.P134R Chr.14: 92797838-92797838 on Build GRCh38
1323330 c.438G>C p.K146N Chr.14: 92797875-92797875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054299736 Chr.14: 92798260-92798263 on Build GRCh38
rs897184705 Chr.14: 92798240-92798240 on Build GRCh38
rs141963625 Chr.14: 92797845-92797845 on Build GRCh38
rs372821434 Chr.14: 92797971-92797971 on Build GRCh38
rs760030435 Chr.14: 92797927-92797927 on Build GRCh38
rs763264755 Chr.14: 92798265-92798267 on Build GRCh38
rs1055192773 Chr.14: 92798150-92798150 on Build GRCh38
rs751560647 Chr.14: 92797862-92797862 on Build GRCh38
rs1007613639 Chr.14: 92798027-92798027 on Build GRCh38
rs764175277 Chr.14: 92797964-92797964 on Build GRCh38
rs764349513 Chr.14: 92797843-92797843 on Build GRCh38
rs779797527 Chr.14: 92797881-92797881 on Build GRCh38
rs757380809 Chr.14: 92797880-92797880 on Build GRCh38
rs74072488 Chr.14: 92798111-92798111 on Build GRCh38
rs749255638 Chr.14: 92797885-92797885 on Build GRCh38
rs145809323 Chr.14: 92797905-92797905 on Build GRCh38
rs114386227 Chr.14: 92798118-92798118 on Build GRCh38
rs149196374 Chr.14: 92797838-92797838 on Build GRCh38
rs993796751 Chr.14: 92798046-92798046 on Build GRCh38
rs370631881 Chr.14: 92797970-92797970 on Build GRCh38
rs568209097 Chr.14: 92798207-92798207 on Build GRCh38
rs566422501 Chr.14: 92798109-92798109 on Build GRCh38
rs950036691 Chr.14: 92797958-92797958 on Build GRCh38
rs952237045 Chr.14: 92798103-92798103 on Build GRCh38
rs376035342 Chr.14: 92797884-92797884 on Build GRCh38
rs76861952 Chr.14: 92797824-92797824 on Build GRCh38
rs961384993 Chr.14: 92797897-92797897 on Build GRCh38
rs2284956 Chr.14: 92798201-92798201 on Build GRCh38
rs935876619 Chr.14: 92798247-92798247 on Build GRCh38
rs995475197 Chr.14: 92798248-92798248 on Build GRCh38
rs775810727 Chr.14: 92797942-92797942 on Build GRCh38
rs776933561 Chr.14: 92797920-92797920 on Build GRCh38
rs1027087793 Chr.14: 92798274-92798274 on Build GRCh38
rs770485468 Chr.14: 92797916-92797916 on Build GRCh38
rs778840316 Chr.14: 92797893-92797893 on Build GRCh38
rs35729851 Chr.14: 92797906-92797906 on Build GRCh38
rs753126089 Chr.14: 92797842-92797842 on Build GRCh38
rs988725389 Chr.14: 92798047-92798047 on Build GRCh38
rs765790077 Chr.14: 92797928-92797928 on Build GRCh38
rs372642243 Chr.14: 92798029-92798029 on Build GRCh38
rs7148156 Chr.14: 92798188-92798188 on Build GRCh38
rs759991894 Chr.14: 92798070-92798070 on Build GRCh38
rs757219754 Chr.14: 92797969-92797969 on Build GRCh38
rs777229794 Chr.14: 92798002-92798002 on Build GRCh38
rs770554079 Chr.14: 92798018-92798018 on Build GRCh38
rs751820228 Chr.14: 92797966-92797966 on Build GRCh38
rs532791576 Chr.14: 92798117-92798117 on Build GRCh38
rs564069214 Chr.14: 92798115-92798115 on Build GRCh38
rs912926947 Chr.14: 92798056-92798056 on Build GRCh38
rs901836383 Chr.14: 92798040-92798040 on Build GRCh38
rs80136374 Chr.14: 92797991-92797991 on Build GRCh38
rs947064973 Chr.14: 92798063-92798063 on Build GRCh38
rs559808338 Chr.14: 92798172-92798172 on Build GRCh38
rs753099540 Chr.14: 92798000-92798001 on Build GRCh38
rs771822697 Chr.14: 92797901-92797901 on Build GRCh38
rs958452364 Chr.14: 92798136-92798136 on Build GRCh38
rs202187684 Chr.14: 92797896-92797896 on Build GRCh38
rs746675878 Chr.14: 92798005-92798005 on Build GRCh38
rs991197866 Chr.14: 92798154-92798154 on Build GRCh38
rs759759384 Chr.14: 92797832-92797832 on Build GRCh38
rs368368226 Chr.14: 92798025-92798025 on Build GRCh38
rs755034600 Chr.14: 92797889-92797889 on Build GRCh38
rs142921167 Chr.14: 92797931-92797931 on Build GRCh38
rs370774857 Chr.14: 92797980-92797980 on Build GRCh38
rs748009930 Chr.14: 92797894-92797894 on Build GRCh38
rs763346887 Chr.14: 92797943-92797943 on Build GRCh38
rs143588478 Chr.14: 92797977-92797977 on Build GRCh38
rs561751889 Chr.14: 92797998-92797998 on Build GRCh38

View Full Product Details