Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 94446250-94446748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINA11
Entrez Gene ID
256394
Gene Name
serpin family A member 11
Gene Aliases
-
UniGene
Hs.145890
Gene Chromosome Location
Chr.14: 94442438-94452857 on Build GRCh38
Transcripts
NM_001080451.1 XM_006720105.3

SNP Detail

SNP ID Chromosome Location
rs138841070 Chr.14: 94446477-94446477 on Build GRCh38
rs769704598 Chr.14: 94446569-94446569 on Build GRCh38
rs986588653 Chr.14: 94446633-94446633 on Build GRCh38
rs115771804 Chr.14: 94446367-94446367 on Build GRCh38
rs756666965 Chr.14: 94446630-94446630 on Build GRCh38
rs764493200 Chr.14: 94446283-94446283 on Build GRCh38
rs762134550 Chr.14: 94446351-94446351 on Build GRCh38
rs571628177 Chr.14: 94446704-94446704 on Build GRCh38
rs761888389 Chr.14: 94446564-94446564 on Build GRCh38
rs774585274 Chr.14: 94446286-94446286 on Build GRCh38
rs559305199 Chr.14: 94446403-94446403 on Build GRCh38
rs754676495 Chr.14: 94446624-94446624 on Build GRCh38
rs758348433 Chr.14: 94446300-94446300 on Build GRCh38
rs1885137 Chr.14: 94446559-94446559 on Build GRCh38
rs900110609 Chr.14: 94446696-94446696 on Build GRCh38
rs369984611 Chr.14: 94446609-94446609 on Build GRCh38
rs747420042 Chr.14: 94446538-94446538 on Build GRCh38
rs749743526 Chr.14: 94446347-94446347 on Build GRCh38
rs771162722 Chr.14: 94446428-94446428 on Build GRCh38
rs763984763 Chr.14: 94446390-94446390 on Build GRCh38
rs753921372 Chr.14: 94446387-94446387 on Build GRCh38
rs755780778 Chr.14: 94446465-94446465 on Build GRCh38
rs748549168 Chr.14: 94446525-94446525 on Build GRCh38
rs914076396 Chr.14: 94446593-94446593 on Build GRCh38
rs766391996 Chr.14: 94446384-94446384 on Build GRCh38
rs1014911147 Chr.14: 94446299-94446299 on Build GRCh38
rs750223613 Chr.14: 94446590-94446590 on Build GRCh38
rs764084100 Chr.14: 94446452-94446452 on Build GRCh38
rs758280945 Chr.14: 94446389-94446389 on Build GRCh38
rs756231348 Chr.14: 94446295-94446295 on Build GRCh38
rs776902350 Chr.14: 94446560-94446560 on Build GRCh38
rs143678178 Chr.14: 94446461-94446461 on Build GRCh38
rs576980680 Chr.14: 94446416-94446416 on Build GRCh38
rs757062491 Chr.14: 94446401-94446401 on Build GRCh38
rs762056575 Chr.14: 94446289-94446289 on Build GRCh38
rs148183767 Chr.14: 94446358-94446358 on Build GRCh38
rs750494944 Chr.14: 94446293-94446293 on Build GRCh38
rs775643756 Chr.14: 94446576-94446576 on Build GRCh38
rs779669422 Chr.14: 94446414-94446414 on Build GRCh38
rs892495151 Chr.14: 94446285-94446285 on Build GRCh38
rs1001652869 Chr.14: 94446514-94446514 on Build GRCh38
rs761615166 Chr.14: 94446580-94446580 on Build GRCh38
rs778074226 Chr.14: 94446537-94446537 on Build GRCh38
rs769485876 Chr.14: 94446342-94446342 on Build GRCh38
rs778288945 Chr.14: 94446653-94446653 on Build GRCh38
rs769320189 Chr.14: 94446349-94446349 on Build GRCh38
rs12888805 Chr.14: 94446462-94446462 on Build GRCh38
rs752285581 Chr.14: 94446636-94446636 on Build GRCh38
rs371851401 Chr.14: 94446303-94446303 on Build GRCh38
rs767657264 Chr.14: 94446292-94446292 on Build GRCh38
rs374314209 Chr.14: 94446619-94446619 on Build GRCh38
rs563316230 Chr.14: 94446427-94446427 on Build GRCh38
rs758807200 Chr.14: 94446640-94446640 on Build GRCh38
rs752692786 Chr.14: 94446298-94446298 on Build GRCh38
rs765158363 Chr.14: 94446297-94446297 on Build GRCh38
rs1014498630 Chr.14: 94446542-94446542 on Build GRCh38
rs767415961 Chr.14: 94446488-94446488 on Build GRCh38
rs751334885 Chr.14: 94446392-94446392 on Build GRCh38
rs114788552 Chr.14: 94446581-94446581 on Build GRCh38
rs745366755 Chr.14: 94446336-94446336 on Build GRCh38
rs187815829 Chr.14: 94446486-94446486 on Build GRCh38
rs115117747 Chr.14: 94446315-94446315 on Build GRCh38
rs911094145 Chr.14: 94446654-94446654 on Build GRCh38
rs138198677 Chr.14: 94446432-94446432 on Build GRCh38
rs114610098 Chr.14: 94446420-94446420 on Build GRCh38
rs746799410 Chr.14: 94446310-94446310 on Build GRCh38
rs143044592 Chr.14: 94446344-94446344 on Build GRCh38
rs928480008 Chr.14: 94446470-94446470 on Build GRCh38
rs745458726 Chr.14: 94446405-94446405 on Build GRCh38
rs759648291 Chr.14: 94446443-94446443 on Build GRCh38
rs756908148 Chr.14: 94446312-94446312 on Build GRCh38
rs752461871 Chr.14: 94446602-94446603 on Build GRCh38
rs760587317 Chr.14: 94446599-94446599 on Build GRCh38
rs368468516 Chr.14: 94446426-94446426 on Build GRCh38
rs776327381 Chr.14: 94446406-94446406 on Build GRCh38
rs76197647 Chr.14: 94446629-94446629 on Build GRCh38
rs769184552 Chr.14: 94446418-94446418 on Build GRCh38
rs774729784 Chr.14: 94446350-94446350 on Build GRCh38
rs772641229 Chr.14: 94446528-94446528 on Build GRCh38
rs920677594 Chr.14: 94446338-94446338 on Build GRCh38
rs753369215 Chr.14: 94446478-94446478 on Build GRCh38
rs754513043 Chr.14: 94446480-94446480 on Build GRCh38
rs147000089 Chr.14: 94446375-94446375 on Build GRCh38

View Full Product Details