Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95184296-95184794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLMN
Entrez Gene ID
79789
Gene Name
calmin
Gene Aliases
-
UniGene
Hs.301478
Gene Chromosome Location
Chr.14: 95181939-95319908 on Build GRCh38
Transcripts
NM_024734.3 XM_011537158.1 XM_017021646.1

SNP Detail

SNP ID Chromosome Location
rs914773069 Chr.14: 95184449-95184449 on Build GRCh38
rs916336211 Chr.14: 95184587-95184587 on Build GRCh38
rs1055156561 Chr.14: 95184370-95184370 on Build GRCh38
rs1046584656 Chr.14: 95184717-95184717 on Build GRCh38
rs768650253 Chr.14: 95184650-95184650 on Build GRCh38
rs1047042341 Chr.14: 95184669-95184669 on Build GRCh38
rs1018064276 Chr.14: 95184571-95184571 on Build GRCh38
rs189356669 Chr.14: 95184558-95184558 on Build GRCh38
rs936698574 Chr.14: 95184671-95184671 on Build GRCh38
rs939035887 Chr.14: 95184574-95184574 on Build GRCh38
rs761571823 Chr.14: 95184344-95184344 on Build GRCh38
rs1025082736 Chr.14: 95184412-95184412 on Build GRCh38
rs1042815559 Chr.14: 95184327-95184327 on Build GRCh38
rs368662048 Chr.14: 95184418-95184418 on Build GRCh38
rs895259030 Chr.14: 95184698-95184698 on Build GRCh38
rs967646275 Chr.14: 95184514-95184514 on Build GRCh38
rs199567429 Chr.14: 95184329-95184330 on Build GRCh38
rs745606242 Chr.14: 95184497-95184497 on Build GRCh38
rs1013736462 Chr.14: 95184387-95184387 on Build GRCh38
rs757363700 Chr.14: 95184385-95184385 on Build GRCh38
rs371681209 Chr.14: 95184427-95184427 on Build GRCh38
rs530985991 Chr.14: 95184542-95184542 on Build GRCh38
rs994424264 Chr.14: 95184466-95184466 on Build GRCh38
rs535953200 Chr.14: 95184405-95184405 on Build GRCh38
rs1055037239 Chr.14: 95184691-95184691 on Build GRCh38
rs867366264 Chr.14: 95184663-95184663 on Build GRCh38
rs571815505 Chr.14: 95184485-95184485 on Build GRCh38
rs945835555 Chr.14: 95184631-95184631 on Build GRCh38
rs1018384032 Chr.14: 95184367-95184367 on Build GRCh38
rs553928634 Chr.14: 95184428-95184428 on Build GRCh38
rs761404154 Chr.14: 95184749-95184749 on Build GRCh38
rs913062089 Chr.14: 95184630-95184630 on Build GRCh38
rs985093474 Chr.14: 95184549-95184549 on Build GRCh38
rs897441329 Chr.14: 95184417-95184417 on Build GRCh38
rs145338567 Chr.14: 95184509-95184509 on Build GRCh38
rs551746666 Chr.14: 95184368-95184368 on Build GRCh38
rs749207859 Chr.14: 95184618-95184618 on Build GRCh38
rs549918878 Chr.14: 95184695-95184695 on Build GRCh38
rs558763349 Chr.14: 95184575-95184575 on Build GRCh38
rs895206990 Chr.14: 95184386-95184386 on Build GRCh38
rs1027618280 Chr.14: 95184474-95184474 on Build GRCh38
rs903861118 Chr.14: 95184340-95184340 on Build GRCh38
rs898362728 Chr.14: 95184645-95184645 on Build GRCh38
rs897331717 Chr.14: 95184735-95184735 on Build GRCh38
rs568328173 Chr.14: 95184381-95184381 on Build GRCh38
rs549784524 Chr.14: 95184376-95184376 on Build GRCh38
rs61217816 Chr.14: 95184710-95184710 on Build GRCh38

View Full Product Details