Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95185613-95186107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLMN
Entrez Gene ID
79789
Gene Name
calmin
Gene Aliases
-
UniGene
Hs.301478
Gene Chromosome Location
Chr.14: 95181939-95319908 on Build GRCh38
Transcripts
NM_024734.3 XM_011537158.1 XM_017021646.1

SNP Detail

SNP ID Chromosome Location
rs971267542 Chr.14: 95185910-95185910 on Build GRCh38
rs140298942 Chr.14: 95185698-95185698 on Build GRCh38
rs1024202426 Chr.14: 95185894-95185894 on Build GRCh38
rs151287769 Chr.14: 95185645-95185645 on Build GRCh38
rs562280419 Chr.14: 95185729-95185729 on Build GRCh38
rs965170227 Chr.14: 95186032-95186032 on Build GRCh38
rs778848757 Chr.14: 95185946-95185946 on Build GRCh38
rs182034420 Chr.14: 95185787-95185787 on Build GRCh38
rs942806222 Chr.14: 95185648-95185648 on Build GRCh38
rs541493341 Chr.14: 95185636-95185636 on Build GRCh38
rs536881617 Chr.14: 95186005-95186005 on Build GRCh38
rs1032730281 Chr.14: 95185839-95185839 on Build GRCh38
rs543605692 Chr.14: 95185728-95185728 on Build GRCh38
rs1027062740 Chr.14: 95185984-95185984 on Build GRCh38
rs191698759 Chr.14: 95185949-95185949 on Build GRCh38
rs933674107 Chr.14: 95185719-95185719 on Build GRCh38
rs796139344 Chr.14: 95185761-95185761 on Build GRCh38
rs371696300 Chr.14: 95185774-95185774 on Build GRCh38
rs369758817 Chr.14: 95186021-95186021 on Build GRCh38
rs749089252 Chr.14: 95185895-95185895 on Build GRCh38
rs577781526 Chr.14: 95185843-95185843 on Build GRCh38
rs185801710 Chr.14: 95185867-95185867 on Build GRCh38
rs576180797 Chr.14: 95185725-95185725 on Build GRCh38
rs975662283 Chr.14: 95186043-95186043 on Build GRCh38
rs1003949396 Chr.14: 95185996-95185996 on Build GRCh38
rs909944200 Chr.14: 95186000-95186000 on Build GRCh38
rs1057174089 Chr.14: 95185875-95185875 on Build GRCh38
rs1012682178 Chr.14: 95185892-95185892 on Build GRCh38
rs114567749 Chr.14: 95185929-95185929 on Build GRCh38
rs984613309 Chr.14: 95185985-95185985 on Build GRCh38
rs559339905 Chr.14: 95185849-95185849 on Build GRCh38
rs756841434 Chr.14: 95185724-95185727 on Build GRCh38
rs965233638 Chr.14: 95185716-95185716 on Build GRCh38
rs895959899 Chr.14: 95185886-95185886 on Build GRCh38
rs529569819 Chr.14: 95185779-95185779 on Build GRCh38
rs183158282 Chr.14: 95185959-95185959 on Build GRCh38
rs995133704 Chr.14: 95185973-95185973 on Build GRCh38
rs975566725 Chr.14: 95186060-95186060 on Build GRCh38
rs527821236 Chr.14: 95185840-95185840 on Build GRCh38
rs8005908 Chr.14: 95185788-95185788 on Build GRCh38
rs571827340 Chr.14: 95185653-95185653 on Build GRCh38
rs951049098 Chr.14: 95185988-95185988 on Build GRCh38
rs750633788 Chr.14: 95185753-95185753 on Build GRCh38
rs564348497 Chr.14: 95185702-95185702 on Build GRCh38
rs951378316 Chr.14: 95185947-95185947 on Build GRCh38
rs547496640 Chr.14: 95185786-95185786 on Build GRCh38

View Full Product Details