Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95186967-95187462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLMN
Entrez Gene ID
79789
Gene Name
calmin
Gene Aliases
-
UniGene
Hs.301478
Gene Chromosome Location
Chr.14: 95181939-95319908 on Build GRCh38
Transcripts
NM_024734.3 XM_011537158.1 XM_017021646.1

SNP Detail

SNP ID Chromosome Location
rs767915771 Chr.14: 95187163-95187163 on Build GRCh38
rs905786597 Chr.14: 95187133-95187133 on Build GRCh38
rs150085800 Chr.14: 95187207-95187207 on Build GRCh38
rs909048668 Chr.14: 95187096-95187096 on Build GRCh38
rs796849237 Chr.14: 95187225-95187225 on Build GRCh38
rs116794212 Chr.14: 95187363-95187363 on Build GRCh38
rs553583843 Chr.14: 95187416-95187416 on Build GRCh38
rs745556077 Chr.14: 95187014-95187014 on Build GRCh38
rs766548212 Chr.14: 95187204-95187204 on Build GRCh38
rs764958123 Chr.14: 95187268-95187268 on Build GRCh38
rs1032141346 Chr.14: 95187198-95187198 on Build GRCh38
rs1023198414 Chr.14: 95187220-95187220 on Build GRCh38
rs551226354 Chr.14: 95186991-95186991 on Build GRCh38
rs188582267 Chr.14: 95187244-95187244 on Build GRCh38
rs980115105 Chr.14: 95187279-95187279 on Build GRCh38
rs1052829802 Chr.14: 95187035-95187035 on Build GRCh38
rs192485502 Chr.14: 95187100-95187100 on Build GRCh38
rs1035359918 Chr.14: 95187292-95187292 on Build GRCh38
rs937263898 Chr.14: 95187424-95187424 on Build GRCh38
rs983729170 Chr.14: 95187018-95187018 on Build GRCh38
rs908925634 Chr.14: 95187346-95187346 on Build GRCh38
rs369328901 Chr.14: 95187152-95187152 on Build GRCh38
rs949851095 Chr.14: 95187393-95187393 on Build GRCh38
rs893319998 Chr.14: 95187206-95187206 on Build GRCh38
rs1046018560 Chr.14: 95187111-95187111 on Build GRCh38
rs529133219 Chr.14: 95187151-95187151 on Build GRCh38
rs922128012 Chr.14: 95187257-95187257 on Build GRCh38
rs550978287 Chr.14: 95187233-95187233 on Build GRCh38
rs536750375 Chr.14: 95187313-95187313 on Build GRCh38
rs773287012 Chr.14: 95187170-95187170 on Build GRCh38
rs774636011 Chr.14: 95187120-95187120 on Build GRCh38
rs752503680 Chr.14: 95187353-95187353 on Build GRCh38
rs953651317 Chr.14: 95187273-95187273 on Build GRCh38
rs1038981159 Chr.14: 95187106-95187106 on Build GRCh38
rs983227640 Chr.14: 95187317-95187317 on Build GRCh38
rs185058061 Chr.14: 95187181-95187181 on Build GRCh38
rs578255136 Chr.14: 95187433-95187433 on Build GRCh38
rs192952129 Chr.14: 95187322-95187322 on Build GRCh38
rs950394229 Chr.14: 95187299-95187299 on Build GRCh38
rs1052079697 Chr.14: 95187419-95187419 on Build GRCh38
rs567242400 Chr.14: 95187359-95187359 on Build GRCh38
rs1045568137 Chr.14: 95187405-95187405 on Build GRCh38
rs377126784 Chr.14: 95187002-95187002 on Build GRCh38

View Full Product Details