Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 100126597-100127000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVL
Entrez Gene ID
51466
Gene Name
Enah/Vasp-like
Gene Aliases
RNB6
UniGene
Hs.125867
Gene Chromosome Location
Chr.14: 99971454-100144236 on Build GRCh38
Transcripts
NM_016337.2 XM_005267749.3 XM_011536828.2 XM_017021363.1 XM_017021364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6296935 c.447G>T p.Q149H Chr.14: 100126731-100126731 on Build GRCh38
1371961 c.452G>A p.R151H Chr.14: 100126736-100126736 on Build GRCh38
4681930 c.451C>T p.R151C Chr.14: 100126735-100126735 on Build GRCh38
6656927 c.466G>T p.E156* Chr.14: 100126750-100126750 on Build GRCh38
7118549 c.466G>A p.E156K Chr.14: 100126750-100126750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774275585 Chr.14: 100126800-100126800 on Build GRCh38
rs375861143 Chr.14: 100126674-100126674 on Build GRCh38
rs539514514 Chr.14: 100126637-100126637 on Build GRCh38
rs751732289 Chr.14: 100126711-100126711 on Build GRCh38
rs147474356 Chr.14: 100126796-100126796 on Build GRCh38
rs756579670 Chr.14: 100126756-100126756 on Build GRCh38
rs574394605 Chr.14: 100126694-100126694 on Build GRCh38
rs866165251 Chr.14: 100126703-100126703 on Build GRCh38
rs1032286692 Chr.14: 100126805-100126807 on Build GRCh38
rs752870060 Chr.14: 100126827-100126827 on Build GRCh38
rs980149686 Chr.14: 100126832-100126832 on Build GRCh38
rs781617821 Chr.14: 100126733-100126733 on Build GRCh38
rs771680079 Chr.14: 100126785-100126785 on Build GRCh38
rs369210011 Chr.14: 100126774-100126774 on Build GRCh38
rs764652252 Chr.14: 100126860-100126860 on Build GRCh38
rs1053334145 Chr.14: 100126950-100126950 on Build GRCh38
rs879751390 Chr.14: 100126621-100126621 on Build GRCh38
rs370415020 Chr.14: 100126823-100126823 on Build GRCh38
rs1042245226 Chr.14: 100126628-100126628 on Build GRCh38
rs548978855 Chr.14: 100126861-100126861 on Build GRCh38
rs1041110271 Chr.14: 100126631-100126631 on Build GRCh38
rs767809844 Chr.14: 100126803-100126803 on Build GRCh38
rs927272322 Chr.14: 100126833-100126833 on Build GRCh38
rs745719022 Chr.14: 100126660-100126660 on Build GRCh38
rs759390194 Chr.14: 100126797-100126797 on Build GRCh38
rs1042604222 Chr.14: 100126866-100126866 on Build GRCh38
rs372980425 Chr.14: 100126802-100126802 on Build GRCh38
rs999548271 Chr.14: 100126947-100126947 on Build GRCh38
rs181270472 Chr.14: 100126666-100126666 on Build GRCh38
rs750787699 Chr.14: 100126736-100126736 on Build GRCh38
rs369045038 Chr.14: 100126653-100126653 on Build GRCh38
rs771853742 Chr.14: 100126725-100126726 on Build GRCh38
rs994606826 Chr.14: 100126946-100126946 on Build GRCh38
rs370245219 Chr.14: 100126820-100126820 on Build GRCh38
rs750697658 Chr.14: 100126808-100126808 on Build GRCh38
rs529219805 Chr.14: 100126859-100126859 on Build GRCh38
rs944517805 Chr.14: 100126885-100126885 on Build GRCh38
rs115783985 Chr.14: 100126965-100126965 on Build GRCh38
rs756116377 Chr.14: 100126856-100126856 on Build GRCh38
rs765279757 Chr.14: 100126799-100126799 on Build GRCh38
rs753737757 Chr.14: 100126681-100126681 on Build GRCh38
rs1043063007 Chr.14: 100126925-100126925 on Build GRCh38
rs759831187 Chr.14: 100126764-100126764 on Build GRCh38
rs726514 Chr.14: 100126622-100126622 on Build GRCh38
rs994577400 Chr.14: 100126767-100126767 on Build GRCh38
rs1048805096 Chr.14: 100126955-100126955 on Build GRCh38
rs780562130 Chr.14: 100126763-100126763 on Build GRCh38
rs900142809 Chr.14: 100126923-100126923 on Build GRCh38
rs772878970 Chr.14: 100126787-100126787 on Build GRCh38
rs541005126 Chr.14: 100126809-100126809 on Build GRCh38
rs1041120304 Chr.14: 100126886-100126886 on Build GRCh38
rs376191873 Chr.14: 100126834-100126834 on Build GRCh38
rs746755774 Chr.14: 100126794-100126794 on Build GRCh38
rs1028151831 Chr.14: 100126634-100126634 on Build GRCh38
rs757541847 Chr.14: 100126727-100126727 on Build GRCh38
rs990168425 Chr.14: 100126855-100126855 on Build GRCh38
rs550242076 Chr.14: 100126824-100126824 on Build GRCh38
rs764089208 Chr.14: 100126702-100126702 on Build GRCh38
rs556025068 Chr.14: 100126659-100126659 on Build GRCh38
rs770721493 Chr.14: 100126795-100126795 on Build GRCh38
rs900275645 Chr.14: 100126757-100126757 on Build GRCh38
rs904510870 Chr.14: 100126929-100126929 on Build GRCh38
rs186705512 Chr.14: 100126810-100126810 on Build GRCh38
rs945176570 Chr.14: 100126862-100126862 on Build GRCh38
rs1008598694 Chr.14: 100126738-100126738 on Build GRCh38
rs2295857 Chr.14: 100126876-100126876 on Build GRCh38
rs535304267 Chr.14: 100126922-100126922 on Build GRCh38
rs944604735 Chr.14: 100126906-100126906 on Build GRCh38

View Full Product Details