Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105354943-105355444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PACS2
Entrez Gene ID
23241
Gene Name
phosphofurin acidic cluster sorting protein 2
Gene Aliases
PACS-2, PACS1L
UniGene
Hs.525626
Gene Chromosome Location
Chr.14: 105300811-105398147 on Build GRCh38
Transcripts
NM_001100913.2 NM_001243127.2 NM_015197.3 XM_006720090.1 XM_006720091.1 XM_006720092.2 XM_017021105.1 XM_017021106.1 XM_017021107.1 XM_017021108.1 XM_017021109.1 XM_017021110.1 XM_017021111.1 XM_017021112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5362529 c.401C>T p.A134V Chr.14: 105355155-105355155 on Build GRCh38
6736754 c.401C>T p.A134V Chr.14: 105355155-105355155 on Build GRCh38
5813738 c.302C>G p.P101R Chr.14: 105355056-105355056 on Build GRCh38
5813737 c.302C>G p.P101R Chr.14: 105355056-105355056 on Build GRCh38
5381971 c.378C>T p.I126I Chr.14: 105355132-105355132 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782203340 Chr.14: 105355074-105355074 on Build GRCh38
rs782228619 Chr.14: 105355221-105355221 on Build GRCh38
rs782063559 Chr.14: 105355093-105355093 on Build GRCh38
rs587629263 Chr.14: 105355248-105355248 on Build GRCh38
rs372618094 Chr.14: 105355035-105355035 on Build GRCh38
rs73359061 Chr.14: 105355342-105355342 on Build GRCh38
rs782525530 Chr.14: 105355002-105355002 on Build GRCh38
rs781949430 Chr.14: 105355208-105355208 on Build GRCh38
rs782292382 Chr.14: 105355204-105355204 on Build GRCh38
rs781857652 Chr.14: 105355314-105355314 on Build GRCh38
rs782452889 Chr.14: 105355019-105355019 on Build GRCh38
rs781876422 Chr.14: 105355009-105355009 on Build GRCh38
rs782524953 Chr.14: 105355324-105355324 on Build GRCh38
rs782050259 Chr.14: 105355146-105355146 on Build GRCh38
rs941994870 Chr.14: 105355278-105355278 on Build GRCh38
rs115631676 Chr.14: 105355397-105355397 on Build GRCh38
rs781793403 Chr.14: 105355014-105355014 on Build GRCh38
rs782308429 Chr.14: 105355222-105355222 on Build GRCh38
rs782672586 Chr.14: 105355197-105355197 on Build GRCh38
rs377453480 Chr.14: 105355155-105355155 on Build GRCh38
rs112557358 Chr.14: 105355050-105355050 on Build GRCh38
rs946212996 Chr.14: 105355343-105355343 on Build GRCh38
rs587648417 Chr.14: 105355283-105355283 on Build GRCh38
rs367940917 Chr.14: 105355018-105355018 on Build GRCh38
rs61738367 Chr.14: 105355120-105355120 on Build GRCh38
rs149722060 Chr.14: 105355156-105355156 on Build GRCh38
rs587682503 Chr.14: 105355351-105355351 on Build GRCh38
rs782456234 Chr.14: 105355412-105355412 on Build GRCh38
rs950582159 Chr.14: 105355254-105355254 on Build GRCh38
rs587593663 Chr.14: 105355273-105355273 on Build GRCh38
rs1000503163 Chr.14: 105355375-105355375 on Build GRCh38
rs983470586 Chr.14: 105355264-105355264 on Build GRCh38
rs782585185 Chr.14: 105355415-105355415 on Build GRCh38
rs938930899 Chr.14: 105355318-105355318 on Build GRCh38
rs782135452 Chr.14: 105355113-105355113 on Build GRCh38
rs979333012 Chr.14: 105354990-105354990 on Build GRCh38
rs782612408 Chr.14: 105355013-105355013 on Build GRCh38
rs909204022 Chr.14: 105355072-105355072 on Build GRCh38
rs782287967 Chr.14: 105355009-105355010 on Build GRCh38
rs782368472 Chr.14: 105355051-105355051 on Build GRCh38
rs111332106 Chr.14: 105355179-105355179 on Build GRCh38
rs74090510 Chr.14: 105355418-105355418 on Build GRCh38
rs782352784 Chr.14: 105355075-105355075 on Build GRCh38
rs781930426 Chr.14: 105355223-105355223 on Build GRCh38
rs1003006997 Chr.14: 105355132-105355132 on Build GRCh38
rs73359063 Chr.14: 105355380-105355380 on Build GRCh38
rs146129304 Chr.14: 105355063-105355063 on Build GRCh38
rs782483501 Chr.14: 105355012-105355012 on Build GRCh38
rs781916206 Chr.14: 105355080-105355080 on Build GRCh38
rs1030261474 Chr.14: 105355398-105355398 on Build GRCh38
rs587688204 Chr.14: 105354983-105354983 on Build GRCh38
rs782318213 Chr.14: 105355196-105355196 on Build GRCh38
rs77365937 Chr.14: 105355237-105355237 on Build GRCh38
rs782375596 Chr.14: 105355207-105355207 on Build GRCh38
rs950531069 Chr.14: 105355411-105355411 on Build GRCh38
rs782303681 Chr.14: 105355373-105355373 on Build GRCh38
rs900187542 Chr.14: 105355381-105355381 on Build GRCh38
rs964553763 Chr.14: 105355002-105355002 on Build GRCh38
rs587659752 Chr.14: 105355413-105355413 on Build GRCh38
rs980515955 Chr.14: 105355286-105355286 on Build GRCh38
rs910473634 Chr.14: 105354997-105354997 on Build GRCh38
rs782677114 Chr.14: 105355036-105355036 on Build GRCh38
rs199643022 Chr.14: 105355028-105355028 on Build GRCh38
rs587717046 Chr.14: 105355265-105355265 on Build GRCh38
rs369984475 Chr.14: 105355195-105355195 on Build GRCh38
rs747435170 Chr.14: 105355240-105355240 on Build GRCh38
rs903608361 Chr.14: 105355369-105355369 on Build GRCh38
rs587729650 Chr.14: 105355339-105355339 on Build GRCh38
rs191402536 Chr.14: 105355416-105355416 on Build GRCh38
rs782569931 Chr.14: 105355153-105355153 on Build GRCh38
rs782405230 Chr.14: 105355096-105355096 on Build GRCh38
rs111940627 Chr.14: 105355005-105355005 on Build GRCh38
rs142209904 Chr.14: 105355102-105355102 on Build GRCh38
rs967679821 Chr.14: 105355205-105355205 on Build GRCh38
rs587691625 Chr.14: 105355189-105355189 on Build GRCh38
rs782466397 Chr.14: 105355188-105355188 on Build GRCh38
rs782345293 Chr.14: 105355135-105355135 on Build GRCh38

View Full Product Details