Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105368343-105368834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PACS2
Entrez Gene ID
23241
Gene Name
phosphofurin acidic cluster sorting protein 2
Gene Aliases
PACS-2, PACS1L
UniGene
Hs.525626
Gene Chromosome Location
Chr.14: 105300811-105398147 on Build GRCh38
Transcripts
NM_001100913.2 NM_001243127.2 NM_015197.3 XM_006720090.1 XM_006720091.1 XM_006720092.2 XM_017021105.1 XM_017021106.1 XM_017021107.1 XM_017021108.1 XM_017021109.1 XM_017021110.1 XM_017021111.1 XM_017021112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3711433 c.670G>A p.E224K Chr.14: 105368468-105368468 on Build GRCh38
4049551 c.706C>T p.R236W Chr.14: 105368504-105368504 on Build GRCh38
5828073 c.686T>C p.V229A Chr.14: 105368484-105368484 on Build GRCh38
5832738 c.690G>A p.G230G Chr.14: 105368488-105368488 on Build GRCh38
5923088 c.731C>T p.S244F Chr.14: 105368529-105368529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782151393 Chr.14: 105368504-105368504 on Build GRCh38
rs782340011 Chr.14: 105368435-105368435 on Build GRCh38
rs782561685 Chr.14: 105368416-105368416 on Build GRCh38
rs781966753 Chr.14: 105368438-105368438 on Build GRCh38
rs371973034 Chr.14: 105368666-105368666 on Build GRCh38
rs375982832 Chr.14: 105368449-105368449 on Build GRCh38
rs781878450 Chr.14: 105368473-105368473 on Build GRCh38
rs782716798 Chr.14: 105368615-105368615 on Build GRCh38
rs782422342 Chr.14: 105368428-105368428 on Build GRCh38
rs968632513 Chr.14: 105368737-105368737 on Build GRCh38
rs369363280 Chr.14: 105368474-105368474 on Build GRCh38
rs1009902584 Chr.14: 105368700-105368700 on Build GRCh38
rs781951709 Chr.14: 105368571-105368571 on Build GRCh38
rs587652611 Chr.14: 105368431-105368431 on Build GRCh38
rs779120119 Chr.14: 105368754-105368754 on Build GRCh38
rs911066522 Chr.14: 105368597-105368597 on Build GRCh38
rs368127909 Chr.14: 105368471-105368471 on Build GRCh38
rs587723997 Chr.14: 105368607-105368607 on Build GRCh38
rs965222107 Chr.14: 105368770-105368770 on Build GRCh38
rs587646885 Chr.14: 105368395-105368395 on Build GRCh38
rs782248654 Chr.14: 105368566-105368566 on Build GRCh38
rs587684556 Chr.14: 105368559-105368559 on Build GRCh38
rs376520036 Chr.14: 105368569-105368569 on Build GRCh38
rs141216726 Chr.14: 105368386-105368386 on Build GRCh38
rs782333563 Chr.14: 105368513-105368513 on Build GRCh38
rs781791295 Chr.14: 105368482-105368482 on Build GRCh38
rs782422954 Chr.14: 105368505-105368505 on Build GRCh38
rs782690320 Chr.14: 105368484-105368484 on Build GRCh38
rs782061496 Chr.14: 105368789-105368789 on Build GRCh38
rs782363413 Chr.14: 105368579-105368579 on Build GRCh38
rs782227644 Chr.14: 105368498-105368498 on Build GRCh38
rs782484100 Chr.14: 105368537-105368537 on Build GRCh38
rs587610730 Chr.14: 105368488-105368488 on Build GRCh38
rs145923328 Chr.14: 105368511-105368511 on Build GRCh38
rs780880602 Chr.14: 105368523-105368523 on Build GRCh38
rs587770602 Chr.14: 105368788-105368788 on Build GRCh38
rs909880730 Chr.14: 105368806-105368806 on Build GRCh38
rs888726495 Chr.14: 105368446-105368446 on Build GRCh38
rs782722006 Chr.14: 105368456-105368456 on Build GRCh38
rs782742051 Chr.14: 105368723-105368723 on Build GRCh38
rs911307386 Chr.14: 105368387-105368387 on Build GRCh38
rs914365988 Chr.14: 105368371-105368371 on Build GRCh38
rs782607701 Chr.14: 105368536-105368536 on Build GRCh38
rs587729444 Chr.14: 105368530-105368530 on Build GRCh38
rs984127819 Chr.14: 105368801-105368801 on Build GRCh38
rs929982891 Chr.14: 105368610-105368610 on Build GRCh38
rs781891896 Chr.14: 105368461-105368461 on Build GRCh38
rs948289172 Chr.14: 105368810-105368810 on Build GRCh38
rs782299087 Chr.14: 105368501-105368501 on Build GRCh38
rs782039359 Chr.14: 105368441-105368441 on Build GRCh38
rs587633704 Chr.14: 105368592-105368592 on Build GRCh38
rs781821323 Chr.14: 105368532-105368532 on Build GRCh38
rs782130739 Chr.14: 105368432-105368432 on Build GRCh38
rs782354768 Chr.14: 105368608-105368608 on Build GRCh38
rs1021743124 Chr.14: 105368706-105368706 on Build GRCh38
rs8016693 Chr.14: 105368410-105368410 on Build GRCh38
rs782139923 Chr.14: 105368512-105368512 on Build GRCh38
rs782235019 Chr.14: 105368574-105368574 on Build GRCh38
rs138129795 Chr.14: 105368462-105368462 on Build GRCh38
rs587705831 Chr.14: 105368483-105368483 on Build GRCh38
rs782188439 Chr.14: 105368480-105368480 on Build GRCh38
rs587625642 Chr.14: 105368554-105368554 on Build GRCh38
rs782084327 Chr.14: 105368688-105368688 on Build GRCh38
rs587670629 Chr.14: 105368602-105368602 on Build GRCh38
rs782769811 Chr.14: 105368467-105368467 on Build GRCh38
rs587633912 Chr.14: 105368631-105368631 on Build GRCh38
rs782279270 Chr.14: 105368427-105368427 on Build GRCh38
rs200023287 Chr.14: 105368415-105368415 on Build GRCh38
rs948336134 Chr.14: 105368618-105368618 on Build GRCh38
rs113565722 Chr.14: 105368563-105368563 on Build GRCh38
rs1006815422 Chr.14: 105368744-105368744 on Build GRCh38
rs191005930 Chr.14: 105368584-105368584 on Build GRCh38
rs782389056 Chr.14: 105368493-105368493 on Build GRCh38
rs138765820 Chr.14: 105368798-105368798 on Build GRCh38
rs782734762 Chr.14: 105368516-105368516 on Build GRCh38
rs782542116 Chr.14: 105368381-105368381 on Build GRCh38
rs782263660 Chr.14: 105368402-105368403 on Build GRCh38
rs781928681 Chr.14: 105368502-105368502 on Build GRCh38
rs1048900082 Chr.14: 105368612-105368612 on Build GRCh38
rs782594693 Chr.14: 105368494-105368494 on Build GRCh38
rs756130055 Chr.14: 105368634-105368634 on Build GRCh38
rs782768000 Chr.14: 105368531-105368531 on Build GRCh38
rs947253074 Chr.14: 105368375-105368375 on Build GRCh38
rs918663455 Chr.14: 105368796-105368796 on Build GRCh38
rs782173977 Chr.14: 105368466-105368466 on Build GRCh38
rs781846060 Chr.14: 105368524-105368524 on Build GRCh38

View Full Product Details