Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 27755155-27755664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OCA2
Entrez Gene ID
4948
Gene Name
OCA2 melanosomal transmembrane protein
Gene Aliases
BEY, BEY1, BEY2, BOCA, D15S12, EYCL, EYCL2, EYCL3, HCL3, P, PED, SHEP1
UniGene
Hs.654411
Gene Chromosome Location
Chr.15: 27719008-28099342 on Build GRCh38
Transcripts
NM_000275.2 NM_001300984.1 XM_011521640.2 XM_017022255.1 XM_017022256.1 XM_017022257.1 XM_017022258.1 XM_017022259.1 XM_017022260.1 XM_017022261.1 XM_017022262.1 XM_017022263.1 XM_017022264.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6554820 c.2472G>A p.G824G Chr.15: 27755433-27755433 on Build GRCh38
5780002 c.2500G>T p.V834L Chr.15: 27755405-27755405 on Build GRCh38
960070 c.2463C>T p.C821C Chr.15: 27755442-27755442 on Build GRCh38
6076996 c.2460C>A p.S820S Chr.15: 27755445-27755445 on Build GRCh38
4950912 c.2438delG p.G813fs*4 Chr.15: 27755467-27755467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773448027 Chr.15: 27755532-27755532 on Build GRCh38
rs377766646 Chr.15: 27755468-27755468 on Build GRCh38
rs138228425 Chr.15: 27755387-27755387 on Build GRCh38
rs748548797 Chr.15: 27755485-27755485 on Build GRCh38
rs943921180 Chr.15: 27755531-27755531 on Build GRCh38
rs748456675 Chr.15: 27755384-27755384 on Build GRCh38
rs759396151 Chr.15: 27755404-27755404 on Build GRCh38
rs779798260 Chr.15: 27755250-27755250 on Build GRCh38
rs747222770 Chr.15: 27755506-27755506 on Build GRCh38
rs745946087 Chr.15: 27755524-27755524 on Build GRCh38
rs760853582 Chr.15: 27755395-27755395 on Build GRCh38
rs554977696 Chr.15: 27755325-27755325 on Build GRCh38
rs899553307 Chr.15: 27755539-27755539 on Build GRCh38
rs776587604 Chr.15: 27755515-27755515 on Build GRCh38
rs183101063 Chr.15: 27755414-27755414 on Build GRCh38
rs927120512 Chr.15: 27755243-27755243 on Build GRCh38
rs751149175 Chr.15: 27755344-27755344 on Build GRCh38
rs373301562 Chr.15: 27755391-27755391 on Build GRCh38
rs962511400 Chr.15: 27755584-27755584 on Build GRCh38
rs1026578659 Chr.15: 27755560-27755560 on Build GRCh38
rs530544648 Chr.15: 27755565-27755565 on Build GRCh38
rs762838711 Chr.15: 27755410-27755410 on Build GRCh38
rs756692515 Chr.15: 27755346-27755346 on Build GRCh38
rs369189827 Chr.15: 27755367-27755367 on Build GRCh38
rs929615250 Chr.15: 27755292-27755292 on Build GRCh38
rs763881975 Chr.15: 27755432-27755432 on Build GRCh38
rs751440917 Chr.15: 27755399-27755401 on Build GRCh38
rs971229767 Chr.15: 27755240-27755240 on Build GRCh38
rs7175266 Chr.15: 27755338-27755338 on Build GRCh38
rs192734230 Chr.15: 27755334-27755334 on Build GRCh38
rs182290687 Chr.15: 27755222-27755222 on Build GRCh38
rs912331573 Chr.15: 27755318-27755318 on Build GRCh38
rs1033427242 Chr.15: 27755206-27755206 on Build GRCh38
rs765257992 Chr.15: 27755187-27755187 on Build GRCh38
rs947269935 Chr.15: 27755409-27755409 on Build GRCh38
rs370954296 Chr.15: 27755491-27755491 on Build GRCh38
rs937351907 Chr.15: 27755221-27755221 on Build GRCh38
rs756885633 Chr.15: 27755443-27755443 on Build GRCh38
rs200396611 Chr.15: 27755408-27755408 on Build GRCh38
rs187972612 Chr.15: 27755595-27755595 on Build GRCh38
rs981566424 Chr.15: 27755192-27755192 on Build GRCh38
rs1019388795 Chr.15: 27755307-27755307 on Build GRCh38
rs200842913 Chr.15: 27755498-27755498 on Build GRCh38
rs770934507 Chr.15: 27755399-27755399 on Build GRCh38
rs745957691 Chr.15: 27755636-27755636 on Build GRCh38
rs958750991 Chr.15: 27755601-27755601 on Build GRCh38
rs1006865482 Chr.15: 27755581-27755581 on Build GRCh38
rs374239058 Chr.15: 27755511-27755511 on Build GRCh38
rs766970716 Chr.15: 27755459-27755459 on Build GRCh38
rs991591068 Chr.15: 27755602-27755602 on Build GRCh38
rs919625825 Chr.15: 27755266-27755266 on Build GRCh38
rs200918662 Chr.15: 27755411-27755411 on Build GRCh38
rs201569748 Chr.15: 27755403-27755403 on Build GRCh38
rs779382711 Chr.15: 27755472-27755472 on Build GRCh38
rs780660298 Chr.15: 27755348-27755348 on Build GRCh38
rs759428753 Chr.15: 27755181-27755181 on Build GRCh38
rs903036089 Chr.15: 27755293-27755293 on Build GRCh38
rs749886695 Chr.15: 27755461-27755461 on Build GRCh38
rs1020315849 Chr.15: 27755235-27755235 on Build GRCh38
rs899500978 Chr.15: 27755308-27755309 on Build GRCh38
rs1035869944 Chr.15: 27755591-27755591 on Build GRCh38
rs766291945 Chr.15: 27755351-27755371 on Build GRCh38
rs761820687 Chr.15: 27755230-27755231 on Build GRCh38
rs200732154 Chr.15: 27755380-27755380 on Build GRCh38
rs187757375 Chr.15: 27755301-27755301 on Build GRCh38
rs769170049 Chr.15: 27755368-27755368 on Build GRCh38
rs140405700 Chr.15: 27755582-27755582 on Build GRCh38
rs370691529 Chr.15: 27755259-27755259 on Build GRCh38
rs914559172 Chr.15: 27755609-27755609 on Build GRCh38
rs981260733 Chr.15: 27755241-27755241 on Build GRCh38
rs996559403 Chr.15: 27755547-27755547 on Build GRCh38
rs982457184 Chr.15: 27755587-27755587 on Build GRCh38
rs751790611 Chr.15: 27755377-27755377 on Build GRCh38
rs530046601 Chr.15: 27755259-27755260 on Build GRCh38
rs957363734 Chr.15: 27755207-27755207 on Build GRCh38
rs751149703 Chr.15: 27755433-27755433 on Build GRCh38
rs374736630 Chr.15: 27755215-27755215 on Build GRCh38
rs34704703 Chr.15: 27755405-27755405 on Build GRCh38

View Full Product Details