Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34233802-34234297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC12A6
Entrez Gene ID
9990
Gene Name
solute carrier family 12 member 6
Gene Aliases
ACCPN, KCC3, KCC3A, KCC3B
UniGene
Hs.510939
Gene Chromosome Location
Chr.15: 34229996-34338064 on Build GRCh38
Transcripts
NM_001042494.1 NM_001042495.1 NM_001042496.1 NM_001042497.1 NM_005135.2 NM_133647.1 XM_006720793.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4425411 c.3329T>C p.L1110P Chr.15: 34233960-34233960 on Build GRCh38
6779462 c.3234C>T p.T1078T Chr.15: 34233947-34233947 on Build GRCh38
6779460 c.3360C>T p.T1120T Chr.15: 34233947-34233947 on Build GRCh38
4425412 c.3221T>C p.L1074P Chr.15: 34233960-34233960 on Build GRCh38
4425410 c.3347T>C p.L1116P Chr.15: 34233960-34233960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969900949 Chr.15: 34233843-34233843 on Build GRCh38
rs948677234 Chr.15: 34234140-34234140 on Build GRCh38
rs184521227 Chr.15: 34233880-34233880 on Build GRCh38
rs199625435 Chr.15: 34233834-34233834 on Build GRCh38
rs980027158 Chr.15: 34234175-34234175 on Build GRCh38
rs901390420 Chr.15: 34234177-34234177 on Build GRCh38
rs1039852702 Chr.15: 34234084-34234085 on Build GRCh38
rs767632019 Chr.15: 34233856-34233856 on Build GRCh38
rs988019151 Chr.15: 34233946-34233946 on Build GRCh38
rs753913744 Chr.15: 34233901-34233901 on Build GRCh38
rs764468587 Chr.15: 34234016-34234016 on Build GRCh38
rs770531965 Chr.15: 34233981-34233981 on Build GRCh38
rs34987939 Chr.15: 34233825-34233826 on Build GRCh38
rs758555514 Chr.15: 34234022-34234022 on Build GRCh38
rs745436661 Chr.15: 34233956-34233956 on Build GRCh38
rs376096845 Chr.15: 34234137-34234137 on Build GRCh38
rs914402815 Chr.15: 34234092-34234092 on Build GRCh38
rs755589866 Chr.15: 34234189-34234190 on Build GRCh38
rs913830616 Chr.15: 34233950-34233950 on Build GRCh38
rs762937772 Chr.15: 34234007-34234007 on Build GRCh38
rs372202037 Chr.15: 34233865-34233865 on Build GRCh38
rs779083807 Chr.15: 34233930-34233930 on Build GRCh38
rs1005570169 Chr.15: 34234261-34234261 on Build GRCh38
rs606231229 Chr.15: 34233934-34233934 on Build GRCh38
rs887340251 Chr.15: 34234228-34234229 on Build GRCh38
rs758754831 Chr.15: 34233933-34233933 on Build GRCh38
rs764043055 Chr.15: 34234005-34234005 on Build GRCh38
rs554416561 Chr.15: 34234120-34234120 on Build GRCh38
rs765508345 Chr.15: 34234015-34234015 on Build GRCh38
rs750825269 Chr.15: 34233932-34233932 on Build GRCh38
rs757443429 Chr.15: 34233911-34233911 on Build GRCh38
rs769137207 Chr.15: 34233944-34233944 on Build GRCh38
rs753914034 Chr.15: 34234009-34234009 on Build GRCh38
rs117864565 Chr.15: 34233997-34233997 on Build GRCh38
rs754501468 Chr.15: 34233977-34233977 on Build GRCh38
rs760605080 Chr.15: 34233857-34233857 on Build GRCh38
rs954279560 Chr.15: 34233900-34233900 on Build GRCh38
rs375876646 Chr.15: 34233994-34233994 on Build GRCh38
rs375277662 Chr.15: 34233992-34233992 on Build GRCh38
rs761919498 Chr.15: 34234011-34234011 on Build GRCh38
rs1007139874 Chr.15: 34233928-34233928 on Build GRCh38
rs201915253 Chr.15: 34233835-34233835 on Build GRCh38
rs1043804124 Chr.15: 34233996-34233996 on Build GRCh38
rs747524478 Chr.15: 34233935-34233935 on Build GRCh38
rs115102582 Chr.15: 34234069-34234069 on Build GRCh38
rs556517856 Chr.15: 34233959-34233959 on Build GRCh38
rs759376614 Chr.15: 34233851-34233851 on Build GRCh38
rs191955067 Chr.15: 34233850-34233850 on Build GRCh38
rs879011763 Chr.15: 34234040-34234040 on Build GRCh38
rs998302071 Chr.15: 34234204-34234204 on Build GRCh38
rs759464363 Chr.15: 34233993-34233993 on Build GRCh38
rs777174639 Chr.15: 34233947-34233947 on Build GRCh38
rs780397219 Chr.15: 34234024-34234024 on Build GRCh38
rs1047332054 Chr.15: 34234223-34234223 on Build GRCh38
rs764086996 Chr.15: 34233864-34233864 on Build GRCh38
rs373027493 Chr.15: 34233998-34233998 on Build GRCh38

View Full Product Details