Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 37936724-37937258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMCO5A
Entrez Gene ID
145942
Gene Name
transmembrane and coiled-coil domains 5A
Gene Aliases
TMCO5
UniGene
Hs.179646
Gene Chromosome Location
Chr.15: 37934607-37997541 on Build GRCh38
Transcripts
NM_152453.3 XM_005254168.3 XM_011521262.2 XM_011521263.1 XM_011521264.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4535846 c.223G>A p.E75K Chr.15: 37936929-37936929 on Build GRCh38
268470 c.166C>T p.R56W Chr.15: 37936872-37936872 on Build GRCh38
285770 c.164C>T p.T55M Chr.15: 37936870-37936870 on Build GRCh38
470543 c.213G>A p.T71T Chr.15: 37936919-37936919 on Build GRCh38
238313 c.201G>A p.E67E Chr.15: 37936907-37936907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753267606 Chr.15: 37936952-37936952 on Build GRCh38
rs766554532 Chr.15: 37936940-37936940 on Build GRCh38
rs562538894 Chr.15: 37936983-37936983 on Build GRCh38
rs528790162 Chr.15: 37937138-37937138 on Build GRCh38
rs746483929 Chr.15: 37936990-37936990 on Build GRCh38
rs765710934 Chr.15: 37937204-37937204 on Build GRCh38
rs1050581627 Chr.15: 37936894-37936894 on Build GRCh38
rs957144699 Chr.15: 37937136-37937136 on Build GRCh38
rs776389062 Chr.15: 37936857-37936857 on Build GRCh38
rs753069248 Chr.15: 37936883-37936883 on Build GRCh38
rs998095237 Chr.15: 37936886-37936886 on Build GRCh38
rs948973091 Chr.15: 37937226-37937226 on Build GRCh38
rs889002578 Chr.15: 37937089-37937089 on Build GRCh38
rs1040673455 Chr.15: 37937187-37937187 on Build GRCh38
rs962696201 Chr.15: 37936784-37936784 on Build GRCh38
rs959946590 Chr.15: 37937048-37937048 on Build GRCh38
rs768065830 Chr.15: 37936947-37936947 on Build GRCh38
rs953458389 Chr.15: 37936794-37936794 on Build GRCh38
rs10520067 Chr.15: 37936772-37936772 on Build GRCh38
rs558575773 Chr.15: 37936819-37936819 on Build GRCh38
rs17557582 Chr.15: 37936791-37936791 on Build GRCh38
rs1031425498 Chr.15: 37937114-37937114 on Build GRCh38
rs768157722 Chr.15: 37936881-37936881 on Build GRCh38
rs745519068 Chr.15: 37936905-37936905 on Build GRCh38
rs977183613 Chr.15: 37936898-37936898 on Build GRCh38
rs774554755 Chr.15: 37936941-37936941 on Build GRCh38
rs201144596 Chr.15: 37936873-37936873 on Build GRCh38
rs758051282 Chr.15: 37936906-37936906 on Build GRCh38
rs948996340 Chr.15: 37936950-37936950 on Build GRCh38
rs781051370 Chr.15: 37937001-37937001 on Build GRCh38
rs1021697843 Chr.15: 37936920-37936920 on Build GRCh38
rs773488924 Chr.15: 37936870-37936870 on Build GRCh38
rs747873511 Chr.15: 37936923-37936923 on Build GRCh38
rs939684286 Chr.15: 37936813-37936813 on Build GRCh38
rs928559240 Chr.15: 37936753-37936753 on Build GRCh38
rs140859377 Chr.15: 37936912-37936912 on Build GRCh38
rs766078599 Chr.15: 37937229-37937229 on Build GRCh38
rs1004223730 Chr.15: 37936987-37936987 on Build GRCh38
rs1028776014 Chr.15: 37937028-37937028 on Build GRCh38
rs989799755 Chr.15: 37937174-37937174 on Build GRCh38
rs772941990 Chr.15: 37936937-37936937 on Build GRCh38
rs773758510 Chr.15: 37936786-37936786 on Build GRCh38
rs749089853 Chr.15: 37937019-37937019 on Build GRCh38
rs148231491 Chr.15: 37937108-37937108 on Build GRCh38
rs144021993 Chr.15: 37936871-37936871 on Build GRCh38
rs1015523027 Chr.15: 37936782-37936782 on Build GRCh38
rs756502146 Chr.15: 37936890-37936890 on Build GRCh38
rs776581165 Chr.15: 37936919-37936919 on Build GRCh38
rs907397677 Chr.15: 37936965-37936965 on Build GRCh38
rs768619479 Chr.15: 37936841-37936841 on Build GRCh38
rs867154521 Chr.15: 37936928-37936928 on Build GRCh38
rs574611932 Chr.15: 37937004-37937004 on Build GRCh38
rs763233987 Chr.15: 37936939-37936939 on Build GRCh38
rs774920399 Chr.15: 37936827-37936827 on Build GRCh38
rs760598743 Chr.15: 37936840-37936840 on Build GRCh38
rs761620512 Chr.15: 37936859-37936859 on Build GRCh38
rs543876878 Chr.15: 37936907-37936907 on Build GRCh38
rs768532037 Chr.15: 37936918-37936918 on Build GRCh38
rs376434269 Chr.15: 37936946-37936946 on Build GRCh38
rs779704010 Chr.15: 37936989-37936989 on Build GRCh38
rs751757881 Chr.15: 37936872-37936872 on Build GRCh38
rs769646187 Chr.15: 37936927-37936927 on Build GRCh38
rs924385960 Chr.15: 37936904-37936904 on Build GRCh38
rs745305683 Chr.15: 37936809-37936809 on Build GRCh38
rs907264917 Chr.15: 37936768-37936768 on Build GRCh38
rs754195014 Chr.15: 37936979-37936979 on Build GRCh38
rs374833527 Chr.15: 37936795-37936795 on Build GRCh38
rs373271370 Chr.15: 37936911-37936911 on Build GRCh38
rs778368891 Chr.15: 37936900-37936900 on Build GRCh38
rs1007908909 Chr.15: 37937094-37937094 on Build GRCh38
rs769690098 Chr.15: 37937010-37937010 on Build GRCh38
rs374551216 Chr.15: 37937015-37937015 on Build GRCh38
rs764514134 Chr.15: 37936968-37936968 on Build GRCh38
rs991741838 Chr.15: 37937065-37937065 on Build GRCh38
rs756633826 Chr.15: 37936953-37936953 on Build GRCh38
rs370901356 Chr.15: 37936992-37936992 on Build GRCh38
rs141746906 Chr.15: 37937110-37937110 on Build GRCh38
rs778655945 Chr.15: 37936804-37936804 on Build GRCh38
rs965855325 Chr.15: 37937109-37937109 on Build GRCh38

View Full Product Details