Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 37947299-37947804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMCO5A
Entrez Gene ID
145942
Gene Name
transmembrane and coiled-coil domains 5A
Gene Aliases
TMCO5
UniGene
Hs.179646
Gene Chromosome Location
Chr.15: 37934607-37997541 on Build GRCh38
Transcripts
NM_152453.3 XM_005254168.3 XM_011521262.2 XM_011521263.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1180146 c.645G>C p.K215N Chr.15: 37947673-37947673 on Build GRCh38
140054 c.629G>A p.G210D Chr.15: 37947657-37947657 on Build GRCh38
7082481 c.663_664insA p.I224fs*58 Chr.15: 37947691-37947692 on Build GRCh38
6988294 c.668+1G>T p.? Chr.15: 37947697-37947697 on Build GRCh38
2268606 c.665A>C p.K222T Chr.15: 37947693-37947693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756001140 Chr.15: 37947729-37947729 on Build GRCh38
rs763074812 Chr.15: 37947639-37947639 on Build GRCh38
rs983271217 Chr.15: 37947371-37947371 on Build GRCh38
rs111501347 Chr.15: 37947571-37947571 on Build GRCh38
rs990958782 Chr.15: 37947483-37947483 on Build GRCh38
rs902653365 Chr.15: 37947551-37947552 on Build GRCh38
rs763126372 Chr.15: 37947627-37947630 on Build GRCh38
rs1035042917 Chr.15: 37947356-37947356 on Build GRCh38
rs113747446 Chr.15: 37947525-37947525 on Build GRCh38
rs566306732 Chr.15: 37947725-37947725 on Build GRCh38
rs548472960 Chr.15: 37947724-37947724 on Build GRCh38
rs193920912 Chr.15: 37947673-37947673 on Build GRCh38
rs558666929 Chr.15: 37947778-37947778 on Build GRCh38
rs1032245852 Chr.15: 37947715-37947715 on Build GRCh38
rs757204108 Chr.15: 37947564-37947564 on Build GRCh38
rs921620763 Chr.15: 37947466-37947466 on Build GRCh38
rs780995013 Chr.15: 37947697-37947697 on Build GRCh38
rs545648365 Chr.15: 37947482-37947482 on Build GRCh38
rs377568962 Chr.15: 37947591-37947591 on Build GRCh38
rs1041720060 Chr.15: 37947662-37947662 on Build GRCh38
rs114078251 Chr.15: 37947578-37947578 on Build GRCh38
rs772594194 Chr.15: 37947739-37947739 on Build GRCh38
rs372649009 Chr.15: 37947716-37947716 on Build GRCh38
rs761294223 Chr.15: 37947744-37947744 on Build GRCh38
rs190352080 Chr.15: 37947379-37947379 on Build GRCh38
rs940935315 Chr.15: 37947543-37947543 on Build GRCh38
rs958109527 Chr.15: 37947754-37947754 on Build GRCh38
rs773335150 Chr.15: 37947634-37947634 on Build GRCh38
rs753041897 Chr.15: 37947666-37947666 on Build GRCh38
rs959602976 Chr.15: 37947360-37947360 on Build GRCh38
rs916372862 Chr.15: 37947500-37947500 on Build GRCh38
rs957926681 Chr.15: 37947455-37947455 on Build GRCh38
rs747932849 Chr.15: 37947702-37947702 on Build GRCh38
rs569129709 Chr.15: 37947678-37947678 on Build GRCh38
rs563958175 Chr.15: 37947511-37947511 on Build GRCh38
rs769990514 Chr.15: 37947627-37947627 on Build GRCh38
rs764896913 Chr.15: 37947676-37947676 on Build GRCh38
rs543006160 Chr.15: 37947552-37947552 on Build GRCh38
rs532160329 Chr.15: 37947665-37947665 on Build GRCh38
rs533790350 Chr.15: 37947751-37947751 on Build GRCh38
rs561081959 Chr.15: 37947569-37947569 on Build GRCh38
rs966603622 Chr.15: 37947404-37947404 on Build GRCh38
rs921673423 Chr.15: 37947763-37947763 on Build GRCh38
rs1012117193 Chr.15: 37947755-37947755 on Build GRCh38
rs1009274967 Chr.15: 37947661-37947661 on Build GRCh38
rs747908601 Chr.15: 37947612-37947612 on Build GRCh38
rs777756060 Chr.15: 37947719-37947719 on Build GRCh38
rs200595347 Chr.15: 37947709-37947709 on Build GRCh38
rs775885679 Chr.15: 37947741-37947741 on Build GRCh38
rs115632768 Chr.15: 37947333-37947333 on Build GRCh38
rs894051904 Chr.15: 37947520-37947520 on Build GRCh38
rs931600489 Chr.15: 37947506-37947506 on Build GRCh38
rs78542351 Chr.15: 37947657-37947657 on Build GRCh38
rs575891716 Chr.15: 37947517-37947517 on Build GRCh38
rs536234536 Chr.15: 37947369-37947369 on Build GRCh38
rs12438391 Chr.15: 37947380-37947380 on Build GRCh38
rs751128025 Chr.15: 37947683-37947683 on Build GRCh38
rs12442232 Chr.15: 37947362-37947362 on Build GRCh38
rs578194652 Chr.15: 37947432-37947432 on Build GRCh38
rs778580136 Chr.15: 37947437-37947441 on Build GRCh38
rs1004015958 Chr.15: 37947339-37947339 on Build GRCh38
rs374513949 Chr.15: 37947353-37947353 on Build GRCh38
rs774518269 Chr.15: 37947735-37947735 on Build GRCh38
rs768542961 Chr.15: 37947605-37947605 on Build GRCh38
rs556490322 Chr.15: 37947425-37947425 on Build GRCh38
rs990739273 Chr.15: 37947361-37947361 on Build GRCh38
rs779535585 Chr.15: 37947680-37947680 on Build GRCh38
rs754885248 Chr.15: 37947696-37947696 on Build GRCh38
rs1042554229 Chr.15: 37947550-37947550 on Build GRCh38
rs747643615 Chr.15: 37947770-37947770 on Build GRCh38
rs767488329 Chr.15: 37947656-37947656 on Build GRCh38
rs751459081 Chr.15: 37947509-37947509 on Build GRCh38
rs76650148 Chr.15: 37947418-37947418 on Build GRCh38
rs546655893 Chr.15: 37947610-37947610 on Build GRCh38
rs759695817 Chr.15: 37947737-37947737 on Build GRCh38

View Full Product Details