Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45198999-45199496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHF
Entrez Gene ID
90525
Gene Name
Src homology 2 domain containing F
Gene Aliases
-
UniGene
Hs.310399
Gene Chromosome Location
Chr.15: 45167214-45201175 on Build GRCh38
Transcripts
NM_138356.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6327517 c.150G>A p.A50A Chr.15: 45199096-45199096 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs937622017 Chr.15: 45199435-45199435 on Build GRCh38
rs569877109 Chr.15: 45199292-45199292 on Build GRCh38
rs749797742 Chr.15: 45199159-45199159 on Build GRCh38
rs904836754 Chr.15: 45199183-45199183 on Build GRCh38
rs762404401 Chr.15: 45199104-45199104 on Build GRCh38
rs1043381038 Chr.15: 45199169-45199169 on Build GRCh38
rs985421981 Chr.15: 45199219-45199219 on Build GRCh38
rs905119975 Chr.15: 45199405-45199405 on Build GRCh38
rs752024439 Chr.15: 45199020-45199020 on Build GRCh38
rs370501642 Chr.15: 45199072-45199072 on Build GRCh38
rs138444775 Chr.15: 45199379-45199379 on Build GRCh38
rs758588697 Chr.15: 45199142-45199142 on Build GRCh38
rs573264428 Chr.15: 45199339-45199339 on Build GRCh38
rs528505493 Chr.15: 45199350-45199350 on Build GRCh38
rs752974867 Chr.15: 45199029-45199029 on Build GRCh38
rs915907271 Chr.15: 45199369-45199369 on Build GRCh38
rs904621263 Chr.15: 45199408-45199408 on Build GRCh38
rs950522989 Chr.15: 45199253-45199253 on Build GRCh38
rs765885395 Chr.15: 45199108-45199108 on Build GRCh38
rs973126070 Chr.15: 45199210-45199210 on Build GRCh38
rs1017673843 Chr.15: 45199204-45199204 on Build GRCh38
rs531017740 Chr.15: 45199416-45199416 on Build GRCh38
rs1053676734 Chr.15: 45199439-45199439 on Build GRCh38
rs757317522 Chr.15: 45199046-45199046 on Build GRCh38
rs746026274 Chr.15: 45199184-45199184 on Build GRCh38
rs769342136 Chr.15: 45199088-45199088 on Build GRCh38
rs999009152 Chr.15: 45199413-45199413 on Build GRCh38
rs767936647 Chr.15: 45199139-45199139 on Build GRCh38
rs112077676 Chr.15: 45199185-45199185 on Build GRCh38
rs1021753515 Chr.15: 45199157-45199157 on Build GRCh38
rs867942607 Chr.15: 45199095-45199095 on Build GRCh38
rs923363788 Chr.15: 45199079-45199079 on Build GRCh38
rs760023988 Chr.15: 45199120-45199120 on Build GRCh38
rs1011268875 Chr.15: 45199141-45199142 on Build GRCh38
rs768629536 Chr.15: 45199058-45199058 on Build GRCh38
rs749342999 Chr.15: 45199044-45199044 on Build GRCh38
rs190196718 Chr.15: 45199199-45199199 on Build GRCh38
rs542919799 Chr.15: 45199209-45199209 on Build GRCh38
rs910815170 Chr.15: 45199227-45199227 on Build GRCh38
rs755666588 Chr.15: 45199061-45199061 on Build GRCh38
rs376077702 Chr.15: 45199022-45199022 on Build GRCh38
rs1055079934 Chr.15: 45199355-45199355 on Build GRCh38
rs913748127 Chr.15: 45199295-45199295 on Build GRCh38
rs564851816 Chr.15: 45199208-45199209 on Build GRCh38
rs920349737 Chr.15: 45199217-45199217 on Build GRCh38
rs976055726 Chr.15: 45199245-45199245 on Build GRCh38
rs559759710 Chr.15: 45199322-45199322 on Build GRCh38
rs766819582 Chr.15: 45199019-45199019 on Build GRCh38
rs147509646 Chr.15: 45199193-45199193 on Build GRCh38
rs1648299 Chr.15: 45199196-45199196 on Build GRCh38
rs906452622 Chr.15: 45199057-45199057 on Build GRCh38
rs544905178 Chr.15: 45199054-45199054 on Build GRCh38
rs373900693 Chr.15: 45199119-45199119 on Build GRCh38
rs777985516 Chr.15: 45199033-45199033 on Build GRCh38
rs772870057 Chr.15: 45199092-45199092 on Build GRCh38
rs767850521 Chr.15: 45199025-45199025 on Build GRCh38
rs749647935 Chr.15: 45199257-45199257 on Build GRCh38
rs566479424 Chr.15: 45199056-45199056 on Build GRCh38
rs964646358 Chr.15: 45199232-45199232 on Build GRCh38
rs565411236 Chr.15: 45199406-45199406 on Build GRCh38

View Full Product Details