Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45661877-45662385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SQRDL
Entrez Gene ID
58472
Gene Name
sulfide quinone reductase-like (yeast)
Gene Aliases
CGI-44, PRO1975, SQOR
UniGene
Hs.511251
Gene Chromosome Location
Chr.15: 45631148-45691294 on Build GRCh38
Transcripts
NM_001271213.1 NM_021199.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1478162 c.245A>C p.Y82S Chr.15: 45661965-45661965 on Build GRCh38
6325483 c.301C>T p.R101C Chr.15: 45662021-45662021 on Build GRCh38
5796785 c.308C>A p.T103K Chr.15: 45662028-45662028 on Build GRCh38
962267 c.401A>G p.E134G Chr.15: 45662121-45662121 on Build GRCh38
5024212 c.235-5C>A p.? Chr.15: 45661950-45661950 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372185368 Chr.15: 45662023-45662023 on Build GRCh38
rs758610812 Chr.15: 45662087-45662087 on Build GRCh38
rs11553490 Chr.15: 45661977-45661977 on Build GRCh38
rs765383285 Chr.15: 45661925-45661926 on Build GRCh38
rs767503060 Chr.15: 45662051-45662051 on Build GRCh38
rs750729101 Chr.15: 45661986-45662001 on Build GRCh38
rs775758681 Chr.15: 45661917-45661917 on Build GRCh38
rs148163396 Chr.15: 45661984-45661984 on Build GRCh38
rs750495836 Chr.15: 45662086-45662086 on Build GRCh38
rs887810797 Chr.15: 45662343-45662343 on Build GRCh38
rs114648923 Chr.15: 45662280-45662280 on Build GRCh38
rs751360058 Chr.15: 45661995-45661995 on Build GRCh38
rs143647722 Chr.15: 45662279-45662279 on Build GRCh38
rs917981263 Chr.15: 45662285-45662286 on Build GRCh38
rs899632723 Chr.15: 45662108-45662108 on Build GRCh38
rs759409196 Chr.15: 45662044-45662044 on Build GRCh38
rs978893690 Chr.15: 45662264-45662264 on Build GRCh38
rs141060633 Chr.15: 45662007-45662007 on Build GRCh38
rs778928942 Chr.15: 45662010-45662010 on Build GRCh38
rs944842485 Chr.15: 45662066-45662066 on Build GRCh38
rs770249680 Chr.15: 45662119-45662119 on Build GRCh38
rs988202049 Chr.15: 45662137-45662137 on Build GRCh38
rs1029801093 Chr.15: 45662126-45662126 on Build GRCh38
rs182420793 Chr.15: 45662289-45662289 on Build GRCh38
rs774789741 Chr.15: 45662297-45662297 on Build GRCh38
rs781600806 Chr.15: 45662116-45662116 on Build GRCh38
rs113007362 Chr.15: 45662144-45662144 on Build GRCh38
rs748618342 Chr.15: 45662117-45662117 on Build GRCh38
rs759497232 Chr.15: 45662003-45662003 on Build GRCh38
rs190637341 Chr.15: 45662021-45662021 on Build GRCh38
rs758785676 Chr.15: 45662006-45662007 on Build GRCh38
rs780501293 Chr.15: 45662010-45662010 on Build GRCh38
rs368726687 Chr.15: 45662091-45662091 on Build GRCh38
rs761319949 Chr.15: 45661959-45661959 on Build GRCh38
rs199996931 Chr.15: 45662016-45662016 on Build GRCh38
rs746947739 Chr.15: 45662038-45662038 on Build GRCh38
rs774156131 Chr.15: 45662043-45662043 on Build GRCh38
rs934890624 Chr.15: 45662027-45662027 on Build GRCh38
rs748710009 Chr.15: 45662025-45662025 on Build GRCh38
rs770511605 Chr.15: 45662034-45662034 on Build GRCh38
rs926050018 Chr.15: 45662265-45662265 on Build GRCh38
rs112287930 Chr.15: 45662136-45662136 on Build GRCh38
rs990649175 Chr.15: 45662251-45662251 on Build GRCh38
rs1027721935 Chr.15: 45662120-45662120 on Build GRCh38
rs983409211 Chr.15: 45661902-45661902 on Build GRCh38
rs752679183 Chr.15: 45662052-45662052 on Build GRCh38
rs766162333 Chr.15: 45661991-45661991 on Build GRCh38
rs1004967393 Chr.15: 45662140-45662140 on Build GRCh38
rs115029765 Chr.15: 45662288-45662288 on Build GRCh38
rs752032718 Chr.15: 45662147-45662148 on Build GRCh38
rs762640484 Chr.15: 45662029-45662029 on Build GRCh38
rs372368130 Chr.15: 45662080-45662080 on Build GRCh38
rs769286196 Chr.15: 45662026-45662026 on Build GRCh38
rs140029707 Chr.15: 45662238-45662238 on Build GRCh38
rs755319782 Chr.15: 45662112-45662112 on Build GRCh38
rs759329918 Chr.15: 45662159-45662159 on Build GRCh38
rs769177573 Chr.15: 45661924-45661924 on Build GRCh38
rs780147586 Chr.15: 45662088-45662088 on Build GRCh38
rs200193729 Chr.15: 45661973-45661973 on Build GRCh38
rs17628584 Chr.15: 45662359-45662359 on Build GRCh38
rs772481694 Chr.15: 45661913-45661913 on Build GRCh38
rs145161599 Chr.15: 45662157-45662157 on Build GRCh38
rs747532344 Chr.15: 45661920-45661920 on Build GRCh38
rs781675712 Chr.15: 45662024-45662024 on Build GRCh38
rs945001219 Chr.15: 45662299-45662299 on Build GRCh38
rs922134390 Chr.15: 45662345-45662345 on Build GRCh38
rs764809712 Chr.15: 45661965-45661965 on Build GRCh38
rs150887067 Chr.15: 45662022-45662022 on Build GRCh38
rs114112036 Chr.15: 45662278-45662278 on Build GRCh38
rs770409881 Chr.15: 45662189-45662189 on Build GRCh38
rs926833757 Chr.15: 45661956-45661956 on Build GRCh38
rs746135742 Chr.15: 45661911-45661911 on Build GRCh38
rs748780003 Chr.15: 45662138-45662138 on Build GRCh38
rs970533642 Chr.15: 45662263-45662263 on Build GRCh38
rs772634155 Chr.15: 45662028-45662028 on Build GRCh38
rs767592938 Chr.15: 45662005-45662005 on Build GRCh38
rs760734672 Chr.15: 45662057-45662057 on Build GRCh38
rs143200108 Chr.15: 45661942-45661942 on Build GRCh38
rs1023507652 Chr.15: 45662257-45662257 on Build GRCh38
rs1039171070 Chr.15: 45662301-45662301 on Build GRCh38

View Full Product Details