Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49897927-49898428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8B4
Entrez Gene ID
79895
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Gene Aliases
ATPIM
UniGene
Hs.511311
Gene Chromosome Location
Chr.15: 49858238-50182096 on Build GRCh38
Transcripts
NM_024837.3 NR_073596.1 NR_073597.1 NR_073598.1 XM_011522046.2 XM_011522047.2 XM_011522048.1 XM_011522049.2 XM_011522051.2 XM_011522052.2 XM_011522053.1 XM_011522056.2 XM_011522058.2 XM_011522059.1 XM_011522060.1 XM_011522061.1 XM_011522062.1 XM_011522063.1 XM_011522064.1 XM_011522069.2 XM_011522070.1 XM_017022587.1 XM_017022588.1 XM_017022589.1 XM_017022590.1 XM_017022591.1 XM_017022592.1 XM_017022593.1 XM_017022594.1 XM_017022595.1 XM_017022596.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
248719 c.2312T>A p.V771D Chr.15: 49898229-49898229 on Build GRCh38
4793422 c.2407A>T p.K803* Chr.15: 49898134-49898134 on Build GRCh38
1129185 c.2354T>C p.V785A Chr.15: 49898187-49898187 on Build GRCh38
2156967 c.2319T>A p.N773K Chr.15: 49898222-49898222 on Build GRCh38
309380 c.2386G>T p.A796S Chr.15: 49898155-49898155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576713410 Chr.15: 49898200-49898200 on Build GRCh38
rs762633108 Chr.15: 49898274-49898274 on Build GRCh38
rs745467301 Chr.15: 49898262-49898262 on Build GRCh38
rs757054235 Chr.15: 49898244-49898244 on Build GRCh38
rs777802572 Chr.15: 49898043-49898043 on Build GRCh38
rs756007693 Chr.15: 49898059-49898059 on Build GRCh38
rs774895282 Chr.15: 49898196-49898196 on Build GRCh38
rs576780031 Chr.15: 49898393-49898393 on Build GRCh38
rs373435903 Chr.15: 49898221-49898221 on Build GRCh38
rs766849989 Chr.15: 49898195-49898195 on Build GRCh38
rs749660068 Chr.15: 49898295-49898295 on Build GRCh38
rs982561196 Chr.15: 49897982-49897982 on Build GRCh38
rs75508967 Chr.15: 49898170-49898170 on Build GRCh38
rs747135303 Chr.15: 49898160-49898160 on Build GRCh38
rs764797698 Chr.15: 49897997-49897997 on Build GRCh38
rs766188881 Chr.15: 49898327-49898327 on Build GRCh38
rs377320089 Chr.15: 49898074-49898074 on Build GRCh38
rs776971080 Chr.15: 49898063-49898063 on Build GRCh38
rs949711391 Chr.15: 49898093-49898093 on Build GRCh38
rs1040453933 Chr.15: 49898384-49898384 on Build GRCh38
rs377154477 Chr.15: 49898299-49898299 on Build GRCh38
rs766971164 Chr.15: 49898206-49898206 on Build GRCh38
rs769977264 Chr.15: 49898263-49898263 on Build GRCh38
rs765659718 Chr.15: 49898077-49898077 on Build GRCh38
rs781362293 Chr.15: 49898258-49898258 on Build GRCh38
rs572659126 Chr.15: 49898000-49898000 on Build GRCh38
rs531826217 Chr.15: 49898141-49898141 on Build GRCh38
rs899793941 Chr.15: 49897953-49897953 on Build GRCh38
rs777377084 Chr.15: 49898131-49898131 on Build GRCh38
rs139212703 Chr.15: 49898339-49898339 on Build GRCh38
rs760836659 Chr.15: 49898048-49898048 on Build GRCh38
rs1044624139 Chr.15: 49898223-49898223 on Build GRCh38
rs775370328 Chr.15: 49898046-49898046 on Build GRCh38
rs754468021 Chr.15: 49898102-49898102 on Build GRCh38
rs149008340 Chr.15: 49898100-49898100 on Build GRCh38
rs747990696 Chr.15: 49898026-49898026 on Build GRCh38
rs564870756 Chr.15: 49898383-49898383 on Build GRCh38
rs749255848 Chr.15: 49898147-49898147 on Build GRCh38
rs770207242 Chr.15: 49898173-49898173 on Build GRCh38
rs1029552220 Chr.15: 49897967-49897967 on Build GRCh38
rs753636885 Chr.15: 49898237-49898237 on Build GRCh38
rs752312796 Chr.15: 49898115-49898115 on Build GRCh38
rs768825651 Chr.15: 49898165-49898165 on Build GRCh38
rs971570805 Chr.15: 49898377-49898377 on Build GRCh38
rs951709707 Chr.15: 49898144-49898144 on Build GRCh38
rs151094786 Chr.15: 49898135-49898135 on Build GRCh38
rs761367462 Chr.15: 49897992-49897992 on Build GRCh38
rs370283733 Chr.15: 49898099-49898099 on Build GRCh38
rs951394469 Chr.15: 49897971-49897971 on Build GRCh38
rs1016720234 Chr.15: 49898317-49898317 on Build GRCh38
rs745762613 Chr.15: 49898248-49898248 on Build GRCh38
rs773468761 Chr.15: 49898272-49898272 on Build GRCh38
rs769552729 Chr.15: 49898038-49898038 on Build GRCh38
rs376111714 Chr.15: 49897966-49897966 on Build GRCh38
rs759985070 Chr.15: 49898303-49898303 on Build GRCh38
rs771292535 Chr.15: 49898298-49898298 on Build GRCh38
rs116715364 Chr.15: 49898191-49898191 on Build GRCh38
rs201681268 Chr.15: 49898171-49898171 on Build GRCh38
rs962428123 Chr.15: 49898012-49898012 on Build GRCh38
rs763802802 Chr.15: 49898235-49898235 on Build GRCh38
rs749134718 Chr.15: 49898044-49898044 on Build GRCh38
rs1022682692 Chr.15: 49898373-49898373 on Build GRCh38
rs778900850 Chr.15: 49898156-49898156 on Build GRCh38
rs755931160 Chr.15: 49898125-49898125 on Build GRCh38
rs867404498 Chr.15: 49898086-49898086 on Build GRCh38
rs935603125 Chr.15: 49898148-49898148 on Build GRCh38
rs780814298 Chr.15: 49898017-49898017 on Build GRCh38
rs143804562 Chr.15: 49898103-49898103 on Build GRCh38
rs891868538 Chr.15: 49898347-49898347 on Build GRCh38
rs1027996986 Chr.15: 49898117-49898117 on Build GRCh38
rs778755605 Chr.15: 49898247-49898247 on Build GRCh38
rs1008960413 Chr.15: 49898353-49898353 on Build GRCh38
rs755699504 Chr.15: 49898233-49898233 on Build GRCh38
rs772208640 Chr.15: 49898045-49898045 on Build GRCh38
rs773789627 Chr.15: 49898088-49898088 on Build GRCh38
rs769058642 Chr.15: 49898057-49898057 on Build GRCh38
rs779763250 Chr.15: 49898076-49898076 on Build GRCh38
rs755132808 Chr.15: 49898251-49898251 on Build GRCh38
rs763308952 Chr.15: 49898092-49898092 on Build GRCh38
rs141037209 Chr.15: 49898155-49898155 on Build GRCh38
rs928212432 Chr.15: 49898142-49898142 on Build GRCh38
rs998602654 Chr.15: 49897962-49897962 on Build GRCh38
rs1004120253 Chr.15: 49898305-49898305 on Build GRCh38
rs145377032 Chr.15: 49898178-49898178 on Build GRCh38

View Full Product Details