Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 55181751-55182293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RSL24D1
Entrez Gene ID
51187
Gene Name
ribosomal L24 domain containing 1
Gene Aliases
C15orf15, HRP-L30-iso, L30, RLP24, RPL24, RPL24L, TVAS3
UniGene
Hs.274772
Gene Chromosome Location
Chr.15: 55181314-55197033 on Build GRCh38
Transcripts
NM_016304.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6273052 c.460C>A p.Q154K Chr.15: 55182184-55182184 on Build GRCh38
6273053 c.451C>A p.Q151K Chr.15: 55182193-55182193 on Build GRCh38
1373604 c.441G>T p.E147D Chr.15: 55182203-55182203 on Build GRCh38
138965 c.419G>A p.G140D Chr.15: 55182225-55182225 on Build GRCh38
6263610 c.429A>G p.K143K Chr.15: 55182215-55182215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143479241 Chr.15: 55182072-55182072 on Build GRCh38
rs1053991080 Chr.15: 55181969-55181969 on Build GRCh38
rs747337216 Chr.15: 55182238-55182238 on Build GRCh38
rs920086404 Chr.15: 55181828-55181828 on Build GRCh38
rs1016757425 Chr.15: 55181777-55181777 on Build GRCh38
rs750572411 Chr.15: 55182179-55182179 on Build GRCh38
rs751183994 Chr.15: 55182202-55182202 on Build GRCh38
rs755601961 Chr.15: 55181867-55181867 on Build GRCh38
rs551951434 Chr.15: 55182268-55182268 on Build GRCh38
rs936834385 Chr.15: 55181926-55181926 on Build GRCh38
rs766430929 Chr.15: 55182192-55182192 on Build GRCh38
rs755494238 Chr.15: 55182122-55182122 on Build GRCh38
rs781511293 Chr.15: 55182203-55182203 on Build GRCh38
rs557656206 Chr.15: 55181791-55181791 on Build GRCh38
rs11540399 Chr.15: 55181995-55181995 on Build GRCh38
rs188072191 Chr.15: 55181801-55181801 on Build GRCh38
rs986335100 Chr.15: 55181897-55181897 on Build GRCh38
rs770225609 Chr.15: 55182133-55182133 on Build GRCh38
rs751686612 Chr.15: 55182194-55182194 on Build GRCh38
rs771954608 Chr.15: 55182165-55182165 on Build GRCh38
rs879237383 Chr.15: 55182016-55182016 on Build GRCh38
rs200847511 Chr.15: 55182206-55182206 on Build GRCh38
rs763887148 Chr.15: 55182175-55182175 on Build GRCh38
rs906169033 Chr.15: 55182069-55182069 on Build GRCh38
rs891509592 Chr.15: 55181965-55181965 on Build GRCh38
rs962184498 Chr.15: 55181822-55181822 on Build GRCh38
rs1053282233 Chr.15: 55181946-55181946 on Build GRCh38
rs750630362 Chr.15: 55182201-55182209 on Build GRCh38
rs914065861 Chr.15: 55181974-55181974 on Build GRCh38
rs11540369 Chr.15: 55182078-55182078 on Build GRCh38
rs560014253 Chr.15: 55181884-55181884 on Build GRCh38
rs780702890 Chr.15: 55182050-55182054 on Build GRCh38
rs11540373 Chr.15: 55182108-55182108 on Build GRCh38
rs573369853 Chr.15: 55181898-55181898 on Build GRCh38
rs369049485 Chr.15: 55182162-55182162 on Build GRCh38
rs770766047 Chr.15: 55182239-55182239 on Build GRCh38
rs774073894 Chr.15: 55182160-55182160 on Build GRCh38
rs780406690 Chr.15: 55182232-55182233 on Build GRCh38
rs778288140 Chr.15: 55182224-55182224 on Build GRCh38
rs563585818 Chr.15: 55182147-55182147 on Build GRCh38
rs753779637 Chr.15: 55182012-55182012 on Build GRCh38
rs781596166 Chr.15: 55182123-55182123 on Build GRCh38
rs775329023 Chr.15: 55182168-55182168 on Build GRCh38
rs758714284 Chr.15: 55182230-55182235 on Build GRCh38
rs760350791 Chr.15: 55182170-55182170 on Build GRCh38
rs993571049 Chr.15: 55182245-55182245 on Build GRCh38
rs564276797 Chr.15: 55182022-55182022 on Build GRCh38
rs765521773 Chr.15: 55182128-55182129 on Build GRCh38
rs923232927 Chr.15: 55181868-55181868 on Build GRCh38
rs780107982 Chr.15: 55182110-55182110 on Build GRCh38
rs748501964 Chr.15: 55182124-55182124 on Build GRCh38
rs949012320 Chr.15: 55181843-55181843 on Build GRCh38
rs11540387 Chr.15: 55182176-55182176 on Build GRCh38
rs562019218 Chr.15: 55182047-55182047 on Build GRCh38
rs1046198430 Chr.15: 55182046-55182046 on Build GRCh38
rs539153883 Chr.15: 55182230-55182230 on Build GRCh38
rs906908306 Chr.15: 55181906-55181906 on Build GRCh38
rs748962587 Chr.15: 55182148-55182148 on Build GRCh38
rs778547752 Chr.15: 55182248-55182248 on Build GRCh38
rs373778784 Chr.15: 55182269-55182269 on Build GRCh38
rs770500399 Chr.15: 55182154-55182154 on Build GRCh38
rs954694007 Chr.15: 55182253-55182253 on Build GRCh38
rs755504242 Chr.15: 55182251-55182253 on Build GRCh38
rs546361213 Chr.15: 55181829-55181829 on Build GRCh38
rs17852589 Chr.15: 55182207-55182207 on Build GRCh38
rs763224716 Chr.15: 55182189-55182189 on Build GRCh38
rs940188680 Chr.15: 55181795-55181795 on Build GRCh38
rs920117719 Chr.15: 55182226-55182226 on Build GRCh38
rs900316356 Chr.15: 55181991-55181991 on Build GRCh38
rs938336121 Chr.15: 55181938-55181938 on Build GRCh38
rs972564753 Chr.15: 55181849-55181849 on Build GRCh38
rs372593488 Chr.15: 55181821-55181821 on Build GRCh38
rs987305150 Chr.15: 55181785-55181785 on Build GRCh38
rs756376298 Chr.15: 55182213-55182213 on Build GRCh38
rs369626781 Chr.15: 55182111-55182111 on Build GRCh38

View Full Product Details