Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 55827819-55828364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEDD4
Entrez Gene ID
4734
Gene Name
neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase
Gene Aliases
NEDD4-1, RPF1
UniGene
Hs.1565
Gene Chromosome Location
Chr.15: 55826917-55993746 on Build GRCh38
Transcripts
NM_001284338.1 NM_001284339.1 NM_001284340.1 NM_006154.3 NM_198400.3 NR_104302.1 XM_011521624.2 XM_011521625.2 XM_011521626.1 XM_011521627.1

SNP Detail

SNP ID Chromosome Location
rs370465939 Chr.15: 55828011-55828011 on Build GRCh38
rs1041790319 Chr.15: 55828077-55828077 on Build GRCh38
rs964261660 Chr.15: 55828326-55828326 on Build GRCh38
rs539800643 Chr.15: 55828247-55828247 on Build GRCh38
rs879129690 Chr.15: 55827908-55827908 on Build GRCh38
rs954499312 Chr.15: 55828266-55828266 on Build GRCh38
rs545273218 Chr.15: 55827916-55827916 on Build GRCh38
rs993208410 Chr.15: 55828183-55828183 on Build GRCh38
rs946128346 Chr.15: 55828075-55828075 on Build GRCh38
rs556997746 Chr.15: 55827868-55827868 on Build GRCh38
rs971844711 Chr.15: 55827918-55827918 on Build GRCh38
rs1017711649 Chr.15: 55828317-55828317 on Build GRCh38
rs1054534457 Chr.15: 55828037-55828037 on Build GRCh38
rs577555443 Chr.15: 55828239-55828239 on Build GRCh38
rs985803119 Chr.15: 55828305-55828305 on Build GRCh38
rs1030102795 Chr.15: 55828238-55828238 on Build GRCh38
rs573392163 Chr.15: 55828333-55828333 on Build GRCh38
rs914782188 Chr.15: 55828052-55828052 on Build GRCh38
rs954406966 Chr.15: 55827956-55827956 on Build GRCh38
rs369773552 Chr.15: 55828264-55828264 on Build GRCh38
rs1016100113 Chr.15: 55828171-55828171 on Build GRCh38
rs375967325 Chr.15: 55827848-55827848 on Build GRCh38
rs961770546 Chr.15: 55827881-55827881 on Build GRCh38
rs372414054 Chr.15: 55828161-55828161 on Build GRCh38
rs1006068977 Chr.15: 55827872-55827872 on Build GRCh38
rs911493110 Chr.15: 55827985-55827985 on Build GRCh38
rs942974148 Chr.15: 55827998-55827998 on Build GRCh38
rs370936247 Chr.15: 55828316-55828316 on Build GRCh38
rs546535741 Chr.15: 55827984-55827984 on Build GRCh38
rs907157692 Chr.15: 55828095-55828095 on Build GRCh38
rs894377817 Chr.15: 55828142-55828142 on Build GRCh38
rs938714264 Chr.15: 55828100-55828100 on Build GRCh38
rs1016131270 Chr.15: 55827876-55827876 on Build GRCh38
rs560099999 Chr.15: 55828284-55828284 on Build GRCh38
rs773964525 Chr.15: 55828173-55828173 on Build GRCh38
rs958759954 Chr.15: 55827849-55827849 on Build GRCh38
rs559975541 Chr.15: 55828160-55828160 on Build GRCh38
rs1056088307 Chr.15: 55828101-55828101 on Build GRCh38
rs759874076 Chr.15: 55827979-55827979 on Build GRCh38
rs11550869 Chr.15: 55827934-55827934 on Build GRCh38

View Full Product Details