Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64823692-64824192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIF1
Entrez Gene ID
80119
Gene Name
PIF1 5'-to-3' DNA helicase
Gene Aliases
C15orf20, PIF
UniGene
Hs.112160
Gene Chromosome Location
Chr.15: 64815630-64827173 on Build GRCh38
Transcripts
NM_001286496.1 NM_001286497.1 NM_001286499.1 NM_025049.3 XM_011522083.2 XM_011522084.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3386981 c.402C>T p.S134S Chr.15: 64823934-64823934 on Build GRCh38
3386979 c.402C>T p.S134S Chr.15: 64823934-64823934 on Build GRCh38
6746017 c.500C>T p.P167L Chr.15: 64823836-64823836 on Build GRCh38
6745996 c.441C>T p.F147F Chr.15: 64823895-64823895 on Build GRCh38
1735090 c.450C>G p.I150M Chr.15: 64823886-64823886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191345285 Chr.15: 64824011-64824011 on Build GRCh38
rs897160606 Chr.15: 64824079-64824079 on Build GRCh38
rs770450355 Chr.15: 64823789-64823789 on Build GRCh38
rs140413159 Chr.15: 64823817-64823817 on Build GRCh38
rs780967662 Chr.15: 64823767-64823767 on Build GRCh38
rs767492357 Chr.15: 64823778-64823779 on Build GRCh38
rs986463453 Chr.15: 64824087-64824087 on Build GRCh38
rs1043812042 Chr.15: 64824146-64824146 on Build GRCh38
rs192801793 Chr.15: 64823888-64823888 on Build GRCh38
rs940592477 Chr.15: 64824056-64824056 on Build GRCh38
rs953717130 Chr.15: 64824084-64824084 on Build GRCh38
rs747234245 Chr.15: 64823756-64823756 on Build GRCh38
rs527276492 Chr.15: 64823794-64823794 on Build GRCh38
rs766736590 Chr.15: 64823903-64823903 on Build GRCh38
rs762406482 Chr.15: 64823779-64823779 on Build GRCh38
rs767214998 Chr.15: 64823795-64823795 on Build GRCh38
rs771677536 Chr.15: 64823984-64823984 on Build GRCh38
rs752890933 Chr.15: 64823915-64823915 on Build GRCh38
rs747879567 Chr.15: 64823768-64823768 on Build GRCh38
rs1056397253 Chr.15: 64824173-64824173 on Build GRCh38
rs749267955 Chr.15: 64823861-64823861 on Build GRCh38
rs777651338 Chr.15: 64823944-64823944 on Build GRCh38
rs1019885790 Chr.15: 64824169-64824169 on Build GRCh38
rs182752537 Chr.15: 64823957-64823957 on Build GRCh38
rs566896632 Chr.15: 64823963-64823963 on Build GRCh38
rs532573339 Chr.15: 64823873-64823873 on Build GRCh38
rs530238706 Chr.15: 64823955-64823955 on Build GRCh38
rs752731887 Chr.15: 64824138-64824138 on Build GRCh38
rs1037001132 Chr.15: 64824071-64824075 on Build GRCh38
rs889905916 Chr.15: 64824113-64824113 on Build GRCh38
rs755045906 Chr.15: 64823907-64823907 on Build GRCh38
rs200547278 Chr.15: 64823881-64823881 on Build GRCh38
rs892550022 Chr.15: 64823760-64823760 on Build GRCh38
rs773402462 Chr.15: 64823900-64823900 on Build GRCh38
rs12899195 Chr.15: 64824129-64824129 on Build GRCh38
rs750911515 Chr.15: 64823816-64823816 on Build GRCh38
rs938045409 Chr.15: 64824165-64824165 on Build GRCh38
rs905281567 Chr.15: 64824151-64824151 on Build GRCh38
rs534178442 Chr.15: 64823983-64823983 on Build GRCh38
rs756239008 Chr.15: 64823927-64823927 on Build GRCh38
rs763212025 Chr.15: 64824050-64824050 on Build GRCh38
rs755867842 Chr.15: 64823846-64823846 on Build GRCh38
rs758500085 Chr.15: 64823715-64823715 on Build GRCh38
rs766972415 Chr.15: 64824062-64824062 on Build GRCh38
rs962837745 Chr.15: 64824031-64824031 on Build GRCh38
rs544530355 Chr.15: 64823894-64823894 on Build GRCh38
rs946847737 Chr.15: 64824142-64824142 on Build GRCh38
rs565237510 Chr.15: 64823848-64823848 on Build GRCh38
rs560122536 Chr.15: 64823759-64823759 on Build GRCh38
rs754271431 Chr.15: 64823799-64823799 on Build GRCh38
rs540565125 Chr.15: 64823827-64823827 on Build GRCh38
rs998256497 Chr.15: 64824093-64824093 on Build GRCh38
rs1053078798 Chr.15: 64824126-64824126 on Build GRCh38
rs184599780 Chr.15: 64824105-64824105 on Build GRCh38
rs556294430 Chr.15: 64824063-64824063 on Build GRCh38
rs773636413 Chr.15: 64824039-64824039 on Build GRCh38
rs538053269 Chr.15: 64824018-64824018 on Build GRCh38
rs765730639 Chr.15: 64823806-64823806 on Build GRCh38
rs769780987 Chr.15: 64823769-64823769 on Build GRCh38
rs775332201 Chr.15: 64823989-64823989 on Build GRCh38
rs1015428547 Chr.15: 64824030-64824030 on Build GRCh38
rs908320996 Chr.15: 64823836-64823836 on Build GRCh38
rs1004501304 Chr.15: 64824029-64824029 on Build GRCh38
rs751570097 Chr.15: 64823818-64823818 on Build GRCh38
rs762259681 Chr.15: 64823802-64823802 on Build GRCh38
rs912130791 Chr.15: 64823945-64823945 on Build GRCh38
rs201870033 Chr.15: 64823835-64823835 on Build GRCh38
rs770731971 Chr.15: 64823872-64823872 on Build GRCh38
rs553577928 Chr.15: 64824160-64824160 on Build GRCh38
rs569269488 Chr.15: 64823910-64823910 on Build GRCh38
rs751694903 Chr.15: 64823905-64823905 on Build GRCh38
rs187718134 Chr.15: 64824002-64824002 on Build GRCh38
rs1034885240 Chr.15: 64823810-64823810 on Build GRCh38
rs755205278 Chr.15: 64823823-64823823 on Build GRCh38
rs957577508 Chr.15: 64824148-64824148 on Build GRCh38
rs767605057 Chr.15: 64824136-64824136 on Build GRCh38
rs960648006 Chr.15: 64824022-64824022 on Build GRCh38
rs541483300 Chr.15: 64823737-64823737 on Build GRCh38
rs1002140057 Chr.15: 64824007-64824007 on Build GRCh38
rs1033176780 Chr.15: 64824141-64824141 on Build GRCh38
rs763549463 Chr.15: 64823902-64823902 on Build GRCh38
rs745318914 Chr.15: 64823880-64823880 on Build GRCh38
rs748972052 Chr.15: 64823947-64823947 on Build GRCh38
rs574438134 Chr.15: 64823732-64823732 on Build GRCh38
rs1052972784 Chr.15: 64823748-64823748 on Build GRCh38
rs912183260 Chr.15: 64824103-64824103 on Build GRCh38
rs773192225 Chr.15: 64823775-64823775 on Build GRCh38

View Full Product Details