Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90230831-90231333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIB1
Entrez Gene ID
10519
Gene Name
calcium and integrin binding 1
Gene Aliases
CIB, CIBP, KIP1, PRKDCIP, SIP2-28
UniGene
Hs.715556
Gene Chromosome Location
Chr.15: 90230245-90265759 on Build GRCh38
Transcripts
NM_001277764.1 NM_006384.3 NR_102427.1 NR_102428.1 XM_006720375.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6863170 c.428G>A p.R143Q Chr.15: 90231132-90231132 on Build GRCh38
5384108 c.459C>T p.I153I Chr.15: 90231101-90231101 on Build GRCh38
3505374 c.500C>T p.T167I Chr.15: 90230988-90230988 on Build GRCh38
702141 c.418G>A p.E140K Chr.15: 90231142-90231142 on Build GRCh38
356028 c.431T>C p.L144P Chr.15: 90231129-90231129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751508143 Chr.15: 90231216-90231216 on Build GRCh38
rs779519559 Chr.15: 90231095-90231097 on Build GRCh38
rs868546161 Chr.15: 90231109-90231109 on Build GRCh38
rs748901358 Chr.15: 90231243-90231243 on Build GRCh38
rs141456665 Chr.15: 90231143-90231143 on Build GRCh38
rs888421088 Chr.15: 90230985-90230985 on Build GRCh38
rs778037667 Chr.15: 90230974-90230975 on Build GRCh38
rs771868093 Chr.15: 90230989-90230989 on Build GRCh38
rs377673570 Chr.15: 90231063-90231063 on Build GRCh38
rs868624291 Chr.15: 90230937-90230937 on Build GRCh38
rs758821375 Chr.15: 90230949-90230949 on Build GRCh38
rs753268972 Chr.15: 90231032-90231032 on Build GRCh38
rs1018739778 Chr.15: 90231018-90231018 on Build GRCh38
rs745539161 Chr.15: 90231152-90231152 on Build GRCh38
rs746905362 Chr.15: 90231137-90231137 on Build GRCh38
rs757484105 Chr.15: 90230953-90230953 on Build GRCh38
rs771632804 Chr.15: 90231065-90231065 on Build GRCh38
rs914933274 Chr.15: 90231094-90231094 on Build GRCh38
rs757076488 Chr.15: 90231217-90231217 on Build GRCh38
rs144975236 Chr.15: 90231122-90231122 on Build GRCh38
rs150421465 Chr.15: 90231179-90231179 on Build GRCh38
rs769355989 Chr.15: 90231093-90231093 on Build GRCh38
rs781487047 Chr.15: 90230955-90230955 on Build GRCh38
rs767933367 Chr.15: 90231191-90231191 on Build GRCh38
rs139813837 Chr.15: 90230961-90230961 on Build GRCh38
rs747991133 Chr.15: 90230963-90230963 on Build GRCh38
rs560810851 Chr.15: 90231123-90231123 on Build GRCh38
rs367739456 Chr.15: 90230856-90230856 on Build GRCh38
rs552611897 Chr.15: 90231171-90231171 on Build GRCh38
rs753385402 Chr.15: 90230925-90230925 on Build GRCh38
rs765750540 Chr.15: 90230939-90230939 on Build GRCh38
rs947854661 Chr.15: 90230858-90230858 on Build GRCh38
rs199557034 Chr.15: 90231219-90231219 on Build GRCh38
rs200906231 Chr.15: 90231222-90231222 on Build GRCh38
rs761859158 Chr.15: 90231132-90231132 on Build GRCh38
rs754257962 Chr.15: 90230975-90230978 on Build GRCh38
rs71715019 Chr.15: 90231218-90231221 on Build GRCh38
rs548055723 Chr.15: 90231056-90231056 on Build GRCh38
rs761018255 Chr.15: 90231197-90231197 on Build GRCh38
rs893423761 Chr.15: 90231209-90231210 on Build GRCh38
rs781536323 Chr.15: 90231057-90231057 on Build GRCh38
rs772925077 Chr.15: 90231012-90231012 on Build GRCh38
rs34604050 Chr.15: 90231216-90231219 on Build GRCh38
rs761682424 Chr.15: 90231245-90231245 on Build GRCh38
rs368617389 Chr.15: 90231153-90231153 on Build GRCh38
rs751965559 Chr.15: 90231050-90231050 on Build GRCh38
rs760388156 Chr.15: 90231013-90231013 on Build GRCh38
rs755078702 Chr.15: 90231203-90231203 on Build GRCh38
rs778944136 Chr.15: 90230962-90230962 on Build GRCh38
rs779940261 Chr.15: 90231068-90231068 on Build GRCh38
rs771818894 Chr.15: 90230900-90230900 on Build GRCh38
rs879032034 Chr.15: 90231188-90231188 on Build GRCh38
rs779886155 Chr.15: 90231230-90231230 on Build GRCh38
rs370102744 Chr.15: 90230967-90230967 on Build GRCh38
rs757062860 Chr.15: 90231139-90231139 on Build GRCh38
rs1048420091 Chr.15: 90230966-90230966 on Build GRCh38
rs765061684 Chr.15: 90231142-90231142 on Build GRCh38
rs201909717 Chr.15: 90231218-90231218 on Build GRCh38
rs772448500 Chr.15: 90231248-90231248 on Build GRCh38
rs775259021 Chr.15: 90231046-90231046 on Build GRCh38
rs753043127 Chr.15: 90230942-90230942 on Build GRCh38
rs564545691 Chr.15: 90231275-90231275 on Build GRCh38
rs957577705 Chr.15: 90230938-90230938 on Build GRCh38
rs1044813 Chr.15: 90231027-90231027 on Build GRCh38
rs778123299 Chr.15: 90230874-90230874 on Build GRCh38
rs753956561 Chr.15: 90231200-90231200 on Build GRCh38
rs760263375 Chr.15: 90230918-90230918 on Build GRCh38
rs146621891 Chr.15: 90231097-90231097 on Build GRCh38
rs775122177 Chr.15: 90231170-90231170 on Build GRCh38
rs1011177752 Chr.15: 90231276-90231276 on Build GRCh38
rs760919745 Chr.15: 90231264-90231264 on Build GRCh38
rs757511234 Chr.15: 90230981-90230983 on Build GRCh38
rs745941847 Chr.15: 90230959-90230959 on Build GRCh38
rs34958365 Chr.15: 90231125-90231125 on Build GRCh38
rs1022937677 Chr.15: 90231278-90231278 on Build GRCh38
rs761641487 Chr.15: 90230895-90230895 on Build GRCh38
rs768294875 Chr.15: 90231245-90231245 on Build GRCh38
rs34645714 Chr.15: 90231116-90231116 on Build GRCh38
rs771284505 Chr.15: 90231313-90231313 on Build GRCh38
rs774791760 Chr.15: 90231095-90231095 on Build GRCh38
rs750575510 Chr.15: 90231059-90231059 on Build GRCh38
rs772874521 Chr.15: 90230914-90230914 on Build GRCh38
rs777139775 Chr.15: 90231138-90231138 on Build GRCh38
rs762300810 Chr.15: 90231104-90231104 on Build GRCh38
rs745455439 Chr.15: 90231025-90231025 on Build GRCh38
rs370566116 Chr.15: 90231077-90231077 on Build GRCh38
rs775021192 Chr.15: 90231172-90231172 on Build GRCh38
rs763491927 Chr.15: 90231035-90231035 on Build GRCh38
rs777654090 Chr.15: 90231133-90231133 on Build GRCh38
rs773449398 Chr.15: 90231196-90231196 on Build GRCh38
rs939156685 Chr.15: 90230891-90230891 on Build GRCh38
rs531811652 Chr.15: 90231283-90231283 on Build GRCh38
rs376858222 Chr.15: 90230920-90230920 on Build GRCh38
rs145803459 Chr.15: 90230952-90230952 on Build GRCh38
rs766491256 Chr.15: 90231199-90231199 on Build GRCh38
rs746373849 Chr.15: 90231033-90231036 on Build GRCh38
rs563075894 Chr.15: 90231101-90231101 on Build GRCh38
rs969086687 Chr.15: 90231279-90231279 on Build GRCh38
rs374226647 Chr.15: 90231262-90231262 on Build GRCh38
rs750241867 Chr.15: 90231314-90231314 on Build GRCh38
rs776376956 Chr.15: 90231087-90231087 on Build GRCh38
rs11551251 Chr.15: 90231144-90231144 on Build GRCh38
rs1010477466 Chr.15: 90231215-90231218 on Build GRCh38
rs746706925 Chr.15: 90231144-90231144 on Build GRCh38
rs767726731 Chr.15: 90231113-90231113 on Build GRCh38
rs138227306 Chr.15: 90231206-90231206 on Build GRCh38
rs768388996 Chr.15: 90231150-90231151 on Build GRCh38
rs774066774 Chr.15: 90230890-90230890 on Build GRCh38
rs372376585 Chr.15: 90231031-90231031 on Build GRCh38
rs377200278 Chr.15: 90231081-90231081 on Build GRCh38
rs11551248 Chr.15: 90231005-90231005 on Build GRCh38
rs779403095 Chr.15: 90231033-90231034 on Build GRCh38
rs758328291 Chr.15: 90231076-90231076 on Build GRCh38
rs201298867 Chr.15: 90231100-90231100 on Build GRCh38
rs370911524 Chr.15: 90230902-90230902 on Build GRCh38
rs901289325 Chr.15: 90231225-90231225 on Build GRCh38
rs748735632 Chr.15: 90231246-90231246 on Build GRCh38
rs760017428 Chr.15: 90230926-90230926 on Build GRCh38
rs748121144 Chr.15: 90231069-90231069 on Build GRCh38
rs755837250 Chr.15: 90231226-90231226 on Build GRCh38
rs768628183 Chr.15: 90230887-90230887 on Build GRCh38
rs746782583 Chr.15: 90231079-90231079 on Build GRCh38
rs745462077 Chr.15: 90231089-90231089 on Build GRCh38
rs557391723 Chr.15: 90230872-90230872 on Build GRCh38

View Full Product Details