Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 724242-724744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC78
Entrez Gene ID
124093
Gene Name
coiled-coil domain containing 78
Gene Aliases
C16orf25, CNM4, JFP10, hsCCDC78
UniGene
Hs.381943
Gene Chromosome Location
Chr.16: 722578-726880 on Build GRCh38
Transcripts
NM_001031737.2 XM_006720838.1 XM_006720843.3 XM_011522356.1 XM_011522357.1 XM_011522358.2 XM_011522359.1 XM_011522360.1 XM_011522361.1 XM_011522362.1 XM_011522363.1 XM_011522364.1 XM_011522365.1 XM_011522366.1 XM_011522367.1 XM_011522368.1 XM_011522369.1 XM_011522370.1 XM_011522371.2 XM_017022929.1 XM_017022930.1 XM_017022932.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
435807 c.878C>G p.A293G Chr.16: 724397-724397 on Build GRCh38
6998590 c.811C>T p.R271W Chr.16: 724464-724464 on Build GRCh38
1195849 c.857G>A p.R286H Chr.16: 724418-724418 on Build GRCh38
4062995 c.772G>A p.A258T Chr.16: 724503-724503 on Build GRCh38
5722366 c.812G>A p.R271Q Chr.16: 724463-724463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767497873 Chr.16: 724471-724471 on Build GRCh38
rs777073735 Chr.16: 724316-724316 on Build GRCh38
rs758020711 Chr.16: 724589-724589 on Build GRCh38
rs375855087 Chr.16: 724359-724359 on Build GRCh38
rs1037631909 Chr.16: 724433-724433 on Build GRCh38
rs761833676 Chr.16: 724339-724339 on Build GRCh38
rs764422260 Chr.16: 724463-724463 on Build GRCh38
rs540376434 Chr.16: 724668-724668 on Build GRCh38
rs760618049 Chr.16: 724402-724402 on Build GRCh38
rs767728545 Chr.16: 724557-724557 on Build GRCh38
rs781751298 Chr.16: 724417-724417 on Build GRCh38
rs755323760 Chr.16: 724497-724497 on Build GRCh38
rs147606575 Chr.16: 724709-724709 on Build GRCh38
rs1050848910 Chr.16: 724510-724510 on Build GRCh38
rs958395578 Chr.16: 724393-724393 on Build GRCh38
rs753039480 Chr.16: 724673-724673 on Build GRCh38
rs2071950 Chr.16: 724692-724692 on Build GRCh38
rs151049173 Chr.16: 724352-724352 on Build GRCh38
rs773013782 Chr.16: 724398-724398 on Build GRCh38
rs374100885 Chr.16: 724291-724291 on Build GRCh38
rs557894563 Chr.16: 724593-724593 on Build GRCh38
rs756036249 Chr.16: 724696-724696 on Build GRCh38
rs749080322 Chr.16: 724652-724652 on Build GRCh38
rs61998209 Chr.16: 724338-724338 on Build GRCh38
rs766284775 Chr.16: 724405-724405 on Build GRCh38
rs763349441 Chr.16: 724683-724683 on Build GRCh38
rs775288285 Chr.16: 724543-724543 on Build GRCh38
rs1010425376 Chr.16: 724293-724293 on Build GRCh38
rs775101094 Chr.16: 724327-724327 on Build GRCh38
rs765738306 Chr.16: 724672-724672 on Build GRCh38
rs756646427 Chr.16: 724480-724480 on Build GRCh38
rs1014754418 Chr.16: 724365-724365 on Build GRCh38
rs750138760 Chr.16: 724272-724272 on Build GRCh38
rs567316689 Chr.16: 724568-724568 on Build GRCh38
rs746756519 Chr.16: 724401-724401 on Build GRCh38
rs369207689 Chr.16: 724532-724532 on Build GRCh38
rs201634385 Chr.16: 724430-724430 on Build GRCh38
rs769471057 Chr.16: 724541-724541 on Build GRCh38
rs79863898 Chr.16: 724312-724312 on Build GRCh38
rs760813011 Chr.16: 724662-724662 on Build GRCh38
rs771582682 Chr.16: 724375-724377 on Build GRCh38
rs746174565 Chr.16: 724318-724318 on Build GRCh38
rs376047604 Chr.16: 724348-724348 on Build GRCh38
rs778492814 Chr.16: 724305-724305 on Build GRCh38
rs201136331 Chr.16: 724266-724266 on Build GRCh38
rs748943027 Chr.16: 724384-724384 on Build GRCh38
rs1012692311 Chr.16: 724720-724720 on Build GRCh38
rs371760552 Chr.16: 724409-724409 on Build GRCh38
rs746959349 Chr.16: 724447-724447 on Build GRCh38
rs778504860 Chr.16: 724302-724302 on Build GRCh38
rs376412009 Chr.16: 724717-724717 on Build GRCh38
rs760723287 Chr.16: 724565-724565 on Build GRCh38
rs943428119 Chr.16: 724622-724622 on Build GRCh38
rs758630518 Chr.16: 724353-724353 on Build GRCh38
rs750889592 Chr.16: 724418-724418 on Build GRCh38
rs77707419 Chr.16: 724472-724472 on Build GRCh38
rs769711936 Chr.16: 724645-724645 on Build GRCh38
rs557666577 Chr.16: 724583-724583 on Build GRCh38
rs755260099 Chr.16: 724277-724277 on Build GRCh38
rs372703744 Chr.16: 724465-724465 on Build GRCh38
rs771029694 Chr.16: 724531-724531 on Build GRCh38
rs561702421 Chr.16: 724423-724423 on Build GRCh38
rs772096201 Chr.16: 724395-724395 on Build GRCh38
rs769382946 Chr.16: 724650-724650 on Build GRCh38
rs776636446 Chr.16: 724670-724670 on Build GRCh38
rs764179609 Chr.16: 724413-724413 on Build GRCh38
rs369120396 Chr.16: 724394-724394 on Build GRCh38
rs573005461 Chr.16: 724603-724603 on Build GRCh38
rs751160160 Chr.16: 724594-724594 on Build GRCh38
rs773321960 Chr.16: 724560-724560 on Build GRCh38
rs200865845 Chr.16: 724464-724464 on Build GRCh38
rs761683637 Chr.16: 724638-724638 on Build GRCh38
rs61998220 Chr.16: 724386-724386 on Build GRCh38
rs756810544 Chr.16: 724597-724597 on Build GRCh38
rs750273057 Chr.16: 724345-724345 on Build GRCh38
rs745805323 Chr.16: 724634-724634 on Build GRCh38
rs199893871 Chr.16: 724518-724518 on Build GRCh38
rs1016508703 Chr.16: 724320-724320 on Build GRCh38
rs371030168 Chr.16: 724695-724695 on Build GRCh38
rs747558848 Chr.16: 724309-724309 on Build GRCh38
rs765537870 Chr.16: 724570-724570 on Build GRCh38
rs770130604 Chr.16: 724382-724382 on Build GRCh38
rs775875068 Chr.16: 724660-724660 on Build GRCh38
rs201042133 Chr.16: 724443-724443 on Build GRCh38
rs562625664 Chr.16: 724448-724448 on Build GRCh38
rs747752625 Chr.16: 724431-724431 on Build GRCh38
rs749614530 Chr.16: 724424-724424 on Build GRCh38
rs537432674 Chr.16: 724574-724574 on Build GRCh38
rs751454157 Chr.16: 724307-724307 on Build GRCh38
rs201235859 Chr.16: 724329-724329 on Build GRCh38
rs747763912 Chr.16: 724504-724504 on Build GRCh38
rs375793651 Chr.16: 724665-724665 on Build GRCh38
rs375625161 Chr.16: 724414-724414 on Build GRCh38
rs781517196 Chr.16: 724376-724376 on Build GRCh38
rs377588917 Chr.16: 724300-724300 on Build GRCh38
rs4984921 Chr.16: 724520-724520 on Build GRCh38
rs371640942 Chr.16: 724517-724517 on Build GRCh38
rs776538123 Chr.16: 724584-724585 on Build GRCh38
rs779921377 Chr.16: 724391-724426 on Build GRCh38
rs768495466 Chr.16: 724714-724714 on Build GRCh38
rs1014786972 Chr.16: 724521-724521 on Build GRCh38
rs545339703 Chr.16: 724591-724591 on Build GRCh38
rs751250054 Chr.16: 724690-724690 on Build GRCh38
rs766842372 Chr.16: 724484-724484 on Build GRCh38
rs186738044 Chr.16: 724494-724494 on Build GRCh38
rs920615844 Chr.16: 724664-724664 on Build GRCh38
rs142133229 Chr.16: 724691-724691 on Build GRCh38
rs781344129 Chr.16: 724601-724601 on Build GRCh38
rs903278547 Chr.16: 724282-724282 on Build GRCh38
rs778696812 Chr.16: 724503-724503 on Build GRCh38
rs767319981 Chr.16: 724341-724341 on Build GRCh38
rs1037684307 Chr.16: 724631-724631 on Build GRCh38
rs200154277 Chr.16: 724412-724412 on Build GRCh38
rs757810344 Chr.16: 724364-724364 on Build GRCh38
rs779746748 Chr.16: 724383-724383 on Build GRCh38
rs540243094 Chr.16: 724419-724419 on Build GRCh38
rs761952739 Chr.16: 724544-724544 on Build GRCh38
rs374915151 Chr.16: 724321-724321 on Build GRCh38
rs763894588 Chr.16: 724689-724689 on Build GRCh38
rs755450229 Chr.16: 724569-724569 on Build GRCh38
rs745711662 Chr.16: 724534-724534 on Build GRCh38
rs1043057437 Chr.16: 724299-724299 on Build GRCh38
rs540043699 Chr.16: 724585-724585 on Build GRCh38
rs768619502 Chr.16: 724581-724582 on Build GRCh38
rs768148847 Chr.16: 724335-724335 on Build GRCh38
rs776596806 Chr.16: 724453-724453 on Build GRCh38
rs746291744 Chr.16: 724377-724377 on Build GRCh38
rs768251031 Chr.16: 724385-724385 on Build GRCh38
rs575824271 Chr.16: 724584-724584 on Build GRCh38
rs752077571 Chr.16: 724716-724716 on Build GRCh38
rs960505485 Chr.16: 724572-724572 on Build GRCh38
rs139766874 Chr.16: 724357-724357 on Build GRCh38
rs199752586 Chr.16: 724290-724290 on Build GRCh38
rs372689615 Chr.16: 724653-724653 on Build GRCh38
rs749095737 Chr.16: 724710-724710 on Build GRCh38

View Full Product Details