Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 729530-730032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAGHL
Entrez Gene ID
84264
Gene Name
hydroxyacylglutathione hydrolase-like
Gene Aliases
-
UniGene
Hs.124015
Gene Chromosome Location
Chr.16: 726936-729746 on Build GRCh38
Transcripts
NM_001290137.1 NM_001290139.1 NM_001323635.1 NM_001323636.1 NM_032304.3 XM_005255632.1 XM_011522712.1 XM_017023774.1 NM_001304799.1 NM_022493.2

Gene Symbol
NARFL
Entrez Gene ID
64428
Gene Name
nuclear prelamin A recognition factor like
Gene Aliases
HPRN, IOP1, LET1L, NAR1, PRN
UniGene
Hs.513247
Gene Chromosome Location
Chr.16: 729753-741038 on Build GRCh38
Transcripts
NM_001290137.1 NM_001290139.1 NM_001323635.1 NM_001323636.1 NM_032304.3 XM_005255632.1 XM_011522712.1 XM_017023774.1 NM_001304799.1 NM_022493.2

SNP Detail

SNP ID Chromosome Location
rs114625496 Chr.16: 729947-729947 on Build GRCh38
rs748635847 Chr.16: 729716-729716 on Build GRCh38
rs777966102 Chr.16: 729844-729844 on Build GRCh38
rs1049212745 Chr.16: 729688-729688 on Build GRCh38
rs1012249530 Chr.16: 729908-729908 on Build GRCh38
rs140410680 Chr.16: 729649-729649 on Build GRCh38
rs988422545 Chr.16: 729632-729632 on Build GRCh38
rs955769791 Chr.16: 729810-729810 on Build GRCh38
rs567723975 Chr.16: 729707-729707 on Build GRCh38
rs542759502 Chr.16: 729933-729933 on Build GRCh38
rs116485521 Chr.16: 729960-729960 on Build GRCh38
rs968227487 Chr.16: 729775-729775 on Build GRCh38
rs112746017 Chr.16: 730006-730006 on Build GRCh38
rs766024276 Chr.16: 729554-729554 on Build GRCh38
rs769274126 Chr.16: 729699-729699 on Build GRCh38
rs142241273 Chr.16: 729932-729933 on Build GRCh38
rs1045763169 Chr.16: 729919-729919 on Build GRCh38
rs1034763993 Chr.16: 729683-729683 on Build GRCh38
rs752612606 Chr.16: 729611-729611 on Build GRCh38
rs972141670 Chr.16: 729961-729961 on Build GRCh38
rs986175790 Chr.16: 729580-729580 on Build GRCh38
rs374782244 Chr.16: 729933-729933 on Build GRCh38
rs564585395 Chr.16: 729895-729895 on Build GRCh38
rs1033013740 Chr.16: 729749-729749 on Build GRCh38
rs953398944 Chr.16: 729750-729750 on Build GRCh38
rs372590979 Chr.16: 729679-729679 on Build GRCh38
rs903311376 Chr.16: 729927-729927 on Build GRCh38
rs897761603 Chr.16: 729635-729635 on Build GRCh38
rs765077156 Chr.16: 729610-729610 on Build GRCh38
rs538114997 Chr.16: 729888-729888 on Build GRCh38
rs756047076 Chr.16: 729727-729727 on Build GRCh38
rs896484107 Chr.16: 729903-729903 on Build GRCh38
rs1001010041 Chr.16: 729924-729925 on Build GRCh38
rs1001820836 Chr.16: 729673-729673 on Build GRCh38
rs905108079 Chr.16: 729861-729861 on Build GRCh38
rs556554873 Chr.16: 729741-729741 on Build GRCh38
rs777494488 Chr.16: 729646-729646 on Build GRCh38
rs911836348 Chr.16: 729645-729645 on Build GRCh38
rs944791446 Chr.16: 729813-729813 on Build GRCh38
rs753373624 Chr.16: 729585-729585 on Build GRCh38
rs137867689 Chr.16: 729846-729846 on Build GRCh38
rs1032750863 Chr.16: 729809-729809 on Build GRCh38
rs190993101 Chr.16: 729695-729695 on Build GRCh38
rs932061346 Chr.16: 729561-729561 on Build GRCh38
rs780159066 Chr.16: 729687-729687 on Build GRCh38
rs577999483 Chr.16: 729747-729747 on Build GRCh38
rs545329028 Chr.16: 729931-729931 on Build GRCh38
rs940498796 Chr.16: 729983-729983 on Build GRCh38
rs1000769425 Chr.16: 729928-729928 on Build GRCh38
rs1036187449 Chr.16: 729627-729627 on Build GRCh38
rs951961587 Chr.16: 729977-729977 on Build GRCh38
rs1020426796 Chr.16: 729738-729738 on Build GRCh38
rs75772387 Chr.16: 729780-729780 on Build GRCh38
rs369560316 Chr.16: 729882-729882 on Build GRCh38
rs183595960 Chr.16: 729606-729606 on Build GRCh38
rs74002869 Chr.16: 729930-729930 on Build GRCh38
rs117818762 Chr.16: 729889-729889 on Build GRCh38
rs778476900 Chr.16: 729579-729579 on Build GRCh38
rs1054788 Chr.16: 729689-729689 on Build GRCh38
rs907717839 Chr.16: 729982-729982 on Build GRCh38
rs747449102 Chr.16: 729877-729877 on Build GRCh38
rs1014859343 Chr.16: 729729-729729 on Build GRCh38
rs192160993 Chr.16: 729992-729992 on Build GRCh38
rs151215719 Chr.16: 729638-729638 on Build GRCh38
rs371560469 Chr.16: 729926-729926 on Build GRCh38
rs549721916 Chr.16: 729609-729609 on Build GRCh38
rs951943981 Chr.16: 729998-729998 on Build GRCh38
rs538980155 Chr.16: 729762-729762 on Build GRCh38
rs368368178 Chr.16: 729576-729576 on Build GRCh38
rs779295719 Chr.16: 729703-729703 on Build GRCh38
rs1000982054 Chr.16: 729804-729804 on Build GRCh38
rs537869602 Chr.16: 729740-729740 on Build GRCh38
rs940768444 Chr.16: 729890-729890 on Build GRCh38
rs1048934799 Chr.16: 729847-729847 on Build GRCh38
rs187310062 Chr.16: 729648-729648 on Build GRCh38
rs912009909 Chr.16: 729782-729782 on Build GRCh38
rs919089712 Chr.16: 729980-729980 on Build GRCh38
rs566766216 Chr.16: 729659-729659 on Build GRCh38
rs896459366 Chr.16: 729742-729742 on Build GRCh38
rs934360271 Chr.16: 729629-729629 on Build GRCh38
rs948079996 Chr.16: 730011-730011 on Build GRCh38
rs758141169 Chr.16: 729621-729621 on Build GRCh38
rs930495611 Chr.16: 729878-729878 on Build GRCh38
rs183135292 Chr.16: 729822-729822 on Build GRCh38
rs531236431 Chr.16: 729608-729608 on Build GRCh38
rs904099087 Chr.16: 729921-729921 on Build GRCh38
rs554097140 Chr.16: 729763-729763 on Build GRCh38
rs1024239480 Chr.16: 729574-729574 on Build GRCh38

View Full Product Details