Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3026599-3027091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THOC6
Entrez Gene ID
79228
Gene Name
THO complex 6
Gene Aliases
BBIS, WDR58, fSAP35
UniGene
Hs.412304
Gene Chromosome Location
Chr.16: 3024031-3027755 on Build GRCh38
Transcripts
NM_001142350.1 NM_024339.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5470503 c.562G>A p.E188K Chr.16: 3026757-3026757 on Build GRCh38
6994302 c.494G>A p.R165Q Chr.16: 3026689-3026689 on Build GRCh38
6503013 c.484-16_517del50 p.? Chr.16: 3026663-3026712 on Build GRCh38
969966 c.662G>T p.R221L Chr.16: 3027036-3027036 on Build GRCh38
969967 c.673T>C p.C225R Chr.16: 3027047-3027047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749122739 Chr.16: 3026882-3026882 on Build GRCh38
rs1044067591 Chr.16: 3026726-3026726 on Build GRCh38
rs116675970 Chr.16: 3027029-3027029 on Build GRCh38
rs973852407 Chr.16: 3026906-3026906 on Build GRCh38
rs965411685 Chr.16: 3026849-3026849 on Build GRCh38
rs772862889 Chr.16: 3026706-3026706 on Build GRCh38
rs747608649 Chr.16: 3026873-3026873 on Build GRCh38
rs371389733 Chr.16: 3026922-3026922 on Build GRCh38
rs778561717 Chr.16: 3026651-3026651 on Build GRCh38
rs781413903 Chr.16: 3026654-3026654 on Build GRCh38
rs199550964 Chr.16: 3026832-3026832 on Build GRCh38
rs775702913 Chr.16: 3026899-3026899 on Build GRCh38
rs780072471 Chr.16: 3027018-3027018 on Build GRCh38
rs779415557 Chr.16: 3027052-3027052 on Build GRCh38
rs373205446 Chr.16: 3026712-3026712 on Build GRCh38
rs757559613 Chr.16: 3026862-3026862 on Build GRCh38
rs757827268 Chr.16: 3026741-3026741 on Build GRCh38
rs750336605 Chr.16: 3026633-3026633 on Build GRCh38
rs764389709 Chr.16: 3026908-3026908 on Build GRCh38
rs777604472 Chr.16: 3026931-3026931 on Build GRCh38
rs768568569 Chr.16: 3027024-3027024 on Build GRCh38
rs368947202 Chr.16: 3026914-3026914 on Build GRCh38
rs372573615 Chr.16: 3026984-3026984 on Build GRCh38
rs754480335 Chr.16: 3026885-3026885 on Build GRCh38
rs759725023 Chr.16: 3026812-3026812 on Build GRCh38
rs746516859 Chr.16: 3026688-3026688 on Build GRCh38
rs776287633 Chr.16: 3026692-3026692 on Build GRCh38
rs779325586 Chr.16: 3026759-3026759 on Build GRCh38
rs758077838 Chr.16: 3027013-3027013 on Build GRCh38
rs774385085 Chr.16: 3026810-3026810 on Build GRCh38
rs137895830 Chr.16: 3027067-3027067 on Build GRCh38
rs746533344 Chr.16: 3026762-3026762 on Build GRCh38
rs199795381 Chr.16: 3026764-3026764 on Build GRCh38
rs367721672 Chr.16: 3026723-3026723 on Build GRCh38
rs748477565 Chr.16: 3026658-3026658 on Build GRCh38
rs745643131 Chr.16: 3026942-3026942 on Build GRCh38
rs147476413 Chr.16: 3026713-3026713 on Build GRCh38
rs746181278 Chr.16: 3026936-3026936 on Build GRCh38
rs200185006 Chr.16: 3026724-3026724 on Build GRCh38
rs552599710 Chr.16: 3026747-3026747 on Build GRCh38
rs768682789 Chr.16: 3026956-3026956 on Build GRCh38
rs1030625763 Chr.16: 3026885-3026885 on Build GRCh38
rs147432065 Chr.16: 3026889-3026889 on Build GRCh38
rs758380428 Chr.16: 3026634-3026634 on Build GRCh38
rs996170512 Chr.16: 3026855-3026856 on Build GRCh38
rs370769114 Chr.16: 3026822-3026822 on Build GRCh38
rs373259488 Chr.16: 3026971-3026971 on Build GRCh38
rs900506328 Chr.16: 3026845-3026845 on Build GRCh38
rs769317786 Chr.16: 3026792-3026792 on Build GRCh38
rs766697950 Chr.16: 3027035-3027035 on Build GRCh38
rs200979270 Chr.16: 3026898-3026898 on Build GRCh38
rs895532843 Chr.16: 3026705-3026705 on Build GRCh38
rs751677848 Chr.16: 3026643-3026643 on Build GRCh38
rs372903986 Chr.16: 3026945-3026945 on Build GRCh38
rs368334924 Chr.16: 3026825-3026825 on Build GRCh38
rs750888467 Chr.16: 3026864-3026864 on Build GRCh38
rs768211622 Chr.16: 3026859-3026859 on Build GRCh38
rs751287570 Chr.16: 3026993-3026993 on Build GRCh38
rs193187862 Chr.16: 3027030-3027030 on Build GRCh38
rs372197717 Chr.16: 3026666-3026666 on Build GRCh38
rs770681999 Chr.16: 3026937-3026937 on Build GRCh38
rs749636390 Chr.16: 3026727-3026727 on Build GRCh38
rs142684553 Chr.16: 3027064-3027064 on Build GRCh38
rs745688373 Chr.16: 3027009-3027009 on Build GRCh38
rs901292279 Chr.16: 3027038-3027038 on Build GRCh38
rs149384272 Chr.16: 3026689-3026689 on Build GRCh38
rs769492928 Chr.16: 3026875-3026875 on Build GRCh38
rs766615204 Chr.16: 3026639-3026639 on Build GRCh38
rs376441483 Chr.16: 3026769-3026769 on Build GRCh38
rs547634218 Chr.16: 3026863-3026863 on Build GRCh38
rs374622482 Chr.16: 3026904-3026904 on Build GRCh38
rs777514417 Chr.16: 3026880-3026880 on Build GRCh38
rs771401910 Chr.16: 3026669-3026669 on Build GRCh38
rs757035225 Chr.16: 3026995-3026995 on Build GRCh38
rs924818182 Chr.16: 3026902-3026902 on Build GRCh38
rs760829009 Chr.16: 3026954-3026954 on Build GRCh38
rs763749014 Chr.16: 3026990-3026990 on Build GRCh38
rs369105832 Chr.16: 3026841-3026841 on Build GRCh38
rs765440475 Chr.16: 3026970-3026970 on Build GRCh38
rs768129197 Chr.16: 3027055-3027055 on Build GRCh38
rs373780800 Chr.16: 3027028-3027028 on Build GRCh38
rs765122235 Chr.16: 3026697-3026697 on Build GRCh38
rs752267358 Chr.16: 3026924-3026924 on Build GRCh38
rs934443026 Chr.16: 3026691-3026691 on Build GRCh38
rs750775841 Chr.16: 3026915-3026915 on Build GRCh38
rs760174599 Chr.16: 3027050-3027050 on Build GRCh38
rs983646854 Chr.16: 3026895-3026895 on Build GRCh38
rs766436286 Chr.16: 3026803-3026803 on Build GRCh38
rs371666310 Chr.16: 3026950-3026950 on Build GRCh38
rs201879540 Chr.16: 3026709-3026709 on Build GRCh38
rs201552352 Chr.16: 3026930-3026930 on Build GRCh38
rs963856470 Chr.16: 3026901-3026901 on Build GRCh38
rs746451480 Chr.16: 3026842-3026843 on Build GRCh38
rs773209304 Chr.16: 3026798-3026798 on Build GRCh38
rs748992986 Chr.16: 3026935-3026935 on Build GRCh38
rs780738671 Chr.16: 3026977-3026985 on Build GRCh38
rs763171282 Chr.16: 3026974-3026974 on Build GRCh38
rs146263534 Chr.16: 3026959-3026959 on Build GRCh38
rs775231209 Chr.16: 3026952-3026952 on Build GRCh38
rs767673196 Chr.16: 3026820-3026820 on Build GRCh38
rs779695761 Chr.16: 3026626-3026627 on Build GRCh38
rs763408717 Chr.16: 3027034-3027034 on Build GRCh38
rs780815492 Chr.16: 3026872-3026872 on Build GRCh38
rs761533808 Chr.16: 3026693-3026693 on Build GRCh38
rs764962699 Chr.16: 3027071-3027071 on Build GRCh38
rs778244946 Chr.16: 3026725-3026725 on Build GRCh38
rs934853608 Chr.16: 3026907-3026907 on Build GRCh38
rs377615260 Chr.16: 3027036-3027036 on Build GRCh38
rs762774133 Chr.16: 3026801-3026801 on Build GRCh38
rs746958941 Chr.16: 3027019-3027019 on Build GRCh38
rs754008235 Chr.16: 3026910-3026910 on Build GRCh38
rs368529581 Chr.16: 3026672-3026672 on Build GRCh38
rs148494949 Chr.16: 3026894-3026894 on Build GRCh38
rs368011594 Chr.16: 3026663-3026663 on Build GRCh38
rs778639669 Chr.16: 3027001-3027001 on Build GRCh38
rs758811423 Chr.16: 3026869-3026869 on Build GRCh38
rs996876708 Chr.16: 3027065-3027065 on Build GRCh38
rs755483049 Chr.16: 3026989-3026989 on Build GRCh38
rs369176676 Chr.16: 3026943-3026943 on Build GRCh38
rs377022702 Chr.16: 3026992-3026992 on Build GRCh38
rs1012017750 Chr.16: 3026642-3026642 on Build GRCh38
rs747484760 Chr.16: 3026913-3026913 on Build GRCh38
rs759010888 Chr.16: 3026917-3026917 on Build GRCh38
rs960274 Chr.16: 3026662-3026662 on Build GRCh38
rs566107628 Chr.16: 3026826-3026826 on Build GRCh38
rs140523485 Chr.16: 3026773-3026773 on Build GRCh38

View Full Product Details