Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3495284-3495791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf90
Entrez Gene ID
646174
Gene Name
chromosome 16 open reading frame 90
Gene Aliases
-
UniGene
Hs.632224
Gene Chromosome Location
Chr.16: 3493484-3495521 on Build GRCh38
Transcripts
NM_001080524.1 XM_005255502.3 XM_017023068.1 XM_017023069.1

Gene Symbol
CLUAP1
Entrez Gene ID
23059
Gene Name
clusterin associated protein 1
Gene Aliases
CFAP22, FAP22, IFT38
UniGene
Hs.155995
Gene Chromosome Location
Chr.16: 3495562-3539048 on Build GRCh38
Transcripts
NM_001080524.1 XM_005255502.3 XM_017023068.1 XM_017023069.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3509471 c.3G>A p.M1I Chr.16: 3495419-3495419 on Build GRCh38
3690990 c.42A>G p.R14R Chr.16: 3495380-3495380 on Build GRCh38
5527570 c.26C>T p.S9F Chr.16: 3495396-3495396 on Build GRCh38
5778560 c.3G>T p.M1I Chr.16: 3495419-3495419 on Build GRCh38
5778586 c.46+1G>T p.? Chr.16: 3495375-3495375 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs557543431 Chr.16: 3495500-3495500 on Build GRCh38
rs958887951 Chr.16: 3495661-3495661 on Build GRCh38
rs759215628 Chr.16: 3495440-3495440 on Build GRCh38
rs747965891 Chr.16: 3495490-3495490 on Build GRCh38
rs552559156 Chr.16: 3495305-3495305 on Build GRCh38
rs753159478 Chr.16: 3495420-3495420 on Build GRCh38
rs181339878 Chr.16: 3495626-3495626 on Build GRCh38
rs1053688438 Chr.16: 3495426-3495426 on Build GRCh38
rs1006567171 Chr.16: 3495547-3495547 on Build GRCh38
rs927769254 Chr.16: 3495677-3495677 on Build GRCh38
rs573681049 Chr.16: 3495405-3495405 on Build GRCh38
rs945752695 Chr.16: 3495321-3495321 on Build GRCh38
rs371096440 Chr.16: 3495371-3495371 on Build GRCh38
rs1005260343 Chr.16: 3495751-3495751 on Build GRCh38
rs779793050 Chr.16: 3495478-3495478 on Build GRCh38
rs933366046 Chr.16: 3495381-3495381 on Build GRCh38
rs780946824 Chr.16: 3495488-3495488 on Build GRCh38
rs746948375 Chr.16: 3495479-3495479 on Build GRCh38
rs778545783 Chr.16: 3495358-3495358 on Build GRCh38
rs376213438 Chr.16: 3495391-3495391 on Build GRCh38
rs768180194 Chr.16: 3495521-3495521 on Build GRCh38
rs768641906 Chr.16: 3495484-3495484 on Build GRCh38
rs902325260 Chr.16: 3495316-3495316 on Build GRCh38
rs749852917 Chr.16: 3495449-3495449 on Build GRCh38
rs572494799 Chr.16: 3495617-3495617 on Build GRCh38
rs780064457 Chr.16: 3495367-3495367 on Build GRCh38
rs760186760 Chr.16: 3495409-3495409 on Build GRCh38
rs767978018 Chr.16: 3495410-3495410 on Build GRCh38
rs201278369 Chr.16: 3495385-3495385 on Build GRCh38
rs889636226 Chr.16: 3495540-3495540 on Build GRCh38
rs190153706 Chr.16: 3495576-3495576 on Build GRCh38
rs373672682 Chr.16: 3495544-3495544 on Build GRCh38
rs185168278 Chr.16: 3495658-3495658 on Build GRCh38
rs757939516 Chr.16: 3495477-3495477 on Build GRCh38
rs373677791 Chr.16: 3495375-3495375 on Build GRCh38
rs56205903 Chr.16: 3495364-3495364 on Build GRCh38
rs544288027 Chr.16: 3495498-3495498 on Build GRCh38
rs1033736695 Chr.16: 3495304-3495304 on Build GRCh38
rs1026341504 Chr.16: 3495379-3495379 on Build GRCh38
rs1005211099 Chr.16: 3495503-3495503 on Build GRCh38
rs1002340121 Chr.16: 3495451-3495451 on Build GRCh38
rs984990493 Chr.16: 3495322-3495322 on Build GRCh38
rs369354498 Chr.16: 3495308-3495308 on Build GRCh38
rs754599025 Chr.16: 3495442-3495443 on Build GRCh38
rs752109942 Chr.16: 3495408-3495408 on Build GRCh38
rs927446304 Chr.16: 3495412-3495412 on Build GRCh38
rs910517898 Chr.16: 3495404-3495404 on Build GRCh38
rs958015211 Chr.16: 3495620-3495620 on Build GRCh38
rs892419819 Chr.16: 3495430-3495430 on Build GRCh38
rs745425411 Chr.16: 3495360-3495360 on Build GRCh38
rs370639375 Chr.16: 3495413-3495413 on Build GRCh38
rs760096648 Chr.16: 3495511-3495511 on Build GRCh38
rs913914216 Chr.16: 3495637-3495637 on Build GRCh38
rs936609616 Chr.16: 3495529-3495529 on Build GRCh38
rs147446098 Chr.16: 3495511-3495511 on Build GRCh38
rs771039075 Chr.16: 3495501-3495501 on Build GRCh38
rs577437412 Chr.16: 3495539-3495539 on Build GRCh38
rs1046331474 Chr.16: 3495669-3495669 on Build GRCh38
rs937887538 Chr.16: 3495698-3495698 on Build GRCh38
rs900831040 Chr.16: 3495579-3495579 on Build GRCh38
rs762872866 Chr.16: 3495548-3495548 on Build GRCh38
rs946061681 Chr.16: 3495481-3495481 on Build GRCh38
rs71388519 Chr.16: 3495721-3495721 on Build GRCh38
rs774350966 Chr.16: 3495509-3495509 on Build GRCh38
rs528536488 Chr.16: 3495439-3495439 on Build GRCh38
rs962229007 Chr.16: 3495520-3495520 on Build GRCh38
rs917240078 Chr.16: 3495453-3495453 on Build GRCh38
rs552509617 Chr.16: 3495592-3495592 on Build GRCh38
rs986600273 Chr.16: 3495606-3495606 on Build GRCh38
rs377448632 Chr.16: 3495730-3495730 on Build GRCh38
rs775290099 Chr.16: 3495531-3495531 on Build GRCh38
rs751975898 Chr.16: 3495403-3495404 on Build GRCh38
rs374023919 Chr.16: 3495423-3495423 on Build GRCh38
rs9926502 Chr.16: 3495441-3495441 on Build GRCh38
rs770040336 Chr.16: 3495383-3495383 on Build GRCh38
rs769952351 Chr.16: 3495497-3495497 on Build GRCh38
rs952400071 Chr.16: 3495309-3495309 on Build GRCh38
rs1016500497 Chr.16: 3495341-3495341 on Build GRCh38
rs369623234 Chr.16: 3495369-3495369 on Build GRCh38
rs200404125 Chr.16: 3495370-3495370 on Build GRCh38
rs1055277266 Chr.16: 3495712-3495712 on Build GRCh38
rs757027113 Chr.16: 3495339-3495339 on Build GRCh38

View Full Product Details