Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3724682-3725197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREBBP
Entrez Gene ID
1387
Gene Name
CREB binding protein
Gene Aliases
CBP, KAT3A, RSTS
UniGene
Hs.459759
Gene Chromosome Location
Chr.16: 3725054-3880727 on Build GRCh38
Transcripts
NM_001079846.1 NM_004380.2 XM_005255124.4 XM_005255125.4 XM_006720848.3 XM_011522381.2 XM_017022944.1

SNP Detail

SNP ID Chromosome Location
rs901386544 Chr.16: 3724923-3724923 on Build GRCh38
rs1004335018 Chr.16: 3724916-3724916 on Build GRCh38
rs575897738 Chr.16: 3724909-3724909 on Build GRCh38
rs573389915 Chr.16: 3724791-3724791 on Build GRCh38
rs915313111 Chr.16: 3725019-3725019 on Build GRCh38
rs372416443 Chr.16: 3725015-3725015 on Build GRCh38
rs982055005 Chr.16: 3725014-3725014 on Build GRCh38
rs1046391548 Chr.16: 3724834-3724834 on Build GRCh38
rs1011140894 Chr.16: 3724731-3724731 on Build GRCh38
rs976113751 Chr.16: 3724752-3724752 on Build GRCh38
rs185579262 Chr.16: 3724981-3724981 on Build GRCh38
rs909696568 Chr.16: 3724775-3724776 on Build GRCh38
rs562603716 Chr.16: 3725042-3725042 on Build GRCh38
rs998782510 Chr.16: 3725040-3725040 on Build GRCh38
rs563754434 Chr.16: 3724967-3724967 on Build GRCh38
rs949335579 Chr.16: 3724810-3724810 on Build GRCh38
rs1048449629 Chr.16: 3724889-3724889 on Build GRCh38
rs988009333 Chr.16: 3724792-3724792 on Build GRCh38
rs531651621 Chr.16: 3725095-3725095 on Build GRCh38
rs888633810 Chr.16: 3724704-3724704 on Build GRCh38
rs971023542 Chr.16: 3724991-3724991 on Build GRCh38
rs567712178 Chr.16: 3725154-3725154 on Build GRCh38
rs555831341 Chr.16: 3724890-3724890 on Build GRCh38
rs542809937 Chr.16: 3725035-3725035 on Build GRCh38
rs751907603 Chr.16: 3724774-3724774 on Build GRCh38
rs138424740 Chr.16: 3724848-3724848 on Build GRCh38
rs1046795193 Chr.16: 3724833-3724833 on Build GRCh38
rs1007263647 Chr.16: 3724971-3724971 on Build GRCh38
rs189649723 Chr.16: 3725176-3725176 on Build GRCh38
rs962890212 Chr.16: 3724798-3724798 on Build GRCh38
rs1037576630 Chr.16: 3725018-3725018 on Build GRCh38
rs1015765343 Chr.16: 3724917-3724917 on Build GRCh38
rs900092240 Chr.16: 3724908-3724908 on Build GRCh38
rs1052630002 Chr.16: 3724713-3724714 on Build GRCh38
rs997853651 Chr.16: 3724742-3724742 on Build GRCh38
rs905205892 Chr.16: 3724849-3724849 on Build GRCh38
rs62037852 Chr.16: 3724771-3724771 on Build GRCh38
rs938005793 Chr.16: 3725006-3725006 on Build GRCh38
rs938073229 Chr.16: 3724883-3724883 on Build GRCh38
rs530705911 Chr.16: 3724978-3724978 on Build GRCh38
rs550518462 Chr.16: 3725033-3725033 on Build GRCh38
rs9392 Chr.16: 3725168-3725168 on Build GRCh38
rs953077313 Chr.16: 3724763-3724763 on Build GRCh38
rs140748621 Chr.16: 3724758-3724758 on Build GRCh38
rs926565482 Chr.16: 3725004-3725004 on Build GRCh38
rs901358249 Chr.16: 3725036-3725036 on Build GRCh38
rs888564138 Chr.16: 3724903-3724903 on Build GRCh38
rs953003756 Chr.16: 3724968-3724968 on Build GRCh38
rs868770766 Chr.16: 3724850-3724850 on Build GRCh38
rs1052972704 Chr.16: 3724868-3724868 on Build GRCh38
rs374582882 Chr.16: 3724852-3724855 on Build GRCh38
rs963703509 Chr.16: 3724739-3724739 on Build GRCh38
rs928023872 Chr.16: 3724841-3724841 on Build GRCh38
rs913743742 Chr.16: 3724989-3724989 on Build GRCh38
rs965696336 Chr.16: 3724751-3724751 on Build GRCh38
rs1016252100 Chr.16: 3724737-3724737 on Build GRCh38
rs886768121 Chr.16: 3725149-3725151 on Build GRCh38
rs572855867 Chr.16: 3724877-3724878 on Build GRCh38
rs781643334 Chr.16: 3724982-3724982 on Build GRCh38
rs900059419 Chr.16: 3725094-3725094 on Build GRCh38
rs570738685 Chr.16: 3725148-3725152 on Build GRCh38
rs550968281 Chr.16: 3725118-3725118 on Build GRCh38
rs890078518 Chr.16: 3725053-3725053 on Build GRCh38
rs375372620 Chr.16: 3725089-3725089 on Build GRCh38
rs957742172 Chr.16: 3725173-3725173 on Build GRCh38
rs372001056 Chr.16: 3724987-3724987 on Build GRCh38
rs915870069 Chr.16: 3724806-3724806 on Build GRCh38
rs183262998 Chr.16: 3724966-3724966 on Build GRCh38
rs991735644 Chr.16: 3725000-3725000 on Build GRCh38
rs940115341 Chr.16: 3725016-3725016 on Build GRCh38
rs559906969 Chr.16: 3725009-3725009 on Build GRCh38
rs946913180 Chr.16: 3724899-3724899 on Build GRCh38
rs982070270 Chr.16: 3724994-3724994 on Build GRCh38
rs748838577 Chr.16: 3725065-3725065 on Build GRCh38
rs913807129 Chr.16: 3724800-3724800 on Build GRCh38
rs368255171 Chr.16: 3724716-3724716 on Build GRCh38
rs751020506 Chr.16: 3724824-3724824 on Build GRCh38

View Full Product Details