Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4336906-4337445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIS2
Entrez Gene ID
84662
Gene Name
GLIS family zinc finger 2
Gene Aliases
NKL, NPHP7
UniGene
Hs.592087
Gene Chromosome Location
Chr.16: 4314761-4339597 on Build GRCh38
Transcripts
NM_001318918.1 NM_032575.2 XM_005255641.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6001946 c.1319G>A p.G440D Chr.16: 4337268-4337268 on Build GRCh38
4687192 c.1326C>T p.A442A Chr.16: 4337275-4337275 on Build GRCh38
5985943 c.1216G>A p.V406I Chr.16: 4337165-4337165 on Build GRCh38
3691005 c.1011G>A p.P337P Chr.16: 4336960-4336960 on Build GRCh38
6435563 c.1160G>A p.G387D Chr.16: 4337109-4337109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200720013 Chr.16: 4337054-4337054 on Build GRCh38
rs764220639 Chr.16: 4337135-4337135 on Build GRCh38
rs773863637 Chr.16: 4337124-4337124 on Build GRCh38
rs148236093 Chr.16: 4336944-4336944 on Build GRCh38
rs759416922 Chr.16: 4337126-4337126 on Build GRCh38
rs780482321 Chr.16: 4336994-4336994 on Build GRCh38
rs766960013 Chr.16: 4337189-4337189 on Build GRCh38
rs759658153 Chr.16: 4337408-4337408 on Build GRCh38
rs886667884 Chr.16: 4337239-4337239 on Build GRCh38
rs978655165 Chr.16: 4336986-4336986 on Build GRCh38
rs768415033 Chr.16: 4336949-4336949 on Build GRCh38
rs149580171 Chr.16: 4337413-4337413 on Build GRCh38
rs777660944 Chr.16: 4336972-4336972 on Build GRCh38
rs767819594 Chr.16: 4337275-4337275 on Build GRCh38
rs150464446 Chr.16: 4337299-4337299 on Build GRCh38
rs886726793 Chr.16: 4337082-4337082 on Build GRCh38
rs936495180 Chr.16: 4337276-4337276 on Build GRCh38
rs896610296 Chr.16: 4336955-4336955 on Build GRCh38
rs779624143 Chr.16: 4336979-4336979 on Build GRCh38
rs1019297382 Chr.16: 4337141-4337141 on Build GRCh38
rs1004844924 Chr.16: 4337113-4337113 on Build GRCh38
rs75495782 Chr.16: 4337208-4337208 on Build GRCh38
rs762088067 Chr.16: 4337253-4337253 on Build GRCh38
rs753957878 Chr.16: 4337060-4337060 on Build GRCh38
rs776593060 Chr.16: 4337252-4337252 on Build GRCh38
rs774339298 Chr.16: 4337036-4337036 on Build GRCh38
rs1032864995 Chr.16: 4337344-4337344 on Build GRCh38
rs749477442 Chr.16: 4337393-4337393 on Build GRCh38
rs750906253 Chr.16: 4337077-4337077 on Build GRCh38
rs755453297 Chr.16: 4337272-4337272 on Build GRCh38
rs746968228 Chr.16: 4337131-4337132 on Build GRCh38
rs759474185 Chr.16: 4337041-4337041 on Build GRCh38
rs769519610 Chr.16: 4337017-4337017 on Build GRCh38
rs532478795 Chr.16: 4336928-4336928 on Build GRCh38
rs757885506 Chr.16: 4336949-4336951 on Build GRCh38
rs776535774 Chr.16: 4337304-4337304 on Build GRCh38
rs770985872 Chr.16: 4337341-4337341 on Build GRCh38
rs771583432 Chr.16: 4336943-4336943 on Build GRCh38
rs965094283 Chr.16: 4337143-4337143 on Build GRCh38
rs752555380 Chr.16: 4337050-4337050 on Build GRCh38
rs762143375 Chr.16: 4337149-4337149 on Build GRCh38
rs746003129 Chr.16: 4337121-4337121 on Build GRCh38
rs764396667 Chr.16: 4337417-4337417 on Build GRCh38
rs559478567 Chr.16: 4336958-4336958 on Build GRCh38
rs746233571 Chr.16: 4336982-4336982 on Build GRCh38
rs994806881 Chr.16: 4336948-4336948 on Build GRCh38
rs750562871 Chr.16: 4337165-4337165 on Build GRCh38
rs201646165 Chr.16: 4336981-4336981 on Build GRCh38
rs779142808 Chr.16: 4337072-4337072 on Build GRCh38
rs1014392691 Chr.16: 4336942-4336942 on Build GRCh38
rs972495300 Chr.16: 4337147-4337147 on Build GRCh38
rs765597461 Chr.16: 4337257-4337257 on Build GRCh38
rs990748000 Chr.16: 4337226-4337226 on Build GRCh38
rs756868579 Chr.16: 4337287-4337287 on Build GRCh38
rs546324394 Chr.16: 4336953-4336953 on Build GRCh38
rs748789915 Chr.16: 4337114-4337114 on Build GRCh38
rs756369339 Chr.16: 4336971-4336971 on Build GRCh38
rs201182025 Chr.16: 4337362-4337362 on Build GRCh38
rs909495329 Chr.16: 4337066-4337066 on Build GRCh38
rs971469408 Chr.16: 4337410-4337410 on Build GRCh38
rs377385736 Chr.16: 4337407-4337407 on Build GRCh38
rs747706593 Chr.16: 4336996-4336996 on Build GRCh38
rs758012466 Chr.16: 4337367-4337367 on Build GRCh38
rs1053628540 Chr.16: 4337285-4337285 on Build GRCh38
rs539153071 Chr.16: 4337092-4337092 on Build GRCh38
rs140544340 Chr.16: 4336959-4336959 on Build GRCh38
rs751855769 Chr.16: 4337193-4337193 on Build GRCh38
rs568739931 Chr.16: 4336976-4336976 on Build GRCh38
rs3810817 Chr.16: 4337260-4337260 on Build GRCh38
rs906028584 Chr.16: 4337074-4337074 on Build GRCh38
rs778350155 Chr.16: 4337211-4337211 on Build GRCh38
rs8057701 Chr.16: 4337423-4337423 on Build GRCh38
rs746287094 Chr.16: 4336947-4336947 on Build GRCh38
rs901823188 Chr.16: 4337267-4337267 on Build GRCh38
rs889747669 Chr.16: 4337134-4337134 on Build GRCh38
rs767156568 Chr.16: 4337042-4337042 on Build GRCh38
rs746920921 Chr.16: 4337298-4337298 on Build GRCh38
rs1025422386 Chr.16: 4337162-4337162 on Build GRCh38
rs953853640 Chr.16: 4337137-4337137 on Build GRCh38
rs995065567 Chr.16: 4336956-4336956 on Build GRCh38
rs757861603 Chr.16: 4337296-4337296 on Build GRCh38
rs778583231 Chr.16: 4337291-4337291 on Build GRCh38
rs780241023 Chr.16: 4337088-4337088 on Build GRCh38
rs762497881 Chr.16: 4337027-4337027 on Build GRCh38
rs777441921 Chr.16: 4337389-4337389 on Build GRCh38
rs1006819423 Chr.16: 4337338-4337338 on Build GRCh38
rs747954752 Chr.16: 4337381-4337381 on Build GRCh38
rs748881616 Chr.16: 4337206-4337206 on Build GRCh38
rs934305633 Chr.16: 4337010-4337010 on Build GRCh38
rs780017382 Chr.16: 4336954-4336955 on Build GRCh38
rs752036048 Chr.16: 4337269-4337269 on Build GRCh38
rs751069276 Chr.16: 4337426-4337426 on Build GRCh38
rs528597262 Chr.16: 4337390-4337390 on Build GRCh38
rs1000731089 Chr.16: 4337130-4337130 on Build GRCh38
rs755435780 Chr.16: 4337194-4337194 on Build GRCh38
rs748007729 Chr.16: 4337322-4337322 on Build GRCh38
rs775114398 Chr.16: 4337129-4337129 on Build GRCh38
rs747442748 Chr.16: 4337091-4337091 on Build GRCh38
rs868422852 Chr.16: 4337063-4337063 on Build GRCh38
rs746927998 Chr.16: 4337245-4337245 on Build GRCh38
rs776929938 Chr.16: 4337139-4337139 on Build GRCh38
rs200814551 Chr.16: 4336960-4336960 on Build GRCh38
rs374664753 Chr.16: 4337375-4337375 on Build GRCh38
rs878855164 Chr.16: 4337157-4337158 on Build GRCh38
rs781541345 Chr.16: 4337205-4337205 on Build GRCh38
rs772073582 Chr.16: 4337128-4337128 on Build GRCh38
rs985172860 Chr.16: 4336988-4336989 on Build GRCh38
rs757481865 Chr.16: 4337062-4337062 on Build GRCh38
rs770128271 Chr.16: 4337328-4337328 on Build GRCh38
rs763089390 Chr.16: 4337339-4337339 on Build GRCh38
rs140403969 Chr.16: 4337425-4337425 on Build GRCh38
rs138285254 Chr.16: 4337352-4337352 on Build GRCh38
rs760766546 Chr.16: 4337053-4337053 on Build GRCh38
rs745353547 Chr.16: 4337292-4337292 on Build GRCh38
rs373198635 Chr.16: 4337337-4337337 on Build GRCh38
rs774316267 Chr.16: 4337342-4337342 on Build GRCh38
rs1027891466 Chr.16: 4336957-4336957 on Build GRCh38
rs768048691 Chr.16: 4337345-4337345 on Build GRCh38
rs761082862 Chr.16: 4337353-4337353 on Build GRCh38
rs758801563 Chr.16: 4337186-4337186 on Build GRCh38
rs777458149 Chr.16: 4337098-4337098 on Build GRCh38
rs753999588 Chr.16: 4336975-4336975 on Build GRCh38
rs551177412 Chr.16: 4336987-4336987 on Build GRCh38
rs766789991 Chr.16: 4337268-4337268 on Build GRCh38
rs761985295 Chr.16: 4337284-4337284 on Build GRCh38
rs776041769 Chr.16: 4337414-4337414 on Build GRCh38
rs202243000 Chr.16: 4337233-4337233 on Build GRCh38
rs779581418 Chr.16: 4337369-4337369 on Build GRCh38
rs760564252 Chr.16: 4337133-4337133 on Build GRCh38
rs554958933 Chr.16: 4337258-4337258 on Build GRCh38
rs752794127 Chr.16: 4336969-4336969 on Build GRCh38
rs1049299719 Chr.16: 4337081-4337081 on Build GRCh38
rs779447018 Chr.16: 4337297-4337297 on Build GRCh38
rs751071757 Chr.16: 4337373-4337373 on Build GRCh38
rs768444364 Chr.16: 4337251-4337251 on Build GRCh38
rs767775378 Chr.16: 4336967-4336967 on Build GRCh38
rs1031985166 Chr.16: 4336978-4336978 on Build GRCh38
rs761186959 Chr.16: 4337416-4337416 on Build GRCh38
rs866856287 Chr.16: 4337061-4337061 on Build GRCh38
rs758967813 Chr.16: 4337078-4337078 on Build GRCh38
rs988889512 Chr.16: 4337015-4337015 on Build GRCh38
rs749944065 Chr.16: 4336949-4336949 on Build GRCh38
rs199849300 Chr.16: 4337222-4337222 on Build GRCh38
rs769643827 Chr.16: 4337384-4337384 on Build GRCh38
rs753089156 Chr.16: 4337278-4337278 on Build GRCh38
rs150810544 Chr.16: 4337380-4337380 on Build GRCh38
rs952531818 Chr.16: 4337370-4337370 on Build GRCh38
rs764594630 Chr.16: 4336954-4336954 on Build GRCh38
rs749697026 Chr.16: 4336940-4336940 on Build GRCh38
rs745421196 Chr.16: 4337221-4337221 on Build GRCh38
rs765360012 Chr.16: 4337164-4337164 on Build GRCh38
rs865954949 Chr.16: 4337021-4337021 on Build GRCh38
rs772566892 Chr.16: 4337023-4337023 on Build GRCh38
rs930806642 Chr.16: 4337224-4337224 on Build GRCh38
rs770840077 Chr.16: 4337032-4337032 on Build GRCh38

View Full Product Details