Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 8621571-8622063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL22
Entrez Gene ID
79091
Gene Name
methyltransferase like 22
Gene Aliases
C16orf68
UniGene
Hs.306380
Gene Chromosome Location
Chr.16: 8621640-8673565 on Build GRCh38
Transcripts
NM_024109.3 NR_134608.1 XM_005255570.3 XM_005255571.2 XM_011522657.2 XM_017023659.1 XM_017023660.1 XM_017023661.1 XM_017023662.1

SNP Detail

SNP ID Chromosome Location
rs143770142 Chr.16: 8621714-8621715 on Build GRCh38
rs556133166 Chr.16: 8621921-8621921 on Build GRCh38
rs936241684 Chr.16: 8621600-8621600 on Build GRCh38
rs951948739 Chr.16: 8621648-8621648 on Build GRCh38
rs997440317 Chr.16: 8621809-8621809 on Build GRCh38
rs977889694 Chr.16: 8621855-8621855 on Build GRCh38
rs1031784279 Chr.16: 8621857-8621857 on Build GRCh38
rs957438007 Chr.16: 8622033-8622033 on Build GRCh38
rs866586116 Chr.16: 8621737-8621737 on Build GRCh38
rs749450504 Chr.16: 8621961-8621961 on Build GRCh38
rs754642652 Chr.16: 8621897-8621897 on Build GRCh38
rs1045334577 Chr.16: 8621721-8621721 on Build GRCh38
rs1054754186 Chr.16: 8621618-8621618 on Build GRCh38
rs906887055 Chr.16: 8621723-8621723 on Build GRCh38
rs930576604 Chr.16: 8621836-8621836 on Build GRCh38
rs541091183 Chr.16: 8622014-8622014 on Build GRCh38
rs538691966 Chr.16: 8621755-8621755 on Build GRCh38
rs1050511748 Chr.16: 8622042-8622042 on Build GRCh38
rs907793952 Chr.16: 8621748-8621748 on Build GRCh38
rs981694978 Chr.16: 8621900-8621900 on Build GRCh38
rs950809076 Chr.16: 8621647-8621647 on Build GRCh38
rs552885750 Chr.16: 8621722-8621722 on Build GRCh38
rs62019676 Chr.16: 8621665-8621665 on Build GRCh38
rs1055102986 Chr.16: 8621679-8621679 on Build GRCh38
rs971580835 Chr.16: 8621941-8621941 on Build GRCh38
rs796113672 Chr.16: 8621782-8621782 on Build GRCh38
rs895506068 Chr.16: 8621681-8621681 on Build GRCh38
rs905511869 Chr.16: 8621833-8621833 on Build GRCh38
rs1044322056 Chr.16: 8621701-8621701 on Build GRCh38
rs779711514 Chr.16: 8621607-8621607 on Build GRCh38
rs71377179 Chr.16: 8621843-8621843 on Build GRCh38
rs534331600 Chr.16: 8621685-8621685 on Build GRCh38
rs1049014566 Chr.16: 8621989-8621989 on Build GRCh38
rs375160598 Chr.16: 8621673-8621673 on Build GRCh38
rs887591542 Chr.16: 8621766-8621766 on Build GRCh38
rs551304521 Chr.16: 8621638-8621638 on Build GRCh38
rs1045386424 Chr.16: 8621624-8621624 on Build GRCh38
rs928302161 Chr.16: 8621923-8621923 on Build GRCh38
rs184680824 Chr.16: 8621905-8621905 on Build GRCh38
rs1028992913 Chr.16: 8621772-8621772 on Build GRCh38
rs1044487332 Chr.16: 8621805-8621805 on Build GRCh38
rs977566528 Chr.16: 8621672-8621672 on Build GRCh38
rs367647198 Chr.16: 8621622-8621622 on Build GRCh38
rs983519839 Chr.16: 8621650-8621650 on Build GRCh38
rs536829898 Chr.16: 8621661-8621661 on Build GRCh38
rs1029659549 Chr.16: 8621837-8621837 on Build GRCh38
rs979855698 Chr.16: 8621957-8621957 on Build GRCh38
rs778075187 Chr.16: 8621759-8621759 on Build GRCh38
rs979695060 Chr.16: 8621775-8621775 on Build GRCh38
rs556203687 Chr.16: 8621900-8621901 on Build GRCh38
rs997430413 Chr.16: 8622004-8622004 on Build GRCh38
rs1031186706 Chr.16: 8621909-8621909 on Build GRCh38
rs557075448 Chr.16: 8621800-8621800 on Build GRCh38
rs891107945 Chr.16: 8622019-8622019 on Build GRCh38
rs927174327 Chr.16: 8621812-8621812 on Build GRCh38
rs533439594 Chr.16: 8621633-8621633 on Build GRCh38
rs1032108676 Chr.16: 8622030-8622030 on Build GRCh38
rs555811780 Chr.16: 8621894-8621894 on Build GRCh38
rs1017547485 Chr.16: 8621658-8621658 on Build GRCh38
rs935480694 Chr.16: 8621779-8621779 on Build GRCh38
rs748257138 Chr.16: 8621987-8621987 on Build GRCh38
rs957699494 Chr.16: 8621858-8621858 on Build GRCh38
rs948940475 Chr.16: 8621889-8621889 on Build GRCh38
rs934440250 Chr.16: 8621675-8621676 on Build GRCh38
rs964581981 Chr.16: 8621803-8621803 on Build GRCh38
rs1038850712 Chr.16: 8621993-8621993 on Build GRCh38
rs968740015 Chr.16: 8621773-8621773 on Build GRCh38
rs142245646 Chr.16: 8621610-8621610 on Build GRCh38
rs966010375 Chr.16: 8621654-8621654 on Build GRCh38
rs894127086 Chr.16: 8621621-8621621 on Build GRCh38
rs1006084143 Chr.16: 8621778-8621778 on Build GRCh38
rs559667481 Chr.16: 8622024-8622024 on Build GRCh38
rs998837498 Chr.16: 8622028-8622028 on Build GRCh38
rs966040911 Chr.16: 8621754-8621754 on Build GRCh38
rs990649436 Chr.16: 8622034-8622034 on Build GRCh38
rs906939547 Chr.16: 8621628-8621628 on Build GRCh38
rs922599336 Chr.16: 8621662-8621662 on Build GRCh38
rs1047514581 Chr.16: 8621738-8621738 on Build GRCh38
rs936206466 Chr.16: 8621676-8621676 on Build GRCh38
rs1023082486 Chr.16: 8622036-8622036 on Build GRCh38
rs946909345 Chr.16: 8621664-8621664 on Build GRCh38
rs887508855 Chr.16: 8621981-8621981 on Build GRCh38
rs1040265892 Chr.16: 8622022-8622022 on Build GRCh38
rs964634096 Chr.16: 8621660-8621660 on Build GRCh38
rs980113251 Chr.16: 8621903-8621903 on Build GRCh38
rs8057713 Chr.16: 8621652-8621652 on Build GRCh38
rs1047924415 Chr.16: 8621949-8621949 on Build GRCh38
rs1017871777 Chr.16: 8621792-8621792 on Build GRCh38
rs948991351 Chr.16: 8621704-8621704 on Build GRCh38
rs973664595 Chr.16: 8621807-8621807 on Build GRCh38
rs916135127 Chr.16: 8621700-8621700 on Build GRCh38
rs576735719 Chr.16: 8621824-8621824 on Build GRCh38
rs544026186 Chr.16: 8621893-8621893 on Build GRCh38
rs577685553 Chr.16: 8621747-8621747 on Build GRCh38
rs75317654 Chr.16: 8621940-8621940 on Build GRCh38
rs916082873 Chr.16: 8621875-8621875 on Build GRCh38
rs1004988070 Chr.16: 8621739-8621739 on Build GRCh38
rs963298725 Chr.16: 8621801-8621801 on Build GRCh38
rs916965215 Chr.16: 8621783-8621783 on Build GRCh38
rs748582990 Chr.16: 8621590-8621590 on Build GRCh38
rs900368621 Chr.16: 8622003-8622003 on Build GRCh38
rs990310262 Chr.16: 8621860-8621860 on Build GRCh38

View Full Product Details