Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 22302591-22303104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR3E
Entrez Gene ID
55718
Gene Name
RNA polymerase III subunit E
Gene Aliases
RPC5, SIN
UniGene
Hs.460298
Gene Chromosome Location
Chr.16: 22297366-22335103 on Build GRCh38
Transcripts
NM_001258033.1 NM_001258034.1 NM_001258035.1 NM_001258036.1 NM_018119.3 NR_047581.1 XM_006721063.2 XM_011545892.1 XM_011545893.2 XM_017023443.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1749484 c.4G>A p.A2T Chr.16: 22302972-22302972 on Build GRCh38
6752445 c.19G>A p.D7N Chr.16: 22302987-22302987 on Build GRCh38
702295 c.28G>T p.V10L Chr.16: 22302996-22302996 on Build GRCh38
6564539 c.10G>A p.E4K Chr.16: 22302978-22302978 on Build GRCh38
3507664 c.21C>T p.D7D Chr.16: 22302989-22302989 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs367650640 Chr.16: 22302872-22302872 on Build GRCh38
rs975999621 Chr.16: 22302667-22302667 on Build GRCh38
rs374423645 Chr.16: 22302818-22302818 on Build GRCh38
rs977326184 Chr.16: 22303041-22303041 on Build GRCh38
rs990422366 Chr.16: 22303075-22303075 on Build GRCh38
rs968113683 Chr.16: 22302826-22302826 on Build GRCh38
rs200785701 Chr.16: 22303033-22303033 on Build GRCh38
rs971311616 Chr.16: 22302798-22302798 on Build GRCh38
rs11648734 Chr.16: 22302722-22302722 on Build GRCh38
rs377481250 Chr.16: 22302909-22302909 on Build GRCh38
rs372095706 Chr.16: 22303031-22303031 on Build GRCh38
rs745851713 Chr.16: 22302787-22302787 on Build GRCh38
rs759084624 Chr.16: 22303030-22303030 on Build GRCh38
rs375747112 Chr.16: 22303023-22303023 on Build GRCh38
rs376251397 Chr.16: 22302934-22302934 on Build GRCh38
rs917163273 Chr.16: 22302785-22302785 on Build GRCh38
rs996791880 Chr.16: 22302624-22302624 on Build GRCh38
rs755070175 Chr.16: 22303052-22303052 on Build GRCh38
rs539137750 Chr.16: 22302721-22302721 on Build GRCh38
rs931400178 Chr.16: 22302825-22302825 on Build GRCh38
rs761668795 Chr.16: 22303037-22303037 on Build GRCh38
rs757174494 Chr.16: 22302946-22302946 on Build GRCh38
rs149111360 Chr.16: 22302959-22302959 on Build GRCh38
rs530220765 Chr.16: 22303014-22303014 on Build GRCh38
rs931622543 Chr.16: 22302692-22302692 on Build GRCh38
rs991598016 Chr.16: 22302777-22302777 on Build GRCh38
rs922952773 Chr.16: 22302680-22302680 on Build GRCh38
rs930153191 Chr.16: 22302950-22302950 on Build GRCh38
rs773689740 Chr.16: 22302921-22302921 on Build GRCh38
rs781386213 Chr.16: 22302961-22302961 on Build GRCh38
rs1050455076 Chr.16: 22302895-22302895 on Build GRCh38
rs536622945 Chr.16: 22302682-22302682 on Build GRCh38
rs141261021 Chr.16: 22302710-22302710 on Build GRCh38
rs201915761 Chr.16: 22303044-22303044 on Build GRCh38
rs770114475 Chr.16: 22303004-22303004 on Build GRCh38
rs555746095 Chr.16: 22302883-22302883 on Build GRCh38
rs773867067 Chr.16: 22302928-22302928 on Build GRCh38
rs367565578 Chr.16: 22303028-22303028 on Build GRCh38
rs544745742 Chr.16: 22302917-22302917 on Build GRCh38
rs1050203861 Chr.16: 22302700-22302700 on Build GRCh38
rs147625767 Chr.16: 22302833-22302833 on Build GRCh38
rs147804189 Chr.16: 22302675-22302675 on Build GRCh38
rs773526479 Chr.16: 22303009-22303009 on Build GRCh38
rs890743893 Chr.16: 22302666-22302666 on Build GRCh38
rs372714723 Chr.16: 22302933-22302933 on Build GRCh38
rs765154969 Chr.16: 22303040-22303040 on Build GRCh38
rs953282792 Chr.16: 22303043-22303043 on Build GRCh38
rs997355440 Chr.16: 22303038-22303038 on Build GRCh38
rs575049078 Chr.16: 22302852-22302852 on Build GRCh38
rs375097958 Chr.16: 22302949-22302949 on Build GRCh38
rs896527066 Chr.16: 22302809-22302809 on Build GRCh38
rs765841054 Chr.16: 22302922-22302922 on Build GRCh38
rs747491220 Chr.16: 22302840-22302840 on Build GRCh38
rs753792184 Chr.16: 22302942-22302942 on Build GRCh38
rs369243430 Chr.16: 22302920-22302920 on Build GRCh38
rs773742023 Chr.16: 22303029-22303029 on Build GRCh38
rs375024210 Chr.16: 22302985-22302985 on Build GRCh38
rs112102365 Chr.16: 22302951-22302951 on Build GRCh38
rs769767909 Chr.16: 22302846-22302846 on Build GRCh38
rs199890611 Chr.16: 22302938-22302938 on Build GRCh38
rs751656775 Chr.16: 22303046-22303046 on Build GRCh38
rs559671493 Chr.16: 22303034-22303034 on Build GRCh38
rs1050673160 Chr.16: 22302643-22302643 on Build GRCh38
rs372092330 Chr.16: 22303055-22303055 on Build GRCh38
rs994617012 Chr.16: 22302712-22302712 on Build GRCh38
rs977212049 Chr.16: 22302799-22302799 on Build GRCh38
rs1031273044 Chr.16: 22302844-22302844 on Build GRCh38
rs183791955 Chr.16: 22302711-22302711 on Build GRCh38
rs760583348 Chr.16: 22303032-22303032 on Build GRCh38
rs781435087 Chr.16: 22303053-22303053 on Build GRCh38
rs534989378 Chr.16: 22302822-22302822 on Build GRCh38
rs575732785 Chr.16: 22302916-22302916 on Build GRCh38

View Full Product Details