Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28879385-28879877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2A1
Entrez Gene ID
487
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 1
Gene Aliases
ATP2A, SERCA1
UniGene
Hs.657344
Gene Chromosome Location
Chr.16: 28878488-28904509 on Build GRCh38
Transcripts
NM_004320.4 NM_173201.3 NR_046287.1 NR_046288.1 NR_046289.1 NR_046290.1

Gene Symbol
ATP2A1-AS1
Entrez Gene ID
100289092
Gene Name
ATP2A1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.666631
Gene Chromosome Location
Chr.16: 28878957-28879921 on Build GRCh38
Transcripts
NM_004320.4 NM_173201.3 NR_046287.1 NR_046288.1 NR_046289.1 NR_046290.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4466744 c.144C>T p.T48T Chr.16: 28879508-28879508 on Build GRCh38
279056 c.153G>T p.E51D Chr.16: 28879517-28879517 on Build GRCh38
3817725 c.193C>T p.L65F Chr.16: 28879557-28879557 on Build GRCh38
5203934 c.193C>T p.L65F Chr.16: 28879557-28879557 on Build GRCh38
969561 c.139A>G p.K47E Chr.16: 28879503-28879503 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372102242 Chr.16: 28879592-28879592 on Build GRCh38
rs376632597 Chr.16: 28879616-28879616 on Build GRCh38
rs568186038 Chr.16: 28879692-28879692 on Build GRCh38
rs536744428 Chr.16: 28879728-28879728 on Build GRCh38
rs972241101 Chr.16: 28879671-28879671 on Build GRCh38
rs754157591 Chr.16: 28879511-28879511 on Build GRCh38
rs367744011 Chr.16: 28879624-28879624 on Build GRCh38
rs778511864 Chr.16: 28879515-28879515 on Build GRCh38
rs534844795 Chr.16: 28879651-28879651 on Build GRCh38
rs759667208 Chr.16: 28879495-28879495 on Build GRCh38
rs998602244 Chr.16: 28879701-28879701 on Build GRCh38
rs984970776 Chr.16: 28879590-28879590 on Build GRCh38
rs888015548 Chr.16: 28879806-28879806 on Build GRCh38
rs1031500896 Chr.16: 28879529-28879529 on Build GRCh38
rs771329836 Chr.16: 28879546-28879546 on Build GRCh38
rs754247348 Chr.16: 28879604-28879604 on Build GRCh38
rs756940046 Chr.16: 28879514-28879514 on Build GRCh38
rs1019275964 Chr.16: 28879521-28879521 on Build GRCh38
rs943050403 Chr.16: 28879689-28879689 on Build GRCh38
rs757106087 Chr.16: 28879584-28879584 on Build GRCh38
rs771609729 Chr.16: 28879519-28879519 on Build GRCh38
rs180863892 Chr.16: 28879505-28879505 on Build GRCh38
rs751731229 Chr.16: 28879494-28879494 on Build GRCh38
rs375724457 Chr.16: 28879449-28879449 on Build GRCh38
rs1016672316 Chr.16: 28879848-28879848 on Build GRCh38
rs554451058 Chr.16: 28879675-28879675 on Build GRCh38
rs764938642 Chr.16: 28879608-28879608 on Build GRCh38
rs896779867 Chr.16: 28879746-28879746 on Build GRCh38
rs201212818 Chr.16: 28879497-28879497 on Build GRCh38
rs879300947 Chr.16: 28879682-28879682 on Build GRCh38
rs774708450 Chr.16: 28879552-28879552 on Build GRCh38
rs963471705 Chr.16: 28879852-28879852 on Build GRCh38
rs909302242 Chr.16: 28879634-28879634 on Build GRCh38
rs776599039 Chr.16: 28879524-28879524 on Build GRCh38
rs190863438 Chr.16: 28879472-28879472 on Build GRCh38
rs1052963042 Chr.16: 28879725-28879725 on Build GRCh38
rs368264574 Chr.16: 28879629-28879629 on Build GRCh38
rs757934586 Chr.16: 28879623-28879623 on Build GRCh38
rs761671172 Chr.16: 28879527-28879527 on Build GRCh38
rs777831324 Chr.16: 28879473-28879473 on Build GRCh38
rs970340546 Chr.16: 28879820-28879820 on Build GRCh38
rs778222175 Chr.16: 28879508-28879508 on Build GRCh38
rs947191960 Chr.16: 28879690-28879690 on Build GRCh38
rs1051978010 Chr.16: 28879687-28879687 on Build GRCh38
rs575679420 Chr.16: 28879714-28879714 on Build GRCh38
rs148925426 Chr.16: 28879523-28879523 on Build GRCh38
rs993717161 Chr.16: 28879563-28879563 on Build GRCh38
rs993853708 Chr.16: 28879762-28879762 on Build GRCh38
rs952168452 Chr.16: 28879583-28879583 on Build GRCh38
rs759763916 Chr.16: 28879570-28879570 on Build GRCh38
rs201630491 Chr.16: 28879461-28879461 on Build GRCh38
rs754984005 Chr.16: 28879628-28879628 on Build GRCh38
rs778795132 Chr.16: 28879603-28879604 on Build GRCh38
rs556589307 Chr.16: 28879742-28879742 on Build GRCh38
rs767974799 Chr.16: 28879496-28879496 on Build GRCh38
rs904305803 Chr.16: 28879697-28879697 on Build GRCh38
rs763292322 Chr.16: 28879491-28879491 on Build GRCh38
rs752669373 Chr.16: 28879572-28879572 on Build GRCh38
rs892936101 Chr.16: 28879405-28879405 on Build GRCh38
rs1014160955 Chr.16: 28879566-28879566 on Build GRCh38
rs561462745 Chr.16: 28879424-28879424 on Build GRCh38
rs745438439 Chr.16: 28879516-28879516 on Build GRCh38
rs1023131105 Chr.16: 28879850-28879850 on Build GRCh38
rs1014069201 Chr.16: 28879739-28879739 on Build GRCh38
rs746462326 Chr.16: 28879625-28879625 on Build GRCh38
rs1002218249 Chr.16: 28879842-28879842 on Build GRCh38
rs753865718 Chr.16: 28879557-28879559 on Build GRCh38
rs910769943 Chr.16: 28879688-28879688 on Build GRCh38
rs767648658 Chr.16: 28879571-28879571 on Build GRCh38
rs919425306 Chr.16: 28879681-28879681 on Build GRCh38
rs112987448 Chr.16: 28879635-28879635 on Build GRCh38
rs1041201132 Chr.16: 28879735-28879735 on Build GRCh38
rs867846169 Chr.16: 28879712-28879712 on Build GRCh38
rs764473732 Chr.16: 28879600-28879600 on Build GRCh38
rs760456828 Chr.16: 28879473-28879473 on Build GRCh38
rs543105584 Chr.16: 28879814-28879814 on Build GRCh38
rs768506490 Chr.16: 28879477-28879477 on Build GRCh38
rs201182451 Chr.16: 28879606-28879606 on Build GRCh38
rs775488442 Chr.16: 28879471-28879471 on Build GRCh38
rs766669977 Chr.16: 28879492-28879492 on Build GRCh38
rs776235380 Chr.16: 28879482-28879482 on Build GRCh38
rs565808029 Chr.16: 28879642-28879642 on Build GRCh38

View Full Product Details