Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29744426-29744964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf54
Entrez Gene ID
283897
Gene Name
chromosome 16 open reading frame 54
Gene Aliases
-
UniGene
Hs.331095
Gene Chromosome Location
Chr.16: 29742463-29746019 on Build GRCh38
Transcripts
NM_175900.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5671096 c.47C>A p.P16H Chr.16: 29744905-29744905 on Build GRCh38
5587331 c.232G>A p.G78R Chr.16: 29744720-29744720 on Build GRCh38
969631 c.164G>A p.R55H Chr.16: 29744788-29744788 on Build GRCh38
5580398 c.235G>A p.E79K Chr.16: 29744717-29744717 on Build GRCh38
1377244 c.317G>A p.R106Q Chr.16: 29744635-29744635 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372993933 Chr.16: 29744690-29744690 on Build GRCh38
rs749227397 Chr.16: 29744760-29744760 on Build GRCh38
rs370898520 Chr.16: 29744789-29744789 on Build GRCh38
rs774872519 Chr.16: 29744920-29744920 on Build GRCh38
rs761708869 Chr.16: 29744571-29744571 on Build GRCh38
rs200945943 Chr.16: 29744593-29744593 on Build GRCh38
rs779014114 Chr.16: 29744876-29744876 on Build GRCh38
rs957256763 Chr.16: 29744472-29744472 on Build GRCh38
rs758099729 Chr.16: 29744857-29744858 on Build GRCh38
rs746268461 Chr.16: 29744802-29744802 on Build GRCh38
rs1002996755 Chr.16: 29744627-29744627 on Build GRCh38
rs781355195 Chr.16: 29744852-29744852 on Build GRCh38
rs766804316 Chr.16: 29744824-29744824 on Build GRCh38
rs376393110 Chr.16: 29744909-29744909 on Build GRCh38
rs752887121 Chr.16: 29744855-29744855 on Build GRCh38
rs868589963 Chr.16: 29744695-29744695 on Build GRCh38
rs747945510 Chr.16: 29744636-29744636 on Build GRCh38
rs759257997 Chr.16: 29744670-29744670 on Build GRCh38
rs1009448985 Chr.16: 29744768-29744768 on Build GRCh38
rs768920094 Chr.16: 29744597-29744597 on Build GRCh38
rs770832221 Chr.16: 29744767-29744767 on Build GRCh38
rs765619918 Chr.16: 29744715-29744715 on Build GRCh38
rs760077380 Chr.16: 29744921-29744921 on Build GRCh38
rs755129808 Chr.16: 29744740-29744740 on Build GRCh38
rs201110852 Chr.16: 29744777-29744777 on Build GRCh38
rs569754860 Chr.16: 29744529-29744529 on Build GRCh38
rs760706108 Chr.16: 29744750-29744750 on Build GRCh38
rs762540464 Chr.16: 29744458-29744458 on Build GRCh38
rs757031853 Chr.16: 29744932-29744932 on Build GRCh38
rs780034430 Chr.16: 29744573-29744573 on Build GRCh38
rs769664424 Chr.16: 29744752-29744752 on Build GRCh38
rs750807770 Chr.16: 29744807-29744807 on Build GRCh38
rs760386935 Chr.16: 29744904-29744904 on Build GRCh38
rs944203939 Chr.16: 29744687-29744687 on Build GRCh38
rs373101692 Chr.16: 29744532-29744532 on Build GRCh38
rs143391821 Chr.16: 29744686-29744686 on Build GRCh38
rs779892161 Chr.16: 29744903-29744904 on Build GRCh38
rs745884376 Chr.16: 29744552-29744552 on Build GRCh38
rs1035799601 Chr.16: 29744843-29744843 on Build GRCh38
rs1020766444 Chr.16: 29744594-29744594 on Build GRCh38
rs749041572 Chr.16: 29744864-29744864 on Build GRCh38
rs758874736 Chr.16: 29744728-29744728 on Build GRCh38
rs578093410 Chr.16: 29744739-29744739 on Build GRCh38
rs916939664 Chr.16: 29744702-29744702 on Build GRCh38
rs779355314 Chr.16: 29744466-29744466 on Build GRCh38
rs754320429 Chr.16: 29744528-29744528 on Build GRCh38
rs866437101 Chr.16: 29744641-29744641 on Build GRCh38
rs755076456 Chr.16: 29744844-29744844 on Build GRCh38
rs779485804 Chr.16: 29744551-29744551 on Build GRCh38
rs778891048 Chr.16: 29744936-29744936 on Build GRCh38
rs754718868 Chr.16: 29744483-29744483 on Build GRCh38
rs969741387 Chr.16: 29744926-29744926 on Build GRCh38
rs968310813 Chr.16: 29744904-29744904 on Build GRCh38
rs765900368 Chr.16: 29744465-29744465 on Build GRCh38
rs889798343 Chr.16: 29744804-29744804 on Build GRCh38
rs1047192975 Chr.16: 29744470-29744470 on Build GRCh38
rs766863718 Chr.16: 29744919-29744919 on Build GRCh38
rs745329582 Chr.16: 29744900-29744900 on Build GRCh38
rs573800426 Chr.16: 29744823-29744823 on Build GRCh38
rs996514684 Chr.16: 29744562-29744562 on Build GRCh38
rs775311706 Chr.16: 29744784-29744784 on Build GRCh38
rs933787869 Chr.16: 29744629-29744629 on Build GRCh38
rs374762176 Chr.16: 29744639-29744639 on Build GRCh38
rs753940451 Chr.16: 29744776-29744776 on Build GRCh38
rs535090241 Chr.16: 29744656-29744656 on Build GRCh38
rs374180124 Chr.16: 29744918-29744918 on Build GRCh38
rs543740743 Chr.16: 29744520-29744520 on Build GRCh38
rs998295941 Chr.16: 29744782-29744782 on Build GRCh38
rs775186423 Chr.16: 29744903-29744903 on Build GRCh38
rs768256621 Chr.16: 29744908-29744908 on Build GRCh38
rs975367643 Chr.16: 29744479-29744479 on Build GRCh38
rs539006493 Chr.16: 29744749-29744749 on Build GRCh38
rs908310717 Chr.16: 29744500-29744500 on Build GRCh38
rs767282348 Chr.16: 29744681-29744681 on Build GRCh38
rs376044334 Chr.16: 29744756-29744756 on Build GRCh38
rs770842982 Chr.16: 29744874-29744874 on Build GRCh38
rs773782853 Chr.16: 29744662-29744662 on Build GRCh38
rs943755188 Chr.16: 29744509-29744509 on Build GRCh38
rs781408664 Chr.16: 29744747-29744747 on Build GRCh38
rs771675299 Chr.16: 29744902-29744902 on Build GRCh38
rs756355279 Chr.16: 29744928-29744928 on Build GRCh38
rs756341447 Chr.16: 29744859-29744859 on Build GRCh38
rs752832049 Chr.16: 29744924-29744924 on Build GRCh38
rs750907545 Chr.16: 29744727-29744727 on Build GRCh38
rs780165473 Chr.16: 29744735-29744735 on Build GRCh38
rs200439714 Chr.16: 29744780-29744780 on Build GRCh38
rs773226713 Chr.16: 29744654-29744654 on Build GRCh38
rs145619320 Chr.16: 29744862-29744862 on Build GRCh38
rs796580121 Chr.16: 29744635-29744635 on Build GRCh38
rs551608693 Chr.16: 29744466-29744466 on Build GRCh38
rs1015787790 Chr.16: 29744929-29744929 on Build GRCh38
rs140490612 Chr.16: 29744804-29744804 on Build GRCh38
rs765609481 Chr.16: 29744805-29744805 on Build GRCh38
rs113304298 Chr.16: 29744696-29744696 on Build GRCh38
rs764532004 Chr.16: 29744493-29744493 on Build GRCh38
rs369694968 Chr.16: 29744798-29744798 on Build GRCh38
rs751584562 Chr.16: 29744840-29744840 on Build GRCh38
rs763508571 Chr.16: 29744809-29744809 on Build GRCh38
rs370513110 Chr.16: 29744931-29744931 on Build GRCh38
rs776410013 Chr.16: 29744633-29744633 on Build GRCh38
rs932328042 Chr.16: 29744738-29744738 on Build GRCh38
rs567404785 Chr.16: 29744607-29744607 on Build GRCh38
rs982919079 Chr.16: 29744680-29744680 on Build GRCh38
rs537044537 Chr.16: 29744569-29744569 on Build GRCh38
rs955958636 Chr.16: 29744911-29744911 on Build GRCh38
rs200964810 Chr.16: 29744779-29744779 on Build GRCh38
rs762332624 Chr.16: 29744655-29744655 on Build GRCh38

View Full Product Details