Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30374670-30375069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYLPF
Entrez Gene ID
29895
Gene Name
myosin light chain, phosphorylatable, fast skeletal muscle
Gene Aliases
HUMMLC2B, MLC2B, MRLC2, MYL11
UniGene
Hs.50889
Gene Chromosome Location
Chr.16: 30370934-30377991 on Build GRCh38
Transcripts
NM_001324458.1 NM_001324459.1 NM_013292.4

SNP Detail

SNP ID Chromosome Location
rs919478591 Chr.16: 30374992-30374992 on Build GRCh38
rs773789427 Chr.16: 30374882-30374882 on Build GRCh38
rs910551628 Chr.16: 30375043-30375043 on Build GRCh38
rs376625822 Chr.16: 30374844-30374844 on Build GRCh38
rs1007625709 Chr.16: 30374765-30374765 on Build GRCh38
rs779864271 Chr.16: 30374754-30374754 on Build GRCh38
rs966098125 Chr.16: 30374994-30374994 on Build GRCh38
rs543225974 Chr.16: 30374899-30374899 on Build GRCh38
rs777631350 Chr.16: 30374952-30374952 on Build GRCh38
rs747362091 Chr.16: 30375018-30375018 on Build GRCh38
rs976102750 Chr.16: 30375004-30375004 on Build GRCh38
rs375837285 Chr.16: 30374846-30374846 on Build GRCh38
rs765705752 Chr.16: 30375042-30375042 on Build GRCh38
rs766840213 Chr.16: 30374912-30374912 on Build GRCh38
rs966128778 Chr.16: 30374729-30374729 on Build GRCh38
rs745543663 Chr.16: 30374880-30374881 on Build GRCh38
rs545808529 Chr.16: 30374824-30374824 on Build GRCh38
rs370233751 Chr.16: 30374847-30374847 on Build GRCh38
rs932484130 Chr.16: 30374772-30374772 on Build GRCh38
rs868560822 Chr.16: 30375029-30375029 on Build GRCh38
rs1027885249 Chr.16: 30374880-30374880 on Build GRCh38
rs576201797 Chr.16: 30374866-30374866 on Build GRCh38
rs1043322911 Chr.16: 30374771-30374771 on Build GRCh38
rs556334693 Chr.16: 30374711-30374711 on Build GRCh38
rs755809113 Chr.16: 30374843-30374843 on Build GRCh38
rs773891259 Chr.16: 30374815-30374815 on Build GRCh38
rs570179540 Chr.16: 30374704-30374704 on Build GRCh38
rs200076406 Chr.16: 30374823-30374823 on Build GRCh38
rs749101951 Chr.16: 30374786-30374786 on Build GRCh38
rs933282043 Chr.16: 30374707-30374707 on Build GRCh38
rs915160937 Chr.16: 30374778-30374779 on Build GRCh38
rs8046445 Chr.16: 30374941-30374941 on Build GRCh38
rs902244136 Chr.16: 30374829-30374829 on Build GRCh38
rs564303747 Chr.16: 30374830-30374830 on Build GRCh38
rs1049626809 Chr.16: 30374775-30374775 on Build GRCh38
rs372644320 Chr.16: 30374839-30374839 on Build GRCh38
rs772128467 Chr.16: 30374788-30374788 on Build GRCh38
rs904350249 Chr.16: 30374774-30374774 on Build GRCh38
rs761344072 Chr.16: 30374867-30374867 on Build GRCh38
rs763419150 Chr.16: 30374897-30374897 on Build GRCh38
rs972326587 Chr.16: 30374951-30374951 on Build GRCh38
rs778360661 Chr.16: 30374838-30374839 on Build GRCh38
rs535392155 Chr.16: 30374735-30374735 on Build GRCh38
rs771904411 Chr.16: 30374851-30374851 on Build GRCh38
rs760098826 Chr.16: 30375015-30375015 on Build GRCh38
rs187398906 Chr.16: 30374705-30374705 on Build GRCh38
rs375244676 Chr.16: 30374852-30374852 on Build GRCh38
rs952042228 Chr.16: 30374713-30374713 on Build GRCh38
rs540909955 Chr.16: 30374724-30374724 on Build GRCh38
rs1013847588 Chr.16: 30374842-30374842 on Build GRCh38
rs922479643 Chr.16: 30374771-30374772 on Build GRCh38
rs976537741 Chr.16: 30374762-30374762 on Build GRCh38
rs771743120 Chr.16: 30374937-30374937 on Build GRCh38
rs770499579 Chr.16: 30374845-30374845 on Build GRCh38
rs1003332189 Chr.16: 30374837-30374837 on Build GRCh38
rs369039939 Chr.16: 30374806-30374806 on Build GRCh38
rs953879544 Chr.16: 30375022-30375022 on Build GRCh38
rs922022460 Chr.16: 30375012-30375012 on Build GRCh38
rs889225961 Chr.16: 30374730-30374730 on Build GRCh38
rs747678478 Chr.16: 30374929-30374929 on Build GRCh38

View Full Product Details