Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31084699-31085194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS53
Entrez Gene ID
339105
Gene Name
protease, serine 53
Gene Aliases
POL3S, UNQ308
UniGene
Hs.569575
Gene Chromosome Location
Chr.16: 31083424-31090035 on Build GRCh38
Transcripts
NM_001039503.2 XM_011545816.1 XM_011545817.1 XM_011545818.2 XM_011545819.1 XM_011545820.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3370407 c.982G>T p.G328* Chr.16: 31085162-31085162 on Build GRCh38
6029058 c.1036C>T p.R346C Chr.16: 31085023-31085023 on Build GRCh38
6016156 c.1060G>A p.V354I Chr.16: 31084999-31084999 on Build GRCh38
4060364 c.1080G>A p.P360P Chr.16: 31084979-31084979 on Build GRCh38
5448853 c.1206C>T p.C402C Chr.16: 31084853-31084853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984219627 Chr.16: 31085058-31085058 on Build GRCh38
rs745937671 Chr.16: 31085053-31085053 on Build GRCh38
rs762701898 Chr.16: 31085105-31085105 on Build GRCh38
rs767272926 Chr.16: 31084731-31084731 on Build GRCh38
rs754935653 Chr.16: 31084730-31084730 on Build GRCh38
rs748776465 Chr.16: 31084770-31084770 on Build GRCh38
rs772012439 Chr.16: 31084938-31084938 on Build GRCh38
rs370867880 Chr.16: 31085121-31085121 on Build GRCh38
rs757463088 Chr.16: 31085045-31085045 on Build GRCh38
rs778049800 Chr.16: 31085027-31085027 on Build GRCh38
rs763780353 Chr.16: 31084851-31084851 on Build GRCh38
rs375690216 Chr.16: 31085162-31085162 on Build GRCh38
rs903270955 Chr.16: 31084744-31084744 on Build GRCh38
rs1010739256 Chr.16: 31084912-31084912 on Build GRCh38
rs764965262 Chr.16: 31085086-31085086 on Build GRCh38
rs550474298 Chr.16: 31084864-31084864 on Build GRCh38
rs770954506 Chr.16: 31084815-31084815 on Build GRCh38
rs182403228 Chr.16: 31084733-31084733 on Build GRCh38
rs774398454 Chr.16: 31084930-31084930 on Build GRCh38
rs72785539 Chr.16: 31085174-31085174 on Build GRCh38
rs765570692 Chr.16: 31084819-31084819 on Build GRCh38
rs373736818 Chr.16: 31084794-31084794 on Build GRCh38
rs761742723 Chr.16: 31085082-31085082 on Build GRCh38
rs371773654 Chr.16: 31085147-31085147 on Build GRCh38
rs199760254 Chr.16: 31085168-31085168 on Build GRCh38
rs753699591 Chr.16: 31084975-31084975 on Build GRCh38
rs568817652 Chr.16: 31084916-31084916 on Build GRCh38
rs766507982 Chr.16: 31084925-31084926 on Build GRCh38
rs759408873 Chr.16: 31084829-31084829 on Build GRCh38
rs940292253 Chr.16: 31084809-31084809 on Build GRCh38
rs1054610897 Chr.16: 31085087-31085087 on Build GRCh38
rs756875911 Chr.16: 31084772-31084772 on Build GRCh38
rs201947413 Chr.16: 31085068-31085068 on Build GRCh38
rs779145638 Chr.16: 31085046-31085046 on Build GRCh38
rs767410045 Chr.16: 31084832-31084832 on Build GRCh38
rs376368418 Chr.16: 31084951-31084951 on Build GRCh38
rs893599375 Chr.16: 31084886-31084886 on Build GRCh38
rs149180816 Chr.16: 31084792-31084792 on Build GRCh38
rs370217577 Chr.16: 31084985-31084985 on Build GRCh38
rs376906210 Chr.16: 31084791-31084791 on Build GRCh38
rs771997153 Chr.16: 31085067-31085067 on Build GRCh38
rs773359736 Chr.16: 31084797-31084797 on Build GRCh38
rs559980823 Chr.16: 31084721-31084721 on Build GRCh38
rs763730838 Chr.16: 31084968-31084968 on Build GRCh38
rs747453677 Chr.16: 31084783-31084783 on Build GRCh38
rs376206757 Chr.16: 31085139-31085139 on Build GRCh38
rs1048582936 Chr.16: 31084790-31084790 on Build GRCh38
rs962136507 Chr.16: 31084868-31084868 on Build GRCh38
rs765051260 Chr.16: 31084983-31084983 on Build GRCh38
rs749708168 Chr.16: 31085035-31085035 on Build GRCh38
rs774414921 Chr.16: 31084828-31084828 on Build GRCh38
rs775659363 Chr.16: 31084779-31084779 on Build GRCh38
rs371518777 Chr.16: 31084742-31084742 on Build GRCh38
rs920449776 Chr.16: 31084942-31084942 on Build GRCh38
rs908711750 Chr.16: 31085068-31085073 on Build GRCh38
rs775337954 Chr.16: 31084941-31084941 on Build GRCh38
rs779270659 Chr.16: 31085156-31085156 on Build GRCh38
rs746037110 Chr.16: 31085158-31085158 on Build GRCh38
rs779985396 Chr.16: 31085161-31085161 on Build GRCh38
rs759678499 Chr.16: 31084935-31084935 on Build GRCh38
rs368384747 Chr.16: 31085113-31085113 on Build GRCh38
rs770008199 Chr.16: 31084913-31084913 on Build GRCh38
rs866819500 Chr.16: 31084981-31084981 on Build GRCh38
rs779642455 Chr.16: 31085019-31085019 on Build GRCh38
rs535842116 Chr.16: 31084927-31084927 on Build GRCh38
rs748527691 Chr.16: 31084894-31084894 on Build GRCh38
rs772907169 Chr.16: 31084912-31084913 on Build GRCh38
rs900850086 Chr.16: 31084821-31084821 on Build GRCh38
rs768698044 Chr.16: 31085074-31085074 on Build GRCh38
rs751060113 Chr.16: 31085065-31085066 on Build GRCh38
rs373472672 Chr.16: 31084872-31084872 on Build GRCh38
rs375546280 Chr.16: 31084816-31084816 on Build GRCh38
rs749925191 Chr.16: 31084991-31084991 on Build GRCh38
rs758659965 Chr.16: 31085160-31085160 on Build GRCh38
rs776760159 Chr.16: 31085165-31085165 on Build GRCh38
rs772954131 Chr.16: 31085088-31085088 on Build GRCh38
rs753491933 Chr.16: 31084853-31084853 on Build GRCh38
rs752444691 Chr.16: 31084732-31084732 on Build GRCh38
rs751441935 Chr.16: 31084999-31084999 on Build GRCh38
rs985980206 Chr.16: 31084957-31084957 on Build GRCh38
rs756974227 Chr.16: 31084863-31084863 on Build GRCh38
rs374546739 Chr.16: 31085081-31085081 on Build GRCh38
rs762579729 Chr.16: 31084916-31084918 on Build GRCh38
rs781592805 Chr.16: 31084888-31084888 on Build GRCh38
rs752725867 Chr.16: 31085175-31085180 on Build GRCh38
rs766060256 Chr.16: 31084919-31084920 on Build GRCh38
rs746863050 Chr.16: 31085071-31085071 on Build GRCh38
rs377232515 Chr.16: 31085022-31085022 on Build GRCh38
rs754169438 Chr.16: 31085143-31085143 on Build GRCh38
rs991471159 Chr.16: 31084988-31084988 on Build GRCh38
rs1044646447 Chr.16: 31085124-31085124 on Build GRCh38
rs778137757 Chr.16: 31084915-31084915 on Build GRCh38
rs571044281 Chr.16: 31084798-31084798 on Build GRCh38
rs188342896 Chr.16: 31085140-31085140 on Build GRCh38
rs7199949 Chr.16: 31084843-31084843 on Build GRCh38
rs752530972 Chr.16: 31084841-31084841 on Build GRCh38
rs767525494 Chr.16: 31085130-31085133 on Build GRCh38
rs754561666 Chr.16: 31085067-31085068 on Build GRCh38
rs750013125 Chr.16: 31084866-31084866 on Build GRCh38
rs770029482 Chr.16: 31084795-31084795 on Build GRCh38
rs572401297 Chr.16: 31084979-31084979 on Build GRCh38
rs371720280 Chr.16: 31085163-31085163 on Build GRCh38

View Full Product Details