Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 48083769-48084276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC12
Entrez Gene ID
94160
Gene Name
ATP binding cassette subfamily C member 12
Gene Aliases
MRP9
UniGene
Hs.410111
Gene Chromosome Location
Chr.16: 48082973-48146770 on Build GRCh38
Transcripts
NM_033226.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
231683 c.4006G>A p.D1336N Chr.16: 48083789-48083789 on Build GRCh38
1172333 c.3976G>T p.V1326F Chr.16: 48083926-48083926 on Build GRCh38
7332140 c.3897C>G p.I1299M Chr.16: 48084005-48084005 on Build GRCh38
5456545 c.3994G>T p.V1332L Chr.16: 48083801-48083801 on Build GRCh38
2832838 c.3886C>G p.Q1296E Chr.16: 48084016-48084016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746612189 Chr.16: 48084054-48084054 on Build GRCh38
rs372109104 Chr.16: 48084071-48084071 on Build GRCh38
rs1032529073 Chr.16: 48084160-48084160 on Build GRCh38
rs770490894 Chr.16: 48084068-48084068 on Build GRCh38
rs778075310 Chr.16: 48083904-48083904 on Build GRCh38
rs755451106 Chr.16: 48084121-48084121 on Build GRCh38
rs192064700 Chr.16: 48083798-48083798 on Build GRCh38
rs779629498 Chr.16: 48084023-48084023 on Build GRCh38
rs369267834 Chr.16: 48084090-48084090 on Build GRCh38
rs761090320 Chr.16: 48083930-48083931 on Build GRCh38
rs540503825 Chr.16: 48083799-48083799 on Build GRCh38
rs753807498 Chr.16: 48083886-48083886 on Build GRCh38
rs371177642 Chr.16: 48084106-48084106 on Build GRCh38
rs771541274 Chr.16: 48084049-48084049 on Build GRCh38
rs760542016 Chr.16: 48083872-48083872 on Build GRCh38
rs759479096 Chr.16: 48083981-48083981 on Build GRCh38
rs776303012 Chr.16: 48083969-48083969 on Build GRCh38
rs758527358 Chr.16: 48083995-48083995 on Build GRCh38
rs548043231 Chr.16: 48084203-48084203 on Build GRCh38
rs4785376 Chr.16: 48084125-48084125 on Build GRCh38
rs550032247 Chr.16: 48084232-48084232 on Build GRCh38
rs1020744477 Chr.16: 48083836-48083836 on Build GRCh38
rs750114924 Chr.16: 48083896-48083896 on Build GRCh38
rs776446335 Chr.16: 48084069-48084069 on Build GRCh38
rs971641862 Chr.16: 48084244-48084244 on Build GRCh38
rs1028318850 Chr.16: 48084053-48084053 on Build GRCh38
rs572523044 Chr.16: 48084192-48084192 on Build GRCh38
rs75028443 Chr.16: 48084052-48084052 on Build GRCh38
rs200207411 Chr.16: 48083808-48083808 on Build GRCh38
rs765349844 Chr.16: 48083890-48083890 on Build GRCh38
rs746236687 Chr.16: 48084024-48084024 on Build GRCh38
rs374848483 Chr.16: 48084229-48084229 on Build GRCh38
rs756452070 Chr.16: 48084107-48084107 on Build GRCh38
rs762974239 Chr.16: 48084208-48084208 on Build GRCh38
rs775536359 Chr.16: 48084079-48084079 on Build GRCh38
rs773912608 Chr.16: 48083831-48083831 on Build GRCh38
rs374923473 Chr.16: 48084077-48084077 on Build GRCh38
rs767161412 Chr.16: 48083837-48083837 on Build GRCh38
rs201342647 Chr.16: 48083933-48083933 on Build GRCh38
rs961612785 Chr.16: 48083942-48083942 on Build GRCh38
rs772844428 Chr.16: 48083839-48083839 on Build GRCh38
rs368257926 Chr.16: 48083926-48083926 on Build GRCh38
rs751803761 Chr.16: 48083997-48083997 on Build GRCh38
rs771665597 Chr.16: 48083938-48083938 on Build GRCh38
rs752772848 Chr.16: 48083988-48083988 on Build GRCh38
rs771290364 Chr.16: 48083809-48083809 on Build GRCh38
rs368282257 Chr.16: 48083982-48083982 on Build GRCh38
rs747241166 Chr.16: 48083905-48083905 on Build GRCh38
rs144766210 Chr.16: 48083996-48083996 on Build GRCh38
rs757480312 Chr.16: 48084013-48084013 on Build GRCh38
rs773783948 Chr.16: 48083927-48083927 on Build GRCh38
rs781478647 Chr.16: 48083913-48083913 on Build GRCh38
rs768153280 Chr.16: 48083823-48083823 on Build GRCh38
rs147483326 Chr.16: 48084032-48084032 on Build GRCh38
rs763052571 Chr.16: 48084081-48084081 on Build GRCh38
rs766181366 Chr.16: 48083968-48083968 on Build GRCh38
rs747689431 Chr.16: 48083934-48083934 on Build GRCh38
rs141979519 Chr.16: 48083802-48083802 on Build GRCh38
rs749846113 Chr.16: 48084043-48084043 on Build GRCh38
rs757612353 Chr.16: 48083909-48083909 on Build GRCh38
rs140699394 Chr.16: 48083961-48083961 on Build GRCh38
rs752824292 Chr.16: 48083895-48083895 on Build GRCh38
rs200251351 Chr.16: 48084050-48084050 on Build GRCh38
rs776536714 Chr.16: 48084141-48084141 on Build GRCh38
rs779199856 Chr.16: 48083797-48083797 on Build GRCh38
rs762029649 Chr.16: 48084105-48084105 on Build GRCh38
rs11864570 Chr.16: 48084131-48084131 on Build GRCh38
rs764222163 Chr.16: 48084083-48084083 on Build GRCh38
rs183611143 Chr.16: 48083834-48083834 on Build GRCh38
rs746404997 Chr.16: 48083816-48083816 on Build GRCh38
rs777171694 Chr.16: 48083815-48083815 on Build GRCh38
rs959137799 Chr.16: 48084193-48084193 on Build GRCh38
rs780564797 Chr.16: 48084038-48084038 on Build GRCh38
rs780439957 Chr.16: 48084108-48084108 on Build GRCh38
rs377501719 Chr.16: 48083801-48083801 on Build GRCh38
rs1012941207 Chr.16: 48083833-48083833 on Build GRCh38
rs766139127 Chr.16: 48083877-48083877 on Build GRCh38
rs754327170 Chr.16: 48084112-48084112 on Build GRCh38
rs1000586678 Chr.16: 48084149-48084149 on Build GRCh38
rs144810262 Chr.16: 48084045-48084045 on Build GRCh38
rs147250788 Chr.16: 48083939-48083939 on Build GRCh38
rs755022479 Chr.16: 48083889-48083889 on Build GRCh38

View Full Product Details