Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 48547928-48548437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
N4BP1
Entrez Gene ID
9683
Gene Name
NEDD4 binding protein 1
Gene Aliases
-
UniGene
Hs.511839
Gene Chromosome Location
Chr.16: 48538726-48610214 on Build GRCh38
Transcripts
NM_153029.3 XM_011523482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7149867 c.2318G>A p.R773K Chr.16: 48548055-48548055 on Build GRCh38
971046 c.2125C>A p.L709I Chr.16: 48548107-48548107 on Build GRCh38
6372916 c.2150G>T p.G717V Chr.16: 48548082-48548082 on Build GRCh38
5118298 c.2209G>T p.E737* Chr.16: 48548023-48548023 on Build GRCh38
5029721 c.2259-2A>G p.? Chr.16: 48548116-48548116 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761398251 Chr.16: 48548235-48548235 on Build GRCh38
rs779085346 Chr.16: 48547987-48547987 on Build GRCh38
rs945566707 Chr.16: 48548266-48548266 on Build GRCh38
rs373455317 Chr.16: 48548108-48548108 on Build GRCh38
rs567675682 Chr.16: 48548121-48548121 on Build GRCh38
rs1002133118 Chr.16: 48548384-48548384 on Build GRCh38
rs746240776 Chr.16: 48548136-48548136 on Build GRCh38
rs773493128 Chr.16: 48548009-48548009 on Build GRCh38
rs750158798 Chr.16: 48548093-48548093 on Build GRCh38
rs954153101 Chr.16: 48548237-48548237 on Build GRCh38
rs776943277 Chr.16: 48548134-48548134 on Build GRCh38
rs943637868 Chr.16: 48548201-48548201 on Build GRCh38
rs1054148655 Chr.16: 48548379-48548379 on Build GRCh38
rs199745907 Chr.16: 48548003-48548003 on Build GRCh38
rs371515218 Chr.16: 48548096-48548096 on Build GRCh38
rs894136487 Chr.16: 48548396-48548396 on Build GRCh38
rs149338916 Chr.16: 48548074-48548074 on Build GRCh38
rs919324936 Chr.16: 48548117-48548117 on Build GRCh38
rs773787911 Chr.16: 48548058-48548058 on Build GRCh38
rs377240954 Chr.16: 48548221-48548221 on Build GRCh38
rs144195793 Chr.16: 48548086-48548086 on Build GRCh38
rs574931457 Chr.16: 48548142-48548142 on Build GRCh38
rs764158091 Chr.16: 48548179-48548179 on Build GRCh38
rs748177634 Chr.16: 48548110-48548110 on Build GRCh38
rs753705797 Chr.16: 48548102-48548102 on Build GRCh38
rs777850917 Chr.16: 48548125-48548125 on Build GRCh38
rs747291756 Chr.16: 48548129-48548129 on Build GRCh38
rs766244485 Chr.16: 48548097-48548097 on Build GRCh38
rs765494713 Chr.16: 48547990-48547993 on Build GRCh38
rs765768037 Chr.16: 48548037-48548037 on Build GRCh38
rs377563346 Chr.16: 48548131-48548131 on Build GRCh38
rs750109938 Chr.16: 48548128-48548129 on Build GRCh38
rs978629923 Chr.16: 48548286-48548286 on Build GRCh38
rs755085682 Chr.16: 48547969-48547969 on Build GRCh38
rs747318507 Chr.16: 48548011-48548011 on Build GRCh38
rs749279797 Chr.16: 48547968-48547968 on Build GRCh38
rs1028279094 Chr.16: 48548228-48548228 on Build GRCh38
rs762276743 Chr.16: 48547983-48547988 on Build GRCh38
rs775483636 Chr.16: 48547992-48547992 on Build GRCh38
rs878956621 Chr.16: 48548370-48548370 on Build GRCh38
rs1021958487 Chr.16: 48548375-48548375 on Build GRCh38
rs912623269 Chr.16: 48548243-48548243 on Build GRCh38
rs192351951 Chr.16: 48548189-48548189 on Build GRCh38
rs948342098 Chr.16: 48548402-48548402 on Build GRCh38
rs1033565420 Chr.16: 48548412-48548412 on Build GRCh38
rs540840627 Chr.16: 48548160-48548160 on Build GRCh38
rs777108973 Chr.16: 48548174-48548174 on Build GRCh38
rs754926247 Chr.16: 48548107-48548107 on Build GRCh38
rs936914983 Chr.16: 48548323-48548323 on Build GRCh38
rs986859751 Chr.16: 48548240-48548240 on Build GRCh38
rs776035895 Chr.16: 48548151-48548151 on Build GRCh38
rs776970232 Chr.16: 48548017-48548017 on Build GRCh38
rs1009680299 Chr.16: 48548332-48548332 on Build GRCh38
rs755995018 Chr.16: 48547962-48547962 on Build GRCh38
rs757944683 Chr.16: 48548150-48548150 on Build GRCh38
rs759991200 Chr.16: 48548020-48548020 on Build GRCh38
rs189199216 Chr.16: 48547961-48547961 on Build GRCh38
rs369775885 Chr.16: 48548304-48548304 on Build GRCh38
rs760656911 Chr.16: 48548043-48548043 on Build GRCh38
rs771390924 Chr.16: 48548013-48548013 on Build GRCh38
rs779852701 Chr.16: 48547963-48547963 on Build GRCh38

View Full Product Details