Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 72118958-72119482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PMFBP1
Entrez Gene ID
83449
Gene Name
polyamine modulated factor 1 binding protein 1
Gene Aliases
-
UniGene
Hs.714939
Gene Chromosome Location
Chr.16: 72116696-72173908 on Build GRCh38
Transcripts
NM_001160213.1 NM_031293.2 XM_011523357.2 XM_011523358.2 XM_011523360.2 XM_011523361.2 XM_011523362.2

SNP Detail

SNP ID Chromosome Location
rs570577865 Chr.16: 72119036-72119036 on Build GRCh38
rs7186117 Chr.16: 72119336-72119336 on Build GRCh38
rs192109619 Chr.16: 72119074-72119074 on Build GRCh38
rs1018719366 Chr.16: 72119068-72119068 on Build GRCh38
rs4788459 Chr.16: 72119226-72119226 on Build GRCh38
rs574593388 Chr.16: 72119435-72119435 on Build GRCh38
rs149230267 Chr.16: 72119305-72119305 on Build GRCh38
rs541864193 Chr.16: 72119427-72119427 on Build GRCh38
rs150044295 Chr.16: 72119052-72119052 on Build GRCh38
rs757879046 Chr.16: 72119375-72119375 on Build GRCh38
rs956739268 Chr.16: 72119001-72119001 on Build GRCh38
rs958746492 Chr.16: 72119112-72119112 on Build GRCh38
rs768103095 Chr.16: 72119404-72119404 on Build GRCh38
rs7184169 Chr.16: 72119034-72119034 on Build GRCh38
rs879766740 Chr.16: 72119450-72119450 on Build GRCh38
rs537638367 Chr.16: 72119037-72119037 on Build GRCh38
rs554513274 Chr.16: 72119082-72119082 on Build GRCh38
rs749960431 Chr.16: 72119345-72119345 on Build GRCh38
rs1047881893 Chr.16: 72119018-72119018 on Build GRCh38
rs375093491 Chr.16: 72119376-72119376 on Build GRCh38
rs373084763 Chr.16: 72119398-72119398 on Build GRCh38
rs755541885 Chr.16: 72119353-72119353 on Build GRCh38
rs868199857 Chr.16: 72119084-72119084 on Build GRCh38
rs774780565 Chr.16: 72119401-72119401 on Build GRCh38
rs899395566 Chr.16: 72118997-72118997 on Build GRCh38
rs370319198 Chr.16: 72119342-72119342 on Build GRCh38
rs560056177 Chr.16: 72119428-72119428 on Build GRCh38
rs546122660 Chr.16: 72119441-72119441 on Build GRCh38
rs967985376 Chr.16: 72119085-72119085 on Build GRCh38
rs745990549 Chr.16: 72119385-72119385 on Build GRCh38
rs886614716 Chr.16: 72119029-72119029 on Build GRCh38
rs777521636 Chr.16: 72119381-72119381 on Build GRCh38
rs968124153 Chr.16: 72119454-72119454 on Build GRCh38
rs770686702 Chr.16: 72119149-72119149 on Build GRCh38
rs760840596 Chr.16: 72119351-72119351 on Build GRCh38
rs112577633 Chr.16: 72119227-72119227 on Build GRCh38
rs199921562 Chr.16: 72119389-72119389 on Build GRCh38
rs1019406423 Chr.16: 72118987-72118987 on Build GRCh38
rs1022248738 Chr.16: 72119053-72119053 on Build GRCh38
rs369966848 Chr.16: 72119362-72119362 on Build GRCh38
rs770160834 Chr.16: 72119304-72119304 on Build GRCh38
rs775904301 Chr.16: 72119390-72119390 on Build GRCh38
rs754375802 Chr.16: 72119370-72119370 on Build GRCh38
rs1036053908 Chr.16: 72119242-72119242 on Build GRCh38
rs920245769 Chr.16: 72119256-72119256 on Build GRCh38
rs886578740 Chr.16: 72119354-72119354 on Build GRCh38
rs568992312 Chr.16: 72119071-72119071 on Build GRCh38
rs377435824 Chr.16: 72119400-72119400 on Build GRCh38
rs767841982 Chr.16: 72119343-72119343 on Build GRCh38
rs978424271 Chr.16: 72119098-72119098 on Build GRCh38
rs35323244 Chr.16: 72118982-72118983 on Build GRCh38
rs562600266 Chr.16: 72119330-72119330 on Build GRCh38
rs1044399866 Chr.16: 72119240-72119240 on Build GRCh38
rs754826221 Chr.16: 72119371-72119371 on Build GRCh38
rs139345862 Chr.16: 72118999-72118999 on Build GRCh38
rs527548297 Chr.16: 72119030-72119030 on Build GRCh38
rs573473805 Chr.16: 72119139-72119139 on Build GRCh38
rs776778654 Chr.16: 72119296-72119296 on Build GRCh38
rs1021930593 Chr.16: 72119438-72119438 on Build GRCh38
rs544806021 Chr.16: 72119306-72119306 on Build GRCh38
rs1040537490 Chr.16: 72119356-72119356 on Build GRCh38
rs927169597 Chr.16: 72119097-72119097 on Build GRCh38
rs1044818434 Chr.16: 72118989-72118989 on Build GRCh38
rs764648660 Chr.16: 72119359-72119359 on Build GRCh38
rs923755745 Chr.16: 72119122-72119122 on Build GRCh38
rs184474711 Chr.16: 72119303-72119303 on Build GRCh38
rs767690169 Chr.16: 72119317-72119317 on Build GRCh38
rs992842884 Chr.16: 72119175-72119175 on Build GRCh38
rs762024048 Chr.16: 72119403-72119403 on Build GRCh38

View Full Product Details