Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 74887344-74887839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR59
Entrez Gene ID
79726
Gene Name
WD repeat domain 59
Gene Aliases
FP977
UniGene
Hs.280951
Gene Chromosome Location
Chr.16: 74873568-74985146 on Build GRCh38
Transcripts
NM_030581.3 XM_005256146.3 XM_011523331.2 XM_011523332.2 XM_017023667.1 XM_017023668.1 XM_017023669.1 XM_017023670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7127213 c.2386G>A p.D796N Chr.16: 74887716-74887716 on Build GRCh38
6145515 c.2401A>T p.T801S Chr.16: 74887701-74887701 on Build GRCh38
6282511 c.2390C>A p.P797Q Chr.16: 74887712-74887712 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746381544 Chr.16: 74887520-74887520 on Build GRCh38
rs931586347 Chr.16: 74887420-74887420 on Build GRCh38
rs748296150 Chr.16: 74887654-74887654 on Build GRCh38
rs1028051542 Chr.16: 74887746-74887746 on Build GRCh38
rs2288042 Chr.16: 74887495-74887495 on Build GRCh38
rs948081585 Chr.16: 74887403-74887403 on Build GRCh38
rs551943845 Chr.16: 74887396-74887396 on Build GRCh38
rs9940014 Chr.16: 74887763-74887763 on Build GRCh38
rs200583394 Chr.16: 74887664-74887664 on Build GRCh38
rs78606888 Chr.16: 74887689-74887689 on Build GRCh38
rs551217179 Chr.16: 74887414-74887414 on Build GRCh38
rs986500870 Chr.16: 74887523-74887523 on Build GRCh38
rs934935727 Chr.16: 74887576-74887576 on Build GRCh38
rs377226670 Chr.16: 74887657-74887657 on Build GRCh38
rs867940701 Chr.16: 74887471-74887471 on Build GRCh38
rs200906442 Chr.16: 74887799-74887799 on Build GRCh38
rs1014477501 Chr.16: 74887742-74887742 on Build GRCh38
rs947467511 Chr.16: 74887633-74887633 on Build GRCh38
rs761594850 Chr.16: 74887776-74887776 on Build GRCh38
rs115786808 Chr.16: 74887626-74887626 on Build GRCh38
rs2288041 Chr.16: 74887548-74887548 on Build GRCh38
rs142406901 Chr.16: 74887503-74887503 on Build GRCh38
rs749950891 Chr.16: 74887722-74887722 on Build GRCh38
rs200203554 Chr.16: 74887665-74887665 on Build GRCh38
rs370985980 Chr.16: 74887790-74887790 on Build GRCh38
rs772511361 Chr.16: 74887521-74887521 on Build GRCh38
rs550723863 Chr.16: 74887700-74887700 on Build GRCh38
rs1035374946 Chr.16: 74887464-74887464 on Build GRCh38
rs566721028 Chr.16: 74887768-74887768 on Build GRCh38
rs934800664 Chr.16: 74887401-74887401 on Build GRCh38
rs758454602 Chr.16: 74887707-74887707 on Build GRCh38
rs148419261 Chr.16: 74887708-74887708 on Build GRCh38
rs148763098 Chr.16: 74887443-74887443 on Build GRCh38
rs1047188624 Chr.16: 74887439-74887439 on Build GRCh38
rs573831821 Chr.16: 74887630-74887630 on Build GRCh38
rs779073434 Chr.16: 74887646-74887646 on Build GRCh38
rs781524824 Chr.16: 74887720-74887720 on Build GRCh38
rs980717810 Chr.16: 74887553-74887553 on Build GRCh38
rs919930967 Chr.16: 74887564-74887564 on Build GRCh38
rs776861493 Chr.16: 74887795-74887795 on Build GRCh38
rs769494495 Chr.16: 74887675-74887675 on Build GRCh38
rs1019279134 Chr.16: 74887777-74887777 on Build GRCh38
rs144551057 Chr.16: 74887696-74887696 on Build GRCh38
rs76119554 Chr.16: 74887418-74887418 on Build GRCh38
rs774109792 Chr.16: 74887690-74887690 on Build GRCh38
rs932909660 Chr.16: 74887385-74887385 on Build GRCh38
rs147330178 Chr.16: 74887411-74887411 on Build GRCh38
rs1053229358 Chr.16: 74887402-74887402 on Build GRCh38
rs1051809761 Chr.16: 74887388-74887388 on Build GRCh38
rs201948681 Chr.16: 74887680-74887680 on Build GRCh38
rs140773440 Chr.16: 74887514-74887514 on Build GRCh38
rs752793279 Chr.16: 74887705-74887705 on Build GRCh38
rs908757291 Chr.16: 74887808-74887808 on Build GRCh38
rs35789099 Chr.16: 74887441-74887442 on Build GRCh38
rs1004170313 Chr.16: 74887693-74887693 on Build GRCh38
rs145527335 Chr.16: 74887670-74887670 on Build GRCh38
rs746298290 Chr.16: 74887732-74887732 on Build GRCh38
rs773914059 Chr.16: 74887775-74887775 on Build GRCh38
rs772190403 Chr.16: 74887786-74887786 on Build GRCh38
rs918032759 Chr.16: 74887542-74887542 on Build GRCh38
rs747375393 Chr.16: 74887659-74887659 on Build GRCh38
rs61733726 Chr.16: 74887695-74887695 on Build GRCh38
rs760585409 Chr.16: 74887793-74887793 on Build GRCh38
rs751945903 Chr.16: 74887714-74887714 on Build GRCh38
rs1011620918 Chr.16: 74887413-74887413 on Build GRCh38
rs751720003 Chr.16: 74887807-74887807 on Build GRCh38
rs774980727 Chr.16: 74887676-74887676 on Build GRCh38
rs764023778 Chr.16: 74887803-74887803 on Build GRCh38
rs748754122 Chr.16: 74887446-74887446 on Build GRCh38
rs530312789 Chr.16: 74887755-74887755 on Build GRCh38
rs369043615 Chr.16: 74887717-74887717 on Build GRCh38
rs181642992 Chr.16: 74887513-74887513 on Build GRCh38
rs536829615 Chr.16: 74887480-74887480 on Build GRCh38
rs765621304 Chr.16: 74887702-74887702 on Build GRCh38
rs752495536 Chr.16: 74887399-74887399 on Build GRCh38
rs931548886 Chr.16: 74887591-74887591 on Build GRCh38
rs763243822 Chr.16: 74887800-74887800 on Build GRCh38
rs962449096 Chr.16: 74887505-74887505 on Build GRCh38
rs143321761 Chr.16: 74887595-74887595 on Build GRCh38
rs960182813 Chr.16: 74887519-74887519 on Build GRCh38
rs571663711 Chr.16: 74887408-74887408 on Build GRCh38
rs140242354 Chr.16: 74887656-74887656 on Build GRCh38
rs1022773343 Chr.16: 74887753-74887753 on Build GRCh38
rs78189903 Chr.16: 74887506-74887506 on Build GRCh38

View Full Product Details