Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75530449-75530957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST5
Entrez Gene ID
23563
Gene Name
carbohydrate sulfotransferase 5
Gene Aliases
I-GlcNAc-6-ST, I-GlcNAc6ST, glcNAc6ST-3, gn6st-3, hIGn6ST
UniGene
Hs.745424
Gene Chromosome Location
Chr.16: 75528530-75535170 on Build GRCh38
Transcripts
NM_024533.4

SNP Detail

SNP ID Chromosome Location
rs754246994 Chr.16: 75530825-75530825 on Build GRCh38
rs116186243 Chr.16: 75530736-75530736 on Build GRCh38
rs144885830 Chr.16: 75530519-75530519 on Build GRCh38
rs893291380 Chr.16: 75530495-75530495 on Build GRCh38
rs533198503 Chr.16: 75530528-75530528 on Build GRCh38
rs993337909 Chr.16: 75530868-75530869 on Build GRCh38
rs1008670856 Chr.16: 75530932-75530932 on Build GRCh38
rs565740114 Chr.16: 75530643-75530643 on Build GRCh38
rs1041884411 Chr.16: 75530536-75530536 on Build GRCh38
rs932688943 Chr.16: 75530604-75530604 on Build GRCh38
rs1002631865 Chr.16: 75530527-75530527 on Build GRCh38
rs542656962 Chr.16: 75530492-75530492 on Build GRCh38
rs376829948 Chr.16: 75530735-75530735 on Build GRCh38
rs921795378 Chr.16: 75530599-75530599 on Build GRCh38
rs528834616 Chr.16: 75530581-75530581 on Build GRCh38
rs947565338 Chr.16: 75530532-75530532 on Build GRCh38
rs551096343 Chr.16: 75530700-75530700 on Build GRCh38
rs974579239 Chr.16: 75530593-75530593 on Build GRCh38
rs1026071958 Chr.16: 75530649-75530649 on Build GRCh38
rs977593891 Chr.16: 75530761-75530761 on Build GRCh38
rs141745551 Chr.16: 75530777-75530777 on Build GRCh38
rs987272311 Chr.16: 75530845-75530845 on Build GRCh38
rs886084902 Chr.16: 75530766-75530766 on Build GRCh38
rs1016621735 Chr.16: 75530628-75530628 on Build GRCh38
rs1017793349 Chr.16: 75530934-75530934 on Build GRCh38
rs954580394 Chr.16: 75530844-75530844 on Build GRCh38
rs755599443 Chr.16: 75530638-75530638 on Build GRCh38
rs1012907700 Chr.16: 75530606-75530606 on Build GRCh38
rs543645677 Chr.16: 75530936-75530936 on Build GRCh38
rs986920857 Chr.16: 75530619-75530619 on Build GRCh38
rs563482481 Chr.16: 75530546-75530546 on Build GRCh38
rs879077077 Chr.16: 75530917-75530917 on Build GRCh38
rs996051399 Chr.16: 75530835-75530835 on Build GRCh38
rs914860378 Chr.16: 75530669-75530669 on Build GRCh38
rs988777318 Chr.16: 75530881-75530881 on Build GRCh38
rs76223162 Chr.16: 75530661-75530661 on Build GRCh38
rs1044546707 Chr.16: 75530707-75530707 on Build GRCh38
rs772144527 Chr.16: 75530839-75530839 on Build GRCh38
rs112749155 Chr.16: 75530500-75530500 on Build GRCh38
rs778311351 Chr.16: 75530646-75530646 on Build GRCh38
rs1056913401 Chr.16: 75530749-75530749 on Build GRCh38
rs1002920034 Chr.16: 75530566-75530566 on Build GRCh38
rs370729745 Chr.16: 75530906-75530906 on Build GRCh38
rs746776710 Chr.16: 75530740-75530740 on Build GRCh38
rs1016157971 Chr.16: 75530563-75530563 on Build GRCh38
rs1044666629 Chr.16: 75530916-75530916 on Build GRCh38
rs147957433 Chr.16: 75530705-75530705 on Build GRCh38
rs987218536 Chr.16: 75530678-75530678 on Build GRCh38
rs894543079 Chr.16: 75530833-75530833 on Build GRCh38
rs567872560 Chr.16: 75530776-75530776 on Build GRCh38
rs1057235991 Chr.16: 75530592-75530592 on Build GRCh38
rs116646060 Chr.16: 75530858-75530858 on Build GRCh38
rs971855000 Chr.16: 75530481-75530481 on Build GRCh38
rs188918599 Chr.16: 75530814-75530814 on Build GRCh38
rs573274088 Chr.16: 75530501-75530501 on Build GRCh38
rs2550915 Chr.16: 75530770-75530770 on Build GRCh38
rs75581112 Chr.16: 75530907-75530907 on Build GRCh38
rs567075099 Chr.16: 75530864-75530864 on Build GRCh38
rs541200762 Chr.16: 75530502-75530502 on Build GRCh38
rs989141288 Chr.16: 75530739-75530739 on Build GRCh38
rs575489467 Chr.16: 75530741-75530741 on Build GRCh38
rs74780301 Chr.16: 75530631-75530631 on Build GRCh38
rs1016125258 Chr.16: 75530771-75530771 on Build GRCh38
rs1053363308 Chr.16: 75530667-75530667 on Build GRCh38
rs147098256 Chr.16: 75530879-75530879 on Build GRCh38
rs555226959 Chr.16: 75530544-75530544 on Build GRCh38
rs371850857 Chr.16: 75530908-75530909 on Build GRCh38
rs557027402 Chr.16: 75530886-75530886 on Build GRCh38
rs943045733 Chr.16: 75530513-75530513 on Build GRCh38
rs747130973 Chr.16: 75530922-75530922 on Build GRCh38
rs181562007 Chr.16: 75530871-75530871 on Build GRCh38
rs1037680559 Chr.16: 75530846-75530846 on Build GRCh38

View Full Product Details