Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 79597638-79598142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAF
Entrez Gene ID
4094
Gene Name
MAF bZIP transcription factor
Gene Aliases
AYGRP, CCA4, CTRCT21, c-MAF
UniGene
Hs.134859
Gene Chromosome Location
Chr.16: 79202624-79600725 on Build GRCh38
Transcripts
NM_001031804.2 NM_005360.4 XM_017023233.1 XM_017023234.1 XM_017023235.1

SNP Detail

SNP ID Chromosome Location
rs1055278187 Chr.16: 79597764-79597764 on Build GRCh38
rs1802313 Chr.16: 79598091-79598091 on Build GRCh38
rs941213314 Chr.16: 79597901-79597901 on Build GRCh38
rs1042730603 Chr.16: 79597722-79597722 on Build GRCh38
rs558549327 Chr.16: 79597898-79597898 on Build GRCh38
rs1046729 Chr.16: 79598018-79598018 on Build GRCh38
rs978451085 Chr.16: 79598006-79598006 on Build GRCh38
rs967161332 Chr.16: 79598002-79598002 on Build GRCh38
rs978480751 Chr.16: 79597732-79597732 on Build GRCh38
rs1018186035 Chr.16: 79597970-79597970 on Build GRCh38
rs538954870 Chr.16: 79597887-79597887 on Build GRCh38
rs747825948 Chr.16: 79597842-79597842 on Build GRCh38
rs929400717 Chr.16: 79597989-79597989 on Build GRCh38
rs965622239 Chr.16: 79597990-79597990 on Build GRCh38
rs748148134 Chr.16: 79597957-79597957 on Build GRCh38
rs67402322 Chr.16: 79597753-79597753 on Build GRCh38
rs573473952 Chr.16: 79597823-79597823 on Build GRCh38
rs575283169 Chr.16: 79597740-79597740 on Build GRCh38
rs34724349 Chr.16: 79598049-79598050 on Build GRCh38
rs529493684 Chr.16: 79597951-79597951 on Build GRCh38
rs878887549 Chr.16: 79597801-79597801 on Build GRCh38
rs1026748226 Chr.16: 79597661-79597661 on Build GRCh38
rs114706140 Chr.16: 79597658-79597658 on Build GRCh38
rs200380882 Chr.16: 79597800-79597800 on Build GRCh38
rs1013743760 Chr.16: 79597820-79597820 on Build GRCh38
rs200762268 Chr.16: 79597812-79597812 on Build GRCh38
rs397720736 Chr.16: 79598026-79598027 on Build GRCh38
rs936986908 Chr.16: 79598020-79598020 on Build GRCh38
rs397855831 Chr.16: 79597811-79597812 on Build GRCh38
rs1047685173 Chr.16: 79598000-79598000 on Build GRCh38
rs34194845 Chr.16: 79598019-79598020 on Build GRCh38
rs1006831500 Chr.16: 79597965-79597965 on Build GRCh38
rs952662633 Chr.16: 79597925-79597925 on Build GRCh38
rs34352423 Chr.16: 79597800-79597801 on Build GRCh38
rs543711042 Chr.16: 79597718-79597718 on Build GRCh38
rs970236532 Chr.16: 79597731-79597731 on Build GRCh38
rs1026996353 Chr.16: 79597913-79597919 on Build GRCh38
rs916825150 Chr.16: 79598058-79598058 on Build GRCh38
rs1014502383 Chr.16: 79598084-79598084 on Build GRCh38
rs530572600 Chr.16: 79598111-79598111 on Build GRCh38
rs200105851 Chr.16: 79597751-79597751 on Build GRCh38
rs1032744441 Chr.16: 79597736-79597736 on Build GRCh38
rs922278497 Chr.16: 79597768-79597768 on Build GRCh38
rs936936117 Chr.16: 79597753-79597753 on Build GRCh38
rs535589329 Chr.16: 79597696-79597696 on Build GRCh38
rs185063462 Chr.16: 79597811-79597811 on Build GRCh38
rs1014925091 Chr.16: 79597689-79597689 on Build GRCh38
rs1046563134 Chr.16: 79598101-79598102 on Build GRCh38
rs1046953115 Chr.16: 79597828-79597828 on Build GRCh38
rs201847885 Chr.16: 79597809-79597810 on Build GRCh38
rs933883263 Chr.16: 79597882-79597882 on Build GRCh38
rs372048932 Chr.16: 79598019-79598019 on Build GRCh38
rs895339228 Chr.16: 79597767-79597767 on Build GRCh38
rs1055335300 Chr.16: 79598021-79598021 on Build GRCh38
rs200906263 Chr.16: 79597761-79597761 on Build GRCh38
rs994622413 Chr.16: 79597912-79597913 on Build GRCh38
rs865792469 Chr.16: 79598081-79598081 on Build GRCh38
rs949625502 Chr.16: 79598061-79598061 on Build GRCh38
rs1037776362 Chr.16: 79597923-79597923 on Build GRCh38
rs1023214993 Chr.16: 79598089-79598089 on Build GRCh38
rs899220021 Chr.16: 79597933-79597933 on Build GRCh38
rs180693486 Chr.16: 79597855-79597855 on Build GRCh38
rs567894712 Chr.16: 79597672-79597672 on Build GRCh38
rs912972520 Chr.16: 79597699-79597699 on Build GRCh38
rs545882302 Chr.16: 79597759-79597759 on Build GRCh38
rs189125015 Chr.16: 79597851-79597851 on Build GRCh38
rs897282908 Chr.16: 79598102-79598102 on Build GRCh38
rs988464810 Chr.16: 79597741-79597741 on Build GRCh38
rs943829949 Chr.16: 79597684-79597684 on Build GRCh38
rs976610192 Chr.16: 79597997-79597997 on Build GRCh38
rs562169075 Chr.16: 79597747-79597757 on Build GRCh38
rs541985105 Chr.16: 79597984-79597984 on Build GRCh38
rs913017139 Chr.16: 79597999-79597999 on Build GRCh38

View Full Product Details